Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 86578892-86579384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXL1
Entrez Gene ID
2300
Gene Name
forkhead box L1
Gene Aliases
FKH6, FKHL11, FREAC7
UniGene
Hs.533830
Gene Chromosome Location
Chr.16: 86578509-86581698 on Build GRCh38
Transcripts
NM_005250.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
311271 c.277T>C p.W93R Chr.16: 86579000-86579000 on Build GRCh38
5089886 c.447C>T p.P149P Chr.16: 86579170-86579170 on Build GRCh38
3513108 c.174C>T p.I58I Chr.16: 86578897-86578897 on Build GRCh38
1252718 c.282G>T p.Q94H Chr.16: 86579005-86579005 on Build GRCh38
6666379 c.312C>T p.N104N Chr.16: 86579035-86579035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777342351 Chr.16: 86579069-86579069 on Build GRCh38
rs1036789070 Chr.16: 86579171-86579171 on Build GRCh38
rs995005160 Chr.16: 86579233-86579233 on Build GRCh38
rs778118009 Chr.16: 86578921-86578922 on Build GRCh38
rs371951781 Chr.16: 86579068-86579068 on Build GRCh38
rs984679500 Chr.16: 86579335-86579335 on Build GRCh38
rs754730388 Chr.16: 86578922-86578922 on Build GRCh38
rs751373929 Chr.16: 86579231-86579231 on Build GRCh38
rs1023990181 Chr.16: 86579293-86579293 on Build GRCh38
rs947860795 Chr.16: 86579273-86579273 on Build GRCh38
rs990013377 Chr.16: 86579328-86579328 on Build GRCh38
rs986259812 Chr.16: 86578960-86578960 on Build GRCh38
rs530798138 Chr.16: 86579341-86579341 on Build GRCh38
rs1030469738 Chr.16: 86578912-86578912 on Build GRCh38
rs796781281 Chr.16: 86579364-86579364 on Build GRCh38
rs766315027 Chr.16: 86579101-86579101 on Build GRCh38
rs374419803 Chr.16: 86579096-86579096 on Build GRCh38
rs977675943 Chr.16: 86579228-86579228 on Build GRCh38
rs763736974 Chr.16: 86579226-86579226 on Build GRCh38
rs769289807 Chr.16: 86579081-86579081 on Build GRCh38
rs756946675 Chr.16: 86579155-86579155 on Build GRCh38
rs762059430 Chr.16: 86578986-86578986 on Build GRCh38
rs1045302700 Chr.16: 86579247-86579247 on Build GRCh38
rs935083656 Chr.16: 86579074-86579074 on Build GRCh38
rs917705905 Chr.16: 86579238-86579238 on Build GRCh38
rs557103644 Chr.16: 86579199-86579199 on Build GRCh38
rs1021757346 Chr.16: 86579216-86579216 on Build GRCh38
rs773325629 Chr.16: 86579005-86579005 on Build GRCh38
rs916432655 Chr.16: 86579243-86579243 on Build GRCh38
rs887697855 Chr.16: 86579116-86579116 on Build GRCh38
rs766514423 Chr.16: 86579031-86579031 on Build GRCh38
rs542168189 Chr.16: 86579333-86579333 on Build GRCh38
rs897801258 Chr.16: 86579173-86579173 on Build GRCh38
rs924846251 Chr.16: 86579229-86579229 on Build GRCh38
rs199982325 Chr.16: 86578933-86578933 on Build GRCh38
rs1027894858 Chr.16: 86579053-86579053 on Build GRCh38
rs550925255 Chr.16: 86579343-86579343 on Build GRCh38
rs1036310261 Chr.16: 86579311-86579311 on Build GRCh38
rs1006097053 Chr.16: 86579165-86579165 on Build GRCh38
rs971513801 Chr.16: 86579298-86579298 on Build GRCh38
rs566337140 Chr.16: 86579104-86579104 on Build GRCh38
rs112864402 Chr.16: 86579266-86579266 on Build GRCh38
rs941955968 Chr.16: 86579305-86579305 on Build GRCh38
rs776561080 Chr.16: 86579202-86579202 on Build GRCh38
rs200323545 Chr.16: 86579138-86579138 on Build GRCh38
rs555070012 Chr.16: 86579163-86579163 on Build GRCh38
rs141410677 Chr.16: 86579102-86579102 on Build GRCh38
rs749832074 Chr.16: 86578961-86578961 on Build GRCh38
rs577050474 Chr.16: 86579203-86579203 on Build GRCh38
rs969395972 Chr.16: 86579220-86579220 on Build GRCh38
rs537362211 Chr.16: 86579181-86579181 on Build GRCh38
rs769091297 Chr.16: 86578962-86578962 on Build GRCh38
rs530724388 Chr.16: 86578942-86578942 on Build GRCh38
rs764948768 Chr.16: 86579057-86579057 on Build GRCh38
rs944737366 Chr.16: 86579353-86579353 on Build GRCh38
rs545875290 Chr.16: 86579232-86579232 on Build GRCh38
rs767254315 Chr.16: 86579242-86579242 on Build GRCh38
rs749927112 Chr.16: 86579245-86579245 on Build GRCh38
rs772279289 Chr.16: 86579185-86579185 on Build GRCh38
rs752467864 Chr.16: 86579115-86579115 on Build GRCh38
rs746498124 Chr.16: 86579070-86579070 on Build GRCh38
rs778216107 Chr.16: 86579192-86579192 on Build GRCh38
rs771036897 Chr.16: 86579348-86579348 on Build GRCh38
rs561955120 Chr.16: 86579338-86579338 on Build GRCh38
rs749019038 Chr.16: 86579220-86579231 on Build GRCh38
rs774849120 Chr.16: 86579084-86579084 on Build GRCh38
rs201070716 Chr.16: 86579072-86579072 on Build GRCh38
rs760840650 Chr.16: 86579131-86579131 on Build GRCh38
rs573260354 Chr.16: 86579332-86579332 on Build GRCh38
rs746122400 Chr.16: 86579363-86579363 on Build GRCh38
rs113108019 Chr.16: 86579038-86579038 on Build GRCh38
rs753200424 Chr.16: 86579295-86579295 on Build GRCh38
rs1012965454 Chr.16: 86579274-86579274 on Build GRCh38
rs762898236 Chr.16: 86579217-86579217 on Build GRCh38
rs750019437 Chr.16: 86579164-86579164 on Build GRCh38
rs771255715 Chr.16: 86579200-86579200 on Build GRCh38
rs957175837 Chr.16: 86579313-86579313 on Build GRCh38
rs550762225 Chr.16: 86578943-86578943 on Build GRCh38
rs769689778 Chr.16: 86579205-86579205 on Build GRCh38
rs924397043 Chr.16: 86579309-86579309 on Build GRCh38
rs763982001 Chr.16: 86579140-86579140 on Build GRCh38
rs772398110 Chr.16: 86579091-86579091 on Build GRCh38
rs760715335 Chr.16: 86579029-86579029 on Build GRCh38
rs953571485 Chr.16: 86578940-86578940 on Build GRCh38
rs753643668 Chr.16: 86578921-86578921 on Build GRCh38
rs765217572 Chr.16: 86579110-86579110 on Build GRCh38
rs749755371 Chr.16: 86579077-86579077 on Build GRCh38
rs767914376 Chr.16: 86578990-86578990 on Build GRCh38
rs751122912 Chr.16: 86579141-86579141 on Build GRCh38
rs775902167 Chr.16: 86578954-86578954 on Build GRCh38
rs755723445 Chr.16: 86579246-86579246 on Build GRCh38
rs544656837 Chr.16: 86579095-86579095 on Build GRCh38
rs754925600 Chr.16: 86579041-86579041 on Build GRCh38
rs201954519 Chr.16: 86579178-86579178 on Build GRCh38
rs781433069 Chr.16: 86579362-86579362 on Build GRCh38
rs778705560 Chr.16: 86578924-86578924 on Build GRCh38
rs775367514 Chr.16: 86579209-86579209 on Build GRCh38
rs752555977 Chr.16: 86579067-86579067 on Build GRCh38
rs546754065 Chr.16: 86579089-86579089 on Build GRCh38
rs759468418 Chr.16: 86579106-86579106 on Build GRCh38
rs947630904 Chr.16: 86579060-86579060 on Build GRCh38
rs774869568 Chr.16: 86578963-86578963 on Build GRCh38
rs374413539 Chr.16: 86579174-86579174 on Build GRCh38

View Full Product Details