Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 86581077-86581578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXL1
Entrez Gene ID
2300
Gene Name
forkhead box L1
Gene Aliases
FKH6, FKHL11, FREAC7
UniGene
Hs.533830
Gene Chromosome Location
Chr.16: 86578509-86581698 on Build GRCh38
Transcripts
NM_005250.2

SNP Detail

SNP ID Chromosome Location
rs887318763 Chr.16: 86581103-86581106 on Build GRCh38
rs981245401 Chr.16: 86581496-86581496 on Build GRCh38
rs555919840 Chr.16: 86581326-86581326 on Build GRCh38
rs750046315 Chr.16: 86581215-86581215 on Build GRCh38
rs934077711 Chr.16: 86581304-86581304 on Build GRCh38
rs866425108 Chr.16: 86581119-86581119 on Build GRCh38
rs979705300 Chr.16: 86581505-86581505 on Build GRCh38
rs544696491 Chr.16: 86581413-86581413 on Build GRCh38
rs975347392 Chr.16: 86581524-86581524 on Build GRCh38
rs1038598451 Chr.16: 86581376-86581376 on Build GRCh38
rs144212795 Chr.16: 86581495-86581495 on Build GRCh38
rs995501948 Chr.16: 86581313-86581313 on Build GRCh38
rs964267025 Chr.16: 86581412-86581412 on Build GRCh38
rs758885663 Chr.16: 86581547-86581547 on Build GRCh38
rs1047017559 Chr.16: 86581328-86581328 on Build GRCh38
rs989815237 Chr.16: 86581450-86581450 on Build GRCh38
rs779476439 Chr.16: 86581303-86581303 on Build GRCh38
rs111494988 Chr.16: 86581443-86581443 on Build GRCh38
rs529311043 Chr.16: 86581301-86581301 on Build GRCh38
rs941562301 Chr.16: 86581374-86581374 on Build GRCh38
rs543405185 Chr.16: 86581192-86581192 on Build GRCh38
rs575868671 Chr.16: 86581358-86581358 on Build GRCh38
rs182741302 Chr.16: 86581310-86581310 on Build GRCh38
rs1008358993 Chr.16: 86581128-86581128 on Build GRCh38
rs1020103803 Chr.16: 86581395-86581395 on Build GRCh38
rs1047182284 Chr.16: 86581315-86581315 on Build GRCh38
rs1008242852 Chr.16: 86581371-86581371 on Build GRCh38
rs1009313135 Chr.16: 86581432-86581432 on Build GRCh38
rs374477712 Chr.16: 86581194-86581194 on Build GRCh38
rs896204152 Chr.16: 86581423-86581423 on Build GRCh38
rs928356962 Chr.16: 86581538-86581538 on Build GRCh38
rs747563725 Chr.16: 86581528-86581529 on Build GRCh38
rs771716714 Chr.16: 86581299-86581299 on Build GRCh38
rs572113358 Chr.16: 86581474-86581474 on Build GRCh38
rs767056681 Chr.16: 86581173-86581173 on Build GRCh38
rs533984421 Chr.16: 86581279-86581279 on Build GRCh38
rs755757594 Chr.16: 86581276-86581276 on Build GRCh38
rs563710191 Chr.16: 86581199-86581199 on Build GRCh38
rs887195619 Chr.16: 86581360-86581360 on Build GRCh38
rs975718673 Chr.16: 86581159-86581159 on Build GRCh38
rs115763123 Chr.16: 86581105-86581105 on Build GRCh38
rs1017495115 Chr.16: 86581514-86581514 on Build GRCh38
rs922610564 Chr.16: 86581535-86581535 on Build GRCh38
rs1034753335 Chr.16: 86581153-86581153 on Build GRCh38
rs77215492 Chr.16: 86581164-86581164 on Build GRCh38
rs898405518 Chr.16: 86581309-86581309 on Build GRCh38
rs1019741345 Chr.16: 86581150-86581150 on Build GRCh38
rs374873294 Chr.16: 86581531-86581531 on Build GRCh38
rs112987356 Chr.16: 86581166-86581166 on Build GRCh38
rs536162816 Chr.16: 86581318-86581318 on Build GRCh38
rs910485263 Chr.16: 86581213-86581213 on Build GRCh38
rs780803544 Chr.16: 86581526-86581527 on Build GRCh38
rs573819854 Chr.16: 86581317-86581317 on Build GRCh38
rs191022724 Chr.16: 86581110-86581110 on Build GRCh38
rs905471550 Chr.16: 86581400-86581400 on Build GRCh38
rs965007119 Chr.16: 86581516-86581516 on Build GRCh38
rs549562290 Chr.16: 86581104-86581104 on Build GRCh38
rs1019935696 Chr.16: 86581492-86581492 on Build GRCh38
rs964658074 Chr.16: 86581151-86581151 on Build GRCh38
rs563805222 Chr.16: 86581152-86581152 on Build GRCh38
rs1032628069 Chr.16: 86581419-86581419 on Build GRCh38
rs937231230 Chr.16: 86581546-86581546 on Build GRCh38
rs540632235 Chr.16: 86581475-86581475 on Build GRCh38

View Full Product Details