Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 87302706-87303206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf95
Entrez Gene ID
100506581
Gene Name
chromosome 16 open reading frame 95
Gene Aliases
-
UniGene
Hs.729380
Gene Chromosome Location
Chr.16: 87302798-87317420 on Build GRCh38
Transcripts
NM_001195124.1 NM_001195125.1 NM_001256917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7276293 c.706T>C p.C236R Chr.16: 87303071-87303071 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs995483713 Chr.16: 87302813-87302813 on Build GRCh38
rs966961567 Chr.16: 87303090-87303090 on Build GRCh38
rs978424347 Chr.16: 87302907-87302907 on Build GRCh38
rs566762685 Chr.16: 87302936-87302936 on Build GRCh38
rs941955257 Chr.16: 87302790-87302790 on Build GRCh38
rs933263314 Chr.16: 87302981-87302981 on Build GRCh38
rs1862788 Chr.16: 87302820-87302820 on Build GRCh38
rs948787093 Chr.16: 87302949-87302949 on Build GRCh38
rs939982220 Chr.16: 87302974-87302974 on Build GRCh38
rs1037708943 Chr.16: 87302999-87302999 on Build GRCh38
rs543976189 Chr.16: 87303170-87303170 on Build GRCh38
rs746414025 Chr.16: 87303046-87303046 on Build GRCh38
rs970233260 Chr.16: 87303096-87303096 on Build GRCh38
rs1862787 Chr.16: 87302933-87302933 on Build GRCh38
rs149461208 Chr.16: 87302853-87302853 on Build GRCh38
rs1030187080 Chr.16: 87302829-87302829 on Build GRCh38
rs1036722255 Chr.16: 87302805-87302805 on Build GRCh38
rs765505629 Chr.16: 87303021-87303021 on Build GRCh38
rs925198482 Chr.16: 87302932-87302932 on Build GRCh38
rs1032301367 Chr.16: 87302892-87302892 on Build GRCh38
rs763265998 Chr.16: 87303116-87303116 on Build GRCh38
rs1007309517 Chr.16: 87303008-87303008 on Build GRCh38
rs386793825 Chr.16: 87302933-87302934 on Build GRCh38
rs1004853265 Chr.16: 87302766-87302766 on Build GRCh38
rs537966729 Chr.16: 87303002-87303002 on Build GRCh38
rs137887055 Chr.16: 87302806-87302806 on Build GRCh38
rs1051294891 Chr.16: 87302979-87302979 on Build GRCh38
rs907594293 Chr.16: 87303165-87303165 on Build GRCh38
rs183994350 Chr.16: 87303060-87303060 on Build GRCh38
rs1037035133 Chr.16: 87302976-87302976 on Build GRCh38
rs570956672 Chr.16: 87302994-87302994 on Build GRCh38
rs998903586 Chr.16: 87303023-87303023 on Build GRCh38
rs544698155 Chr.16: 87303028-87303028 on Build GRCh38
rs879524095 Chr.16: 87303079-87303079 on Build GRCh38
rs540259195 Chr.16: 87303109-87303109 on Build GRCh38
rs888470996 Chr.16: 87302765-87302765 on Build GRCh38
rs909916136 Chr.16: 87303111-87303111 on Build GRCh38
rs776964286 Chr.16: 87303042-87303042 on Build GRCh38
rs140830551 Chr.16: 87303028-87303029 on Build GRCh38
rs1021352558 Chr.16: 87303009-87303009 on Build GRCh38
rs974419095 Chr.16: 87303115-87303115 on Build GRCh38
rs534230682 Chr.16: 87302961-87302961 on Build GRCh38
rs888837959 Chr.16: 87302849-87302849 on Build GRCh38
rs564744086 Chr.16: 87303112-87303112 on Build GRCh38
rs746709278 Chr.16: 87302983-87302983 on Build GRCh38
rs1048368359 Chr.16: 87302746-87302746 on Build GRCh38
rs562527639 Chr.16: 87303179-87303179 on Build GRCh38
rs968826380 Chr.16: 87302861-87302861 on Build GRCh38
rs977124498 Chr.16: 87302865-87302865 on Build GRCh38
rs560271017 Chr.16: 87303143-87303143 on Build GRCh38
rs554092207 Chr.16: 87303066-87303066 on Build GRCh38
rs959411801 Chr.16: 87302835-87302835 on Build GRCh38
rs984208385 Chr.16: 87302960-87302960 on Build GRCh38
rs1031349295 Chr.16: 87303032-87303032 on Build GRCh38
rs556590691 Chr.16: 87303005-87303005 on Build GRCh38
rs897710884 Chr.16: 87302978-87302978 on Build GRCh38
rs571876957 Chr.16: 87302736-87302736 on Build GRCh38
rs987639921 Chr.16: 87303155-87303155 on Build GRCh38
rs909877884 Chr.16: 87302971-87302971 on Build GRCh38
rs143161296 Chr.16: 87303139-87303139 on Build GRCh38
rs1007340585 Chr.16: 87302851-87302851 on Build GRCh38
rs1024840642 Chr.16: 87303067-87303067 on Build GRCh38
rs558656172 Chr.16: 87302990-87302990 on Build GRCh38
rs536404393 Chr.16: 87302905-87302905 on Build GRCh38
rs922655350 Chr.16: 87303156-87303156 on Build GRCh38
rs572384911 Chr.16: 87303108-87303108 on Build GRCh38
rs188017570 Chr.16: 87303063-87303063 on Build GRCh38
rs563006892 Chr.16: 87303034-87303034 on Build GRCh38
rs961499930 Chr.16: 87303131-87303131 on Build GRCh38
rs921582293 Chr.16: 87303128-87303128 on Build GRCh38
rs1051196004 Chr.16: 87302739-87302739 on Build GRCh38
rs960557339 Chr.16: 87303041-87303041 on Build GRCh38
rs933223193 Chr.16: 87303178-87303178 on Build GRCh38

View Full Product Details