Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88005961-88006368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BANP
Entrez Gene ID
54971
Gene Name
BTG3 associated nuclear protein
Gene Aliases
BEND1, SMAR1, SMARBP1
UniGene
Hs.461705
Gene Chromosome Location
Chr.16: 87949216-88077318 on Build GRCh38
Transcripts
NM_001173539.1 NM_001173540.1 NM_001173541.1 NM_001173542.1 NM_001173543.1 NM_017869.3 NM_079837.2 XM_005256008.3 XM_005256009.3 XM_011523170.2 XM_011523172.2 XM_011523174.2 XM_011523175.2 XM_011523176.2 XM_011523177.2 XM_011523178.2 XM_011523179.2 XM_011523180.2 XM_011523181.2 XM_011523182.1 XM_011523183.2 XM_011523184.2 XM_011523186.2 XM_017023378.1 XM_017023379.1 XM_017023380.1 XM_017023381.1 XM_017023382.1 XM_017023383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6219303 c.363-10T>C p.? Chr.16: 88006080-88006080 on Build GRCh38
4890306 c.388C>T p.P130S Chr.16: 88006097-88006097 on Build GRCh38
1302332 c.522C>G p.H174Q Chr.16: 88006156-88006156 on Build GRCh38
3421225 c.460G>A p.D154N Chr.16: 88006187-88006187 on Build GRCh38
6609891 c.519C>T p.I173I Chr.16: 88006228-88006228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764617068 Chr.16: 88006050-88006050 on Build GRCh38
rs376115895 Chr.16: 88006047-88006047 on Build GRCh38
rs781120281 Chr.16: 88006197-88006197 on Build GRCh38
rs1042725988 Chr.16: 88005997-88005997 on Build GRCh38
rs752305818 Chr.16: 88006053-88006053 on Build GRCh38
rs374469815 Chr.16: 88006269-88006269 on Build GRCh38
rs777185086 Chr.16: 88006317-88006317 on Build GRCh38
rs1015642053 Chr.16: 88006065-88006065 on Build GRCh38
rs756491605 Chr.16: 88006057-88006057 on Build GRCh38
rs1018222772 Chr.16: 88005996-88005996 on Build GRCh38
rs372130199 Chr.16: 88006278-88006278 on Build GRCh38
rs557909608 Chr.16: 88005993-88005993 on Build GRCh38
rs139770048 Chr.16: 88006289-88006289 on Build GRCh38
rs745483519 Chr.16: 88006091-88006091 on Build GRCh38
rs750336505 Chr.16: 88006288-88006288 on Build GRCh38
rs998854359 Chr.16: 88006080-88006080 on Build GRCh38
rs565301883 Chr.16: 88006213-88006213 on Build GRCh38
rs767531732 Chr.16: 88006276-88006277 on Build GRCh38
rs143791417 Chr.16: 88006176-88006176 on Build GRCh38
rs955712609 Chr.16: 88006034-88006034 on Build GRCh38
rs752872887 Chr.16: 88006177-88006177 on Build GRCh38
rs778562002 Chr.16: 88006228-88006228 on Build GRCh38
rs779744719 Chr.16: 88006148-88006148 on Build GRCh38
rs1012727111 Chr.16: 88006043-88006043 on Build GRCh38
rs867351117 Chr.16: 88006023-88006023 on Build GRCh38
rs955910629 Chr.16: 88006308-88006311 on Build GRCh38
rs139021637 Chr.16: 88006119-88006119 on Build GRCh38
rs536848852 Chr.16: 88006322-88006322 on Build GRCh38
rs757140286 Chr.16: 88006195-88006195 on Build GRCh38
rs774861065 Chr.16: 88006102-88006102 on Build GRCh38
rs748919354 Chr.16: 88006217-88006217 on Build GRCh38
rs759885495 Chr.16: 88006171-88006171 on Build GRCh38
rs777858600 Chr.16: 88006134-88006134 on Build GRCh38
rs775227523 Chr.16: 88006152-88006152 on Build GRCh38
rs529472023 Chr.16: 88006307-88006307 on Build GRCh38
rs778845265 Chr.16: 88006078-88006078 on Build GRCh38
rs751692720 Chr.16: 88006191-88006191 on Build GRCh38
rs779973858 Chr.16: 88006297-88006297 on Build GRCh38
rs764132919 Chr.16: 88006187-88006187 on Build GRCh38
rs569769146 Chr.16: 88006016-88006016 on Build GRCh38
rs377102216 Chr.16: 88006106-88006106 on Build GRCh38
rs769483467 Chr.16: 88006151-88006151 on Build GRCh38
rs780839856 Chr.16: 88006145-88006145 on Build GRCh38
rs747628148 Chr.16: 88006229-88006229 on Build GRCh38
rs141911325 Chr.16: 88006261-88006261 on Build GRCh38
rs748293144 Chr.16: 88006082-88006082 on Build GRCh38
rs773807056 Chr.16: 88006104-88006104 on Build GRCh38
rs555222790 Chr.16: 88006036-88006036 on Build GRCh38
rs375544217 Chr.16: 88006298-88006298 on Build GRCh38
rs201658021 Chr.16: 88006058-88006058 on Build GRCh38
rs575733236 Chr.16: 88006105-88006105 on Build GRCh38
rs533062285 Chr.16: 88006162-88006162 on Build GRCh38
rs550783538 Chr.16: 88006305-88006305 on Build GRCh38
rs116714118 Chr.16: 88006090-88006090 on Build GRCh38
rs768472728 Chr.16: 88006226-88006226 on Build GRCh38
rs868302869 Chr.16: 88006012-88006012 on Build GRCh38
rs368963219 Chr.16: 88006118-88006118 on Build GRCh38
rs748066517 Chr.16: 88006342-88006342 on Build GRCh38
rs755150570 Chr.16: 88006114-88006114 on Build GRCh38
rs746913650 Chr.16: 88006135-88006135 on Build GRCh38
rs757868342 Chr.16: 88006103-88006103 on Build GRCh38
rs763529784 Chr.16: 88006306-88006307 on Build GRCh38
rs778429761 Chr.16: 88006003-88006003 on Build GRCh38
rs142706336 Chr.16: 88006046-88006046 on Build GRCh38
rs755639963 Chr.16: 88006304-88006305 on Build GRCh38
rs543231944 Chr.16: 88006156-88006156 on Build GRCh38
rs375644738 Chr.16: 88006163-88006163 on Build GRCh38
rs753967861 Chr.16: 88006064-88006064 on Build GRCh38
rs766622078 Chr.16: 88006107-88006107 on Build GRCh38
rs757264142 Chr.16: 88006138-88006138 on Build GRCh38
rs754286632 Chr.16: 88006109-88006109 on Build GRCh38
rs201182509 Chr.16: 88006306-88006306 on Build GRCh38
rs773435641 Chr.16: 88006073-88006073 on Build GRCh38
rs759454623 Chr.16: 88006106-88006107 on Build GRCh38
rs374539111 Chr.16: 88006084-88006084 on Build GRCh38
rs774276814 Chr.16: 88006101-88006102 on Build GRCh38
rs544797610 Chr.16: 88006086-88006086 on Build GRCh38
rs537289150 Chr.16: 88006025-88006025 on Build GRCh38
rs376758421 Chr.16: 88006279-88006279 on Build GRCh38
rs763095742 Chr.16: 88006072-88006073 on Build GRCh38
rs1032747149 Chr.16: 88006014-88006014 on Build GRCh38
rs766129739 Chr.16: 88006080-88006081 on Build GRCh38
rs947123316 Chr.16: 88005997-88005998 on Build GRCh38
rs147423802 Chr.16: 88006212-88006212 on Build GRCh38
rs761242006 Chr.16: 88006160-88006160 on Build GRCh38
rs752713878 Chr.16: 88006301-88006302 on Build GRCh38
rs771494472 Chr.16: 88006231-88006231 on Build GRCh38
rs920655811 Chr.16: 88006094-88006094 on Build GRCh38
rs747776068 Chr.16: 88006308-88006308 on Build GRCh38
rs771568895 Chr.16: 88006314-88006314 on Build GRCh38
rs780029068 Chr.16: 88006215-88006215 on Build GRCh38
rs185879087 Chr.16: 88006062-88006062 on Build GRCh38
rs373928466 Chr.16: 88006055-88006055 on Build GRCh38
rs370624671 Chr.16: 88006083-88006083 on Build GRCh38
rs372273341 Chr.16: 88006243-88006243 on Build GRCh38
rs769552029 Chr.16: 88006099-88006099 on Build GRCh38
rs774045333 Chr.16: 88006159-88006159 on Build GRCh38
rs767518723 Chr.16: 88006286-88006286 on Build GRCh38
rs373253543 Chr.16: 88006233-88006233 on Build GRCh38
rs748972674 Chr.16: 88006153-88006153 on Build GRCh38
rs759989063 Chr.16: 88006237-88006237 on Build GRCh38
rs528806022 Chr.16: 88006186-88006186 on Build GRCh38
rs961394504 Chr.16: 88006068-88006068 on Build GRCh38
rs149402068 Chr.16: 88006172-88006172 on Build GRCh38
rs758420829 Chr.16: 88006120-88006120 on Build GRCh38
rs367640047 Chr.16: 88006276-88006276 on Build GRCh38
rs761896974 Chr.16: 88006283-88006283 on Build GRCh38

View Full Product Details