Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88071986-88072482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BANP
Entrez Gene ID
54971
Gene Name
BTG3 associated nuclear protein
Gene Aliases
BEND1, SMAR1, SMARBP1
UniGene
Hs.461705
Gene Chromosome Location
Chr.16: 87949216-88077318 on Build GRCh38
Transcripts
NM_001173539.1 NM_001173540.1 NM_001173541.1 NM_001173542.1 NM_001173543.1 NM_017869.3 NM_079837.2 XM_005256008.3 XM_005256009.3 XM_011523170.2 XM_011523172.2 XM_011523174.2 XM_011523175.2 XM_011523176.2 XM_011523177.2 XM_011523178.2 XM_011523179.2 XM_011523180.2 XM_011523181.2 XM_011523183.2 XM_011523184.2 XM_011523186.2 XM_017023378.1 XM_017023379.1 XM_017023380.1 XM_017023381.1 XM_017023382.1 XM_017023383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4665173 c.1309C>T p.P437S Chr.16: 88072183-88072183 on Build GRCh38
6395211 c.1429G>A p.D477N Chr.16: 88072153-88072153 on Build GRCh38
3387660 c.1195-6C>T p.? Chr.16: 88072063-88072063 on Build GRCh38
7267037 c.1372G>A p.V458M Chr.16: 88072096-88072096 on Build GRCh38
4665172 c.1393C>T p.P465S Chr.16: 88072183-88072183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764645448 Chr.16: 88072242-88072242 on Build GRCh38
rs113141769 Chr.16: 88072053-88072053 on Build GRCh38
rs532009997 Chr.16: 88072451-88072451 on Build GRCh38
rs201660824 Chr.16: 88072200-88072200 on Build GRCh38
rs920094979 Chr.16: 88072353-88072353 on Build GRCh38
rs111350348 Chr.16: 88072203-88072203 on Build GRCh38
rs146466657 Chr.16: 88072113-88072113 on Build GRCh38
rs908223621 Chr.16: 88072362-88072362 on Build GRCh38
rs775793011 Chr.16: 88072135-88072135 on Build GRCh38
rs372917438 Chr.16: 88072258-88072258 on Build GRCh38
rs546551447 Chr.16: 88072229-88072229 on Build GRCh38
rs772525766 Chr.16: 88072062-88072062 on Build GRCh38
rs536831807 Chr.16: 88072275-88072275 on Build GRCh38
rs796960548 Chr.16: 88072312-88072312 on Build GRCh38
rs747551259 Chr.16: 88072121-88072121 on Build GRCh38
rs758101982 Chr.16: 88072313-88072313 on Build GRCh38
rs987133588 Chr.16: 88072399-88072399 on Build GRCh38
rs765338048 Chr.16: 88072034-88072034 on Build GRCh38
rs563007703 Chr.16: 88072125-88072125 on Build GRCh38
rs748683985 Chr.16: 88072116-88072116 on Build GRCh38
rs867560277 Chr.16: 88072181-88072181 on Build GRCh38
rs775514112 Chr.16: 88072061-88072061 on Build GRCh38
rs558710850 Chr.16: 88072092-88072092 on Build GRCh38
rs79666647 Chr.16: 88072361-88072361 on Build GRCh38
rs745549832 Chr.16: 88072051-88072051 on Build GRCh38
rs777618514 Chr.16: 88072044-88072044 on Build GRCh38
rs962924985 Chr.16: 88072376-88072376 on Build GRCh38
rs113063252 Chr.16: 88072367-88072367 on Build GRCh38
rs748678999 Chr.16: 88072055-88072055 on Build GRCh38
rs867845868 Chr.16: 88072133-88072133 on Build GRCh38
rs11864872 Chr.16: 88072360-88072360 on Build GRCh38
rs957411734 Chr.16: 88072145-88072145 on Build GRCh38
rs369246788 Chr.16: 88072115-88072115 on Build GRCh38
rs572721583 Chr.16: 88072417-88072417 on Build GRCh38
rs371819708 Chr.16: 88072199-88072199 on Build GRCh38
rs748427088 Chr.16: 88072095-88072095 on Build GRCh38
rs917527464 Chr.16: 88072218-88072218 on Build GRCh38
rs772072018 Chr.16: 88072228-88072228 on Build GRCh38
rs757627515 Chr.16: 88072320-88072320 on Build GRCh38
rs548835726 Chr.16: 88072290-88072290 on Build GRCh38
rs143051522 Chr.16: 88072152-88072152 on Build GRCh38
rs560775175 Chr.16: 88072398-88072398 on Build GRCh38
rs891866443 Chr.16: 88072356-88072356 on Build GRCh38
rs757079708 Chr.16: 88072108-88072108 on Build GRCh38
rs556378889 Chr.16: 88072381-88072381 on Build GRCh38
rs879493804 Chr.16: 88072282-88072282 on Build GRCh38
rs761512343 Chr.16: 88072327-88072327 on Build GRCh38
rs374548756 Chr.16: 88072256-88072257 on Build GRCh38
rs780098631 Chr.16: 88072204-88072204 on Build GRCh38
rs148266591 Chr.16: 88072265-88072265 on Build GRCh38
rs773777017 Chr.16: 88072063-88072063 on Build GRCh38
rs4075180 Chr.16: 88072440-88072440 on Build GRCh38
rs370027319 Chr.16: 88072424-88072424 on Build GRCh38
rs542397736 Chr.16: 88072387-88072387 on Build GRCh38
rs759289165 Chr.16: 88072235-88072235 on Build GRCh38
rs768598312 Chr.16: 88072222-88072222 on Build GRCh38
rs771260984 Chr.16: 88072122-88072122 on Build GRCh38
rs1020337513 Chr.16: 88072283-88072283 on Build GRCh38
rs139148209 Chr.16: 88072126-88072126 on Build GRCh38
rs180881982 Chr.16: 88072234-88072234 on Build GRCh38
rs761891787 Chr.16: 88072144-88072144 on Build GRCh38
rs754411970 Chr.16: 88072176-88072176 on Build GRCh38
rs1040958835 Chr.16: 88072375-88072375 on Build GRCh38
rs929057418 Chr.16: 88072389-88072389 on Build GRCh38
rs893639486 Chr.16: 88072414-88072414 on Build GRCh38
rs1031788698 Chr.16: 88072355-88072355 on Build GRCh38
rs185961232 Chr.16: 88072245-88072245 on Build GRCh38
rs760340600 Chr.16: 88072230-88072230 on Build GRCh38
rs774630779 Chr.16: 88072224-88072224 on Build GRCh38
rs8050209 Chr.16: 88072119-88072119 on Build GRCh38
rs757917104 Chr.16: 88072251-88072251 on Build GRCh38
rs766230465 Chr.16: 88072231-88072231 on Build GRCh38
rs1017582031 Chr.16: 88072368-88072368 on Build GRCh38
rs62046926 Chr.16: 88072035-88072035 on Build GRCh38
rs750844378 Chr.16: 88072329-88072329 on Build GRCh38
rs200731196 Chr.16: 88072038-88072038 on Build GRCh38
rs1041789451 Chr.16: 88072425-88072425 on Build GRCh38
rs968815464 Chr.16: 88072005-88072011 on Build GRCh38
rs567150446 Chr.16: 88072291-88072291 on Build GRCh38
rs577190967 Chr.16: 88072118-88072118 on Build GRCh38
rs767462307 Chr.16: 88072112-88072112 on Build GRCh38
rs1054051240 Chr.16: 88072402-88072402 on Build GRCh38
rs34278685 Chr.16: 88072311-88072311 on Build GRCh38
rs761838766 Chr.16: 88072227-88072227 on Build GRCh38
rs1035962089 Chr.16: 88072445-88072445 on Build GRCh38
rs769840020 Chr.16: 88072132-88072132 on Build GRCh38
rs751967561 Chr.16: 88072316-88072316 on Build GRCh38
rs369510119 Chr.16: 88072246-88072246 on Build GRCh38
rs375744686 Chr.16: 88072048-88072048 on Build GRCh38
rs534469035 Chr.16: 88072301-88072301 on Build GRCh38
rs540115391 Chr.16: 88072431-88072431 on Build GRCh38
rs997059977 Chr.16: 88072273-88072273 on Build GRCh38
rs778532846 Chr.16: 88072182-88072182 on Build GRCh38
rs886770826 Chr.16: 88072253-88072253 on Build GRCh38
rs149029966 Chr.16: 88072137-88072137 on Build GRCh38
rs574629771 Chr.16: 88072382-88072382 on Build GRCh38
rs368041267 Chr.16: 88072045-88072045 on Build GRCh38
rs536844238 Chr.16: 88072036-88072036 on Build GRCh38
rs992039507 Chr.16: 88072346-88072346 on Build GRCh38
rs760554782 Chr.16: 88072159-88072159 on Build GRCh38
rs759111016 Chr.16: 88072266-88072266 on Build GRCh38
rs765284683 Chr.16: 88072099-88072099 on Build GRCh38
rs763098274 Chr.16: 88072106-88072106 on Build GRCh38
rs918980355 Chr.16: 88072277-88072277 on Build GRCh38
rs765892010 Chr.16: 88072170-88072170 on Build GRCh38
rs758321188 Chr.16: 88072039-88072039 on Build GRCh38
rs926404645 Chr.16: 88072050-88072050 on Build GRCh38
rs764610091 Chr.16: 88072314-88072314 on Build GRCh38
rs752436661 Chr.16: 88072312-88072312 on Build GRCh38
rs763698631 Chr.16: 88072274-88072274 on Build GRCh38
rs377306642 Chr.16: 88072107-88072107 on Build GRCh38
rs775499152 Chr.16: 88072102-88072102 on Build GRCh38
rs946209528 Chr.16: 88072031-88072031 on Build GRCh38
rs147469146 Chr.16: 88072153-88072153 on Build GRCh38
rs774126415 Chr.16: 88072139-88072139 on Build GRCh38
rs958923465 Chr.16: 88072326-88072326 on Build GRCh38
rs1017437736 Chr.16: 88072120-88072120 on Build GRCh38
rs991259125 Chr.16: 88072205-88072205 on Build GRCh38
rs753554190 Chr.16: 88072171-88072171 on Build GRCh38
rs534830814 Chr.16: 88072259-88072259 on Build GRCh38
rs749565724 Chr.16: 88072206-88072206 on Build GRCh38
rs752044766 Chr.16: 88072192-88072192 on Build GRCh38
rs755889634 Chr.16: 88072114-88072114 on Build GRCh38
rs981688771 Chr.16: 88072195-88072195 on Build GRCh38
rs545702496 Chr.16: 88072340-88072340 on Build GRCh38
rs1003058865 Chr.16: 88072005-88072018 on Build GRCh38
rs781727956 Chr.16: 88072322-88072322 on Build GRCh38
rs776799978 Chr.16: 88072093-88072093 on Build GRCh38
rs747281915 Chr.16: 88072065-88072065 on Build GRCh38
rs1036314956 Chr.16: 88072321-88072321 on Build GRCh38

View Full Product Details