Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88647961-88648467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYBA
Entrez Gene ID
1535
Gene Name
cytochrome b-245 alpha chain
Gene Aliases
p22-PHOX
UniGene
Hs.513803
Gene Chromosome Location
Chr.16: 88643289-88651084 on Build GRCh38
Transcripts
NM_000101.3 XM_011522905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6638769 c.79G>A p.V27M Chr.16: 88648094-88648094 on Build GRCh38
5458598 c.59-8C>A p.? Chr.16: 88648122-88648122 on Build GRCh38
356824 c.63C>T p.L21L Chr.16: 88648110-88648110 on Build GRCh38
1609768 c.128+9C>A p.? Chr.16: 88648036-88648036 on Build GRCh38
4063496 c.117T>C p.G39G Chr.16: 88648056-88648056 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1036637407 Chr.16: 88648429-88648429 on Build GRCh38
rs554551021 Chr.16: 88648373-88648373 on Build GRCh38
rs759139349 Chr.16: 88648095-88648095 on Build GRCh38
rs72558349 Chr.16: 88648179-88648179 on Build GRCh38
rs878859963 Chr.16: 88648260-88648260 on Build GRCh38
rs748127011 Chr.16: 88648154-88648154 on Build GRCh38
rs375141871 Chr.16: 88648110-88648110 on Build GRCh38
rs939412714 Chr.16: 88648176-88648176 on Build GRCh38
rs916668450 Chr.16: 88648383-88648383 on Build GRCh38
rs1014993594 Chr.16: 88648268-88648268 on Build GRCh38
rs753268783 Chr.16: 88648157-88648157 on Build GRCh38
rs771834031 Chr.16: 88648155-88648155 on Build GRCh38
rs576454526 Chr.16: 88648426-88648426 on Build GRCh38
rs372615824 Chr.16: 88648014-88648014 on Build GRCh38
rs374776337 Chr.16: 88648068-88648068 on Build GRCh38
rs67141749 Chr.16: 88648402-88648402 on Build GRCh38
rs1012426944 Chr.16: 88648224-88648224 on Build GRCh38
rs780473138 Chr.16: 88648137-88648137 on Build GRCh38
rs543852457 Chr.16: 88648308-88648308 on Build GRCh38
rs537984397 Chr.16: 88648419-88648419 on Build GRCh38
rs35099352 Chr.16: 88648414-88648414 on Build GRCh38
rs778002755 Chr.16: 88648404-88648404 on Build GRCh38
rs749332805 Chr.16: 88648040-88648040 on Build GRCh38
rs537563434 Chr.16: 88648203-88648203 on Build GRCh38
rs917339154 Chr.16: 88648318-88648318 on Build GRCh38
rs185846992 Chr.16: 88648119-88648119 on Build GRCh38
rs1006983277 Chr.16: 88648271-88648271 on Build GRCh38
rs375028199 Chr.16: 88648374-88648374 on Build GRCh38
rs867350668 Chr.16: 88648414-88648414 on Build GRCh38
rs536109017 Chr.16: 88648339-88648339 on Build GRCh38
rs967252901 Chr.16: 88648324-88648324 on Build GRCh38
rs781702057 Chr.16: 88648113-88648113 on Build GRCh38
rs918386968 Chr.16: 88648394-88648394 on Build GRCh38
rs2306422 Chr.16: 88648151-88648151 on Build GRCh38
rs746151688 Chr.16: 88648017-88648017 on Build GRCh38
rs561092122 Chr.16: 88648152-88648152 on Build GRCh38
rs572754164 Chr.16: 88647996-88647996 on Build GRCh38
rs1049301647 Chr.16: 88648251-88648251 on Build GRCh38
rs568138482 Chr.16: 88648079-88648079 on Build GRCh38
rs558349758 Chr.16: 88648175-88648175 on Build GRCh38
rs765420485 Chr.16: 88647987-88647987 on Build GRCh38
rs867528553 Chr.16: 88648304-88648304 on Build GRCh38
rs983116270 Chr.16: 88648065-88648065 on Build GRCh38
rs888028625 Chr.16: 88648256-88648256 on Build GRCh38
rs377306683 Chr.16: 88648016-88648016 on Build GRCh38
rs971392363 Chr.16: 88648338-88648338 on Build GRCh38
rs753373754 Chr.16: 88648094-88648094 on Build GRCh38
rs199989792 Chr.16: 88648118-88648118 on Build GRCh38
rs770085128 Chr.16: 88648018-88648018 on Build GRCh38
rs749488868 Chr.16: 88648139-88648139 on Build GRCh38
rs890859549 Chr.16: 88648415-88648415 on Build GRCh38
rs753275155 Chr.16: 88648002-88648002 on Build GRCh38
rs764924055 Chr.16: 88648012-88648012 on Build GRCh38
rs774384826 Chr.16: 88648057-88648057 on Build GRCh38
rs72558369 Chr.16: 88648316-88648316 on Build GRCh38
rs1028755666 Chr.16: 88648297-88648297 on Build GRCh38
rs751990836 Chr.16: 88648009-88648009 on Build GRCh38
rs965765991 Chr.16: 88648276-88648276 on Build GRCh38
rs895284844 Chr.16: 88648221-88648221 on Build GRCh38
rs72558370 Chr.16: 88648360-88648360 on Build GRCh38
rs755407489 Chr.16: 88648035-88648035 on Build GRCh38
rs201647266 Chr.16: 88648157-88648157 on Build GRCh38
rs572665538 Chr.16: 88648377-88648377 on Build GRCh38
rs956215436 Chr.16: 88648309-88648309 on Build GRCh38
rs529905180 Chr.16: 88648104-88648104 on Build GRCh38
rs780285474 Chr.16: 88648023-88648023 on Build GRCh38
rs11547384 Chr.16: 88648051-88648051 on Build GRCh38
rs368862873 Chr.16: 88648078-88648078 on Build GRCh38
rs902540644 Chr.16: 88648159-88648159 on Build GRCh38
rs189240658 Chr.16: 88648396-88648396 on Build GRCh38
rs756441248 Chr.16: 88648399-88648399 on Build GRCh38
rs965313762 Chr.16: 88648286-88648286 on Build GRCh38
rs758938974 Chr.16: 88648007-88648007 on Build GRCh38
rs890775916 Chr.16: 88648269-88648269 on Build GRCh38
rs781763387 Chr.16: 88648283-88648283 on Build GRCh38
rs769306693 Chr.16: 88648434-88648437 on Build GRCh38
rs768595936 Chr.16: 88648052-88648052 on Build GRCh38
rs975888280 Chr.16: 88648087-88648087 on Build GRCh38
rs771957240 Chr.16: 88648059-88648059 on Build GRCh38
rs28941476 Chr.16: 88648103-88648103 on Build GRCh38
rs72549197 Chr.16: 88648025-88648025 on Build GRCh38
rs1804007 Chr.16: 88648060-88648060 on Build GRCh38
rs759109333 Chr.16: 88648165-88648165 on Build GRCh38
rs926147892 Chr.16: 88648015-88648015 on Build GRCh38
rs993907638 Chr.16: 88648250-88648250 on Build GRCh38
rs765972539 Chr.16: 88648084-88648084 on Build GRCh38
rs576572338 Chr.16: 88648198-88648198 on Build GRCh38
rs778297984 Chr.16: 88648008-88648008 on Build GRCh38
rs4268732 Chr.16: 88648382-88648382 on Build GRCh38
rs767870349 Chr.16: 88648110-88648112 on Build GRCh38
rs1804006 Chr.16: 88648088-88648088 on Build GRCh38
rs927214859 Chr.16: 88648146-88648146 on Build GRCh38
rs765884593 Chr.16: 88648001-88648001 on Build GRCh38
rs179363891 Chr.16: 88648099-88648099 on Build GRCh38
rs751896984 Chr.16: 88648021-88648021 on Build GRCh38
rs375616744 Chr.16: 88648010-88648010 on Build GRCh38
rs770694725 Chr.16: 88648161-88648161 on Build GRCh38
rs1018292854 Chr.16: 88647992-88647992 on Build GRCh38
rs35786745 Chr.16: 88648176-88648177 on Build GRCh38
rs115151135 Chr.16: 88648258-88648258 on Build GRCh38

View Full Product Details