Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88709946-88710439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTU2
Entrez Gene ID
348180
Gene Name
cytosolic thiouridylase subunit 2
Gene Aliases
C16orf84, NCS2, UPF0432
UniGene
Hs.592074
Gene Chromosome Location
Chr.16: 88706463-88715386 on Build GRCh38
Transcripts
NM_001012759.2 NM_001012762.2 NM_001318507.1 NM_001318513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6637953 c.166G>A p.V56I Chr.16: 88709960-88709960 on Build GRCh38
7429865 c.234G>A p.A78A Chr.16: 88710234-88710234 on Build GRCh38
7429866 c.234G>A p.A78A Chr.16: 88710234-88710234 on Build GRCh38
4922766 c.162C>G p.F54L Chr.16: 88709956-88709956 on Build GRCh38
4926627 c.153C>T p.F51F Chr.16: 88709947-88709947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766640977 Chr.16: 88710051-88710051 on Build GRCh38
rs984332015 Chr.16: 88710162-88710162 on Build GRCh38
rs765273195 Chr.16: 88710056-88710056 on Build GRCh38
rs375329250 Chr.16: 88710036-88710036 on Build GRCh38
rs1002247519 Chr.16: 88709976-88709976 on Build GRCh38
rs925587613 Chr.16: 88710167-88710167 on Build GRCh38
rs9889176 Chr.16: 88710194-88710194 on Build GRCh38
rs747397487 Chr.16: 88710188-88710188 on Build GRCh38
rs765265257 Chr.16: 88710298-88710302 on Build GRCh38
rs374826247 Chr.16: 88710011-88710011 on Build GRCh38
rs763798441 Chr.16: 88710207-88710208 on Build GRCh38
rs781681529 Chr.16: 88710292-88710292 on Build GRCh38
rs531378397 Chr.16: 88710021-88710021 on Build GRCh38
rs751411884 Chr.16: 88710062-88710062 on Build GRCh38
rs1019135186 Chr.16: 88710317-88710317 on Build GRCh38
rs984874799 Chr.16: 88710377-88710377 on Build GRCh38
rs925679563 Chr.16: 88710042-88710042 on Build GRCh38
rs201488917 Chr.16: 88710192-88710192 on Build GRCh38
rs780649645 Chr.16: 88710028-88710028 on Build GRCh38
rs778284872 Chr.16: 88710233-88710233 on Build GRCh38
rs756950162 Chr.16: 88710172-88710172 on Build GRCh38
rs960760858 Chr.16: 88710117-88710117 on Build GRCh38
rs990482201 Chr.16: 88710127-88710127 on Build GRCh38
rs762193182 Chr.16: 88709977-88709977 on Build GRCh38
rs770154511 Chr.16: 88710247-88710247 on Build GRCh38
rs1033715743 Chr.16: 88709981-88709981 on Build GRCh38
rs371438692 Chr.16: 88710177-88710177 on Build GRCh38
rs778444062 Chr.16: 88710273-88710273 on Build GRCh38
rs779619510 Chr.16: 88710220-88710220 on Build GRCh38
rs760498224 Chr.16: 88710326-88710326 on Build GRCh38
rs549808540 Chr.16: 88710022-88710022 on Build GRCh38
rs764866819 Chr.16: 88710334-88710334 on Build GRCh38
rs953958827 Chr.16: 88710079-88710079 on Build GRCh38
rs114994631 Chr.16: 88710216-88710216 on Build GRCh38
rs1026364180 Chr.16: 88710121-88710121 on Build GRCh38
rs764833613 Chr.16: 88710010-88710010 on Build GRCh38
rs753007504 Chr.16: 88710191-88710191 on Build GRCh38
rs916283388 Chr.16: 88710151-88710151 on Build GRCh38
rs750767961 Chr.16: 88709984-88709984 on Build GRCh38
rs779559096 Chr.16: 88710320-88710320 on Build GRCh38
rs760964617 Chr.16: 88710047-88710047 on Build GRCh38
rs61744010 Chr.16: 88710091-88710091 on Build GRCh38
rs775716328 Chr.16: 88710195-88710195 on Build GRCh38
rs754595424 Chr.16: 88710265-88710265 on Build GRCh38
rs377252732 Chr.16: 88710020-88710020 on Build GRCh38
rs376556556 Chr.16: 88710257-88710257 on Build GRCh38
rs889746448 Chr.16: 88710232-88710232 on Build GRCh38
rs145251953 Chr.16: 88710234-88710234 on Build GRCh38
rs754956537 Chr.16: 88709995-88709995 on Build GRCh38
rs948973748 Chr.16: 88710007-88710007 on Build GRCh38
rs753730609 Chr.16: 88710208-88710208 on Build GRCh38
rs972919607 Chr.16: 88710405-88710405 on Build GRCh38
rs570086641 Chr.16: 88710409-88710409 on Build GRCh38
rs549719454 Chr.16: 88710252-88710252 on Build GRCh38
rs998125719 Chr.16: 88710068-88710068 on Build GRCh38
rs369910606 Chr.16: 88710309-88710309 on Build GRCh38
rs754439278 Chr.16: 88710231-88710231 on Build GRCh38
rs767489235 Chr.16: 88710254-88710254 on Build GRCh38
rs927705325 Chr.16: 88709980-88709980 on Build GRCh38
rs183824126 Chr.16: 88710163-88710163 on Build GRCh38
rs759245573 Chr.16: 88710329-88710329 on Build GRCh38
rs760949995 Chr.16: 88709992-88709992 on Build GRCh38
rs747681590 Chr.16: 88710280-88710280 on Build GRCh38
rs144950335 Chr.16: 88710314-88710314 on Build GRCh38
rs746176941 Chr.16: 88710246-88710246 on Build GRCh38
rs910119594 Chr.16: 88710392-88710392 on Build GRCh38
rs536176195 Chr.16: 88710058-88710058 on Build GRCh38
rs773654933 Chr.16: 88710183-88710183 on Build GRCh38
rs375583556 Chr.16: 88710301-88710301 on Build GRCh38
rs901257037 Chr.16: 88710213-88710213 on Build GRCh38
rs1046023552 Chr.16: 88710072-88710072 on Build GRCh38
rs149997219 Chr.16: 88709994-88709994 on Build GRCh38
rs912990354 Chr.16: 88710228-88710228 on Build GRCh38
rs761639757 Chr.16: 88710240-88710241 on Build GRCh38
rs761888813 Chr.16: 88710312-88710312 on Build GRCh38
rs951667052 Chr.16: 88710337-88710337 on Build GRCh38
rs1038263056 Chr.16: 88710297-88710297 on Build GRCh38
rs748812385 Chr.16: 88710223-88710223 on Build GRCh38
rs372288432 Chr.16: 88710059-88710059 on Build GRCh38
rs757184728 Chr.16: 88710214-88710214 on Build GRCh38
rs78353973 Chr.16: 88710202-88710202 on Build GRCh38
rs949923900 Chr.16: 88710070-88710070 on Build GRCh38
rs755882986 Chr.16: 88710256-88710256 on Build GRCh38
rs543371035 Chr.16: 88710161-88710161 on Build GRCh38
rs760402352 Chr.16: 88710033-88710033 on Build GRCh38
rs899190054 Chr.16: 88710108-88710108 on Build GRCh38
rs147948789 Chr.16: 88709982-88709982 on Build GRCh38
rs374513861 Chr.16: 88710295-88710295 on Build GRCh38
rs751499655 Chr.16: 88710205-88710205 on Build GRCh38
rs1002601583 Chr.16: 88710084-88710084 on Build GRCh38
rs745402860 Chr.16: 88710031-88710031 on Build GRCh38
rs950908207 Chr.16: 88710136-88710136 on Build GRCh38
rs200066636 Chr.16: 88710061-88710061 on Build GRCh38
rs749568507 Chr.16: 88710303-88710303 on Build GRCh38
rs527527613 Chr.16: 88710181-88710181 on Build GRCh38
rs115768982 Chr.16: 88710053-88710053 on Build GRCh38
rs987697680 Chr.16: 88710142-88710142 on Build GRCh38
rs775804412 Chr.16: 88710251-88710251 on Build GRCh38
rs369427791 Chr.16: 88710286-88710286 on Build GRCh38
rs376122307 Chr.16: 88710328-88710328 on Build GRCh38
rs147078272 Chr.16: 88710300-88710300 on Build GRCh38
rs773157162 Chr.16: 88710321-88710321 on Build GRCh38
rs1010746103 Chr.16: 88710089-88710089 on Build GRCh38
rs574363880 Chr.16: 88710166-88710166 on Build GRCh38
rs199634578 Chr.16: 88710308-88710308 on Build GRCh38
rs576352261 Chr.16: 88710143-88710143 on Build GRCh38
rs758380123 Chr.16: 88710016-88710016 on Build GRCh38
rs560067038 Chr.16: 88710179-88710179 on Build GRCh38
rs149072520 Chr.16: 88710397-88710397 on Build GRCh38
rs781684769 Chr.16: 88710109-88710109 on Build GRCh38
rs767261006 Chr.16: 88710215-88710215 on Build GRCh38
rs775030045 Chr.16: 88710041-88710041 on Build GRCh38
rs763066792 Chr.16: 88710201-88710201 on Build GRCh38
rs978343832 Chr.16: 88710110-88710110 on Build GRCh38
rs774669128 Chr.16: 88709974-88709974 on Build GRCh38
rs371609784 Chr.16: 88710245-88710245 on Build GRCh38
rs771447868 Chr.16: 88710238-88710238 on Build GRCh38
rs775710725 Chr.16: 88709965-88709966 on Build GRCh38
rs751652887 Chr.16: 88710009-88710009 on Build GRCh38
rs927722426 Chr.16: 88710116-88710116 on Build GRCh38
rs769216967 Chr.16: 88709973-88709973 on Build GRCh38
rs200836148 Chr.16: 88709993-88709993 on Build GRCh38
rs778933792 Chr.16: 88710000-88710000 on Build GRCh38
rs202164726 Chr.16: 88710173-88710173 on Build GRCh38
rs761711234 Chr.16: 88710253-88710253 on Build GRCh38
rs141766248 Chr.16: 88710074-88710074 on Build GRCh38
rs750193972 Chr.16: 88710255-88710255 on Build GRCh38
rs965265129 Chr.16: 88710334-88710334 on Build GRCh38
rs188101062 Chr.16: 88710411-88710411 on Build GRCh38
rs772557939 Chr.16: 88710045-88710045 on Build GRCh38
rs566299665 Chr.16: 88710069-88710069 on Build GRCh38
rs778036046 Chr.16: 88710267-88710267 on Build GRCh38

View Full Product Details