Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88714014-88714513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTU2
Entrez Gene ID
348180
Gene Name
cytosolic thiouridylase subunit 2
Gene Aliases
C16orf84, NCS2, UPF0432
UniGene
Hs.592074
Gene Chromosome Location
Chr.16: 88706463-88715386 on Build GRCh38
Transcripts
NM_001012759.2 NM_001012762.2 NM_001318507.1 NM_001318513.1 XM_017023210.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6080290 c.1408G>C p.A470P Chr.16: 88714480-88714480 on Build GRCh38
3387672 c.1196C>T p.A399V Chr.16: 88714481-88714481 on Build GRCh38
4675519 c.1031G>A p.R344Q Chr.16: 88714161-88714161 on Build GRCh38
7295006 c.1104T>C p.S368S Chr.16: 88714389-88714389 on Build GRCh38
2922396 c.1163G>A p.C388Y Chr.16: 88714448-88714448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000911929 Chr.16: 88714089-88714089 on Build GRCh38
rs796110405 Chr.16: 88714232-88714233 on Build GRCh38
rs145828182 Chr.16: 88714473-88714473 on Build GRCh38
rs772769195 Chr.16: 88714205-88714205 on Build GRCh38
rs61734068 Chr.16: 88714217-88714217 on Build GRCh38
rs753542455 Chr.16: 88714481-88714481 on Build GRCh38
rs1016014080 Chr.16: 88714142-88714142 on Build GRCh38
rs910169942 Chr.16: 88714312-88714312 on Build GRCh38
rs201768632 Chr.16: 88714263-88714263 on Build GRCh38
rs562672342 Chr.16: 88714228-88714237 on Build GRCh38
rs878935902 Chr.16: 88714274-88714274 on Build GRCh38
rs781218199 Chr.16: 88714236-88714256 on Build GRCh38
rs549166093 Chr.16: 88714309-88714309 on Build GRCh38
rs774126798 Chr.16: 88714149-88714149 on Build GRCh38
rs908959061 Chr.16: 88714051-88714051 on Build GRCh38
rs1039927775 Chr.16: 88714067-88714067 on Build GRCh38
rs754958017 Chr.16: 88714255-88714255 on Build GRCh38
rs569895054 Chr.16: 88714065-88714065 on Build GRCh38
rs778181869 Chr.16: 88714452-88714457 on Build GRCh38
rs761070966 Chr.16: 88714382-88714382 on Build GRCh38
rs906897106 Chr.16: 88714132-88714132 on Build GRCh38
rs977634147 Chr.16: 88714420-88714420 on Build GRCh38
rs987580847 Chr.16: 88714213-88714213 on Build GRCh38
rs770211787 Chr.16: 88714239-88714240 on Build GRCh38
rs575472278 Chr.16: 88714239-88714239 on Build GRCh38
rs377401588 Chr.16: 88714359-88714359 on Build GRCh38
rs776211558 Chr.16: 88714216-88714216 on Build GRCh38
rs368817425 Chr.16: 88714251-88714251 on Build GRCh38
rs767079692 Chr.16: 88714109-88714109 on Build GRCh38
rs758912470 Chr.16: 88714256-88714257 on Build GRCh38
rs557189445 Chr.16: 88714346-88714346 on Build GRCh38
rs894248730 Chr.16: 88714328-88714328 on Build GRCh38
rs1016149911 Chr.16: 88714279-88714279 on Build GRCh38
rs145565756 Chr.16: 88714273-88714282 on Build GRCh38
rs765883350 Chr.16: 88714161-88714161 on Build GRCh38
rs74880807 Chr.16: 88714080-88714080 on Build GRCh38
rs375039500 Chr.16: 88714253-88714253 on Build GRCh38
rs769987116 Chr.16: 88714429-88714429 on Build GRCh38
rs371428735 Chr.16: 88714440-88714440 on Build GRCh38
rs765613896 Chr.16: 88714115-88714115 on Build GRCh38
rs368302599 Chr.16: 88714122-88714122 on Build GRCh38
rs748409186 Chr.16: 88714240-88714243 on Build GRCh38
rs771795709 Chr.16: 88714202-88714202 on Build GRCh38
rs563534005 Chr.16: 88714219-88714219 on Build GRCh38
rs1036076265 Chr.16: 88714053-88714053 on Build GRCh38
rs749052411 Chr.16: 88714482-88714483 on Build GRCh38
rs758789738 Chr.16: 88714272-88714273 on Build GRCh38
rs201076790 Chr.16: 88714101-88714101 on Build GRCh38
rs761458553 Chr.16: 88714150-88714150 on Build GRCh38
rs755522715 Chr.16: 88714114-88714114 on Build GRCh38
rs757808758 Chr.16: 88714487-88714487 on Build GRCh38
rs370141298 Chr.16: 88714370-88714370 on Build GRCh38
rs760164445 Chr.16: 88714044-88714044 on Build GRCh38
rs367965619 Chr.16: 88714336-88714336 on Build GRCh38
rs370722300 Chr.16: 88714104-88714104 on Build GRCh38
rs369446542 Chr.16: 88714262-88714262 on Build GRCh38
rs773623901 Chr.16: 88714337-88714337 on Build GRCh38
rs760324933 Chr.16: 88714206-88714206 on Build GRCh38
rs958242068 Chr.16: 88714434-88714434 on Build GRCh38
rs753902691 Chr.16: 88714254-88714254 on Build GRCh38
rs767878077 Chr.16: 88714248-88714248 on Build GRCh38
rs757757646 Chr.16: 88714230-88714231 on Build GRCh38
rs779893329 Chr.16: 88714140-88714140 on Build GRCh38
rs397761749 Chr.16: 88714233-88714238 on Build GRCh38
rs908340282 Chr.16: 88714464-88714464 on Build GRCh38
rs200076111 Chr.16: 88714438-88714438 on Build GRCh38
rs763543064 Chr.16: 88714168-88714168 on Build GRCh38
rs536806393 Chr.16: 88714300-88714300 on Build GRCh38
rs571090098 Chr.16: 88714232-88714233 on Build GRCh38
rs1026016108 Chr.16: 88714293-88714293 on Build GRCh38
rs776859749 Chr.16: 88714387-88714387 on Build GRCh38
rs1046496720 Chr.16: 88714356-88714356 on Build GRCh38
rs759127335 Chr.16: 88714210-88714211 on Build GRCh38
rs760864835 Chr.16: 88714340-88714340 on Build GRCh38
rs778227960 Chr.16: 88714446-88714446 on Build GRCh38
rs770359913 Chr.16: 88714211-88714211 on Build GRCh38
rs762980841 Chr.16: 88714093-88714093 on Build GRCh38
rs754518249 Chr.16: 88714482-88714482 on Build GRCh38
rs975632390 Chr.16: 88714321-88714321 on Build GRCh38
rs61730419 Chr.16: 88714412-88714412 on Build GRCh38
rs899042540 Chr.16: 88714226-88714227 on Build GRCh38
rs758149117 Chr.16: 88714259-88714259 on Build GRCh38
rs932996175 Chr.16: 88714056-88714056 on Build GRCh38
rs149847195 Chr.16: 88714227-88714228 on Build GRCh38
rs773498927 Chr.16: 88714380-88714380 on Build GRCh38
rs918901849 Chr.16: 88714055-88714055 on Build GRCh38
rs371899645 Chr.16: 88714218-88714218 on Build GRCh38
rs756173624 Chr.16: 88714186-88714186 on Build GRCh38
rs749145713 Chr.16: 88714252-88714265 on Build GRCh38
rs538934008 Chr.16: 88714345-88714345 on Build GRCh38
rs746802588 Chr.16: 88714363-88714363 on Build GRCh38
rs762612396 Chr.16: 88714095-88714095 on Build GRCh38
rs149927003 Chr.16: 88714415-88714415 on Build GRCh38
rs551005079 Chr.16: 88714445-88714445 on Build GRCh38
rs764768032 Chr.16: 88714369-88714369 on Build GRCh38
rs865903079 Chr.16: 88714241-88714241 on Build GRCh38
rs771566209 Chr.16: 88714154-88714154 on Build GRCh38
rs963170461 Chr.16: 88714282-88714282 on Build GRCh38
rs748236272 Chr.16: 88714280-88714280 on Build GRCh38
rs759817868 Chr.16: 88714428-88714428 on Build GRCh38
rs755110952 Chr.16: 88714234-88714239 on Build GRCh38
rs573239956 Chr.16: 88714407-88714407 on Build GRCh38
rs377604379 Chr.16: 88714375-88714375 on Build GRCh38
rs770862024 Chr.16: 88714490-88714491 on Build GRCh38
rs771297629 Chr.16: 88714421-88714421 on Build GRCh38
rs545320542 Chr.16: 88714201-88714201 on Build GRCh38
rs766689276 Chr.16: 88714385-88714385 on Build GRCh38
rs569858310 Chr.16: 88714290-88714290 on Build GRCh38
rs936822224 Chr.16: 88714338-88714338 on Build GRCh38
rs748811574 Chr.16: 88714413-88714413 on Build GRCh38
rs768496364 Chr.16: 88714147-88714147 on Build GRCh38
rs371659918 Chr.16: 88714252-88714252 on Build GRCh38
rs945320585 Chr.16: 88714063-88714063 on Build GRCh38
rs781592397 Chr.16: 88714461-88714461 on Build GRCh38
rs769798425 Chr.16: 88714137-88714137 on Build GRCh38
rs569014529 Chr.16: 88714277-88714278 on Build GRCh38
rs751531565 Chr.16: 88714432-88714432 on Build GRCh38
rs199646051 Chr.16: 88714093-88714093 on Build GRCh38
rs3826076 Chr.16: 88714110-88714110 on Build GRCh38
rs764035913 Chr.16: 88714431-88714431 on Build GRCh38
rs767780142 Chr.16: 88714335-88714335 on Build GRCh38
rs758322993 Chr.16: 88714399-88714399 on Build GRCh38
rs776563195 Chr.16: 88714085-88714085 on Build GRCh38
rs528606137 Chr.16: 88714256-88714256 on Build GRCh38
rs76217997 Chr.16: 88714237-88714237 on Build GRCh38
rs770038426 Chr.16: 88714468-88714468 on Build GRCh38
rs760064522 Chr.16: 88714160-88714160 on Build GRCh38
rs368748040 Chr.16: 88714173-88714173 on Build GRCh38
rs764623963 Chr.16: 88714171-88714171 on Build GRCh38
rs778791403 Chr.16: 88714257-88714257 on Build GRCh38
rs552680350 Chr.16: 88714323-88714323 on Build GRCh38
rs780126764 Chr.16: 88714190-88714190 on Build GRCh38
rs772800919 Chr.16: 88714159-88714159 on Build GRCh38
rs147808203 Chr.16: 88714476-88714476 on Build GRCh38
rs766039051 Chr.16: 88714480-88714480 on Build GRCh38
rs759695208 Chr.16: 88714393-88714393 on Build GRCh38
rs372733301 Chr.16: 88714174-88714174 on Build GRCh38
rs534734692 Chr.16: 88714315-88714315 on Build GRCh38
rs781244722 Chr.16: 88714180-88714180 on Build GRCh38
rs954570510 Chr.16: 88714404-88714404 on Build GRCh38
rs61730418 Chr.16: 88714405-88714405 on Build GRCh38
rs942877278 Chr.16: 88714066-88714066 on Build GRCh38
rs767058128 Chr.16: 88714261-88714287 on Build GRCh38
rs899126271 Chr.16: 88714389-88714389 on Build GRCh38
rs775076153 Chr.16: 88714371-88714371 on Build GRCh38
rs374457332 Chr.16: 88714479-88714479 on Build GRCh38
rs777493178 Chr.16: 88714362-88714362 on Build GRCh38
rs773129655 Chr.16: 88714241-88714242 on Build GRCh38
rs774378446 Chr.16: 88714257-88714273 on Build GRCh38
rs938395699 Chr.16: 88714138-88714138 on Build GRCh38
rs776912579 Chr.16: 88714424-88714424 on Build GRCh38
rs757607044 Chr.16: 88714177-88714177 on Build GRCh38
rs747296499 Chr.16: 88714125-88714125 on Build GRCh38
rs762325925 Chr.16: 88714334-88714334 on Build GRCh38
rs767027896 Chr.16: 88714222-88714223 on Build GRCh38
rs868591049 Chr.16: 88714157-88714157 on Build GRCh38
rs758875967 Chr.16: 88714119-88714119 on Build GRCh38
rs552957377 Chr.16: 88714118-88714118 on Build GRCh38
rs141153247 Chr.16: 88714310-88714310 on Build GRCh38
rs766484216 Chr.16: 88714137-88714137 on Build GRCh38
rs761440186 Chr.16: 88714107-88714107 on Build GRCh38
rs919039314 Chr.16: 88714187-88714187 on Build GRCh38
rs763030593 Chr.16: 88714472-88714472 on Build GRCh38
rs200108886 Chr.16: 88714097-88714097 on Build GRCh38
rs752643561 Chr.16: 88714352-88714352 on Build GRCh38
rs753194868 Chr.16: 88714162-88714162 on Build GRCh38
rs745882276 Chr.16: 88714135-88714135 on Build GRCh38
rs7205626 Chr.16: 88714064-88714064 on Build GRCh38
rs200518996 Chr.16: 88714374-88714374 on Build GRCh38
rs140479732 Chr.16: 88714282-88714283 on Build GRCh38
rs368218174 Chr.16: 88714474-88714474 on Build GRCh38
rs752187784 Chr.16: 88714486-88714486 on Build GRCh38
rs766826191 Chr.16: 88714276-88714281 on Build GRCh38
rs201634219 Chr.16: 88714086-88714087 on Build GRCh38
rs75686921 Chr.16: 88714233-88714233 on Build GRCh38
rs147606000 Chr.16: 88714437-88714437 on Build GRCh38
rs941752727 Chr.16: 88714052-88714052 on Build GRCh38
rs745344367 Chr.16: 88714267-88714267 on Build GRCh38
rs969232228 Chr.16: 88714035-88714035 on Build GRCh38
rs868851958 Chr.16: 88714280-88714280 on Build GRCh38
rs369399187 Chr.16: 88714271-88714277 on Build GRCh38
rs749230204 Chr.16: 88714194-88714194 on Build GRCh38
rs772495350 Chr.16: 88714414-88714414 on Build GRCh38
rs370661454 Chr.16: 88714411-88714411 on Build GRCh38
rs7191439 Chr.16: 88714277-88714277 on Build GRCh38
rs995795235 Chr.16: 88714084-88714084 on Build GRCh38
rs942912304 Chr.16: 88714225-88714225 on Build GRCh38
rs368211946 Chr.16: 88714130-88714130 on Build GRCh38
rs752014699 Chr.16: 88714232-88714232 on Build GRCh38
rs770961865 Chr.16: 88714252-88714252 on Build GRCh38
rs764868235 Chr.16: 88714485-88714485 on Build GRCh38
rs756904891 Chr.16: 88714365-88714365 on Build GRCh38
rs1046906956 Chr.16: 88714452-88714452 on Build GRCh38
rs780574958 Chr.16: 88714230-88714256 on Build GRCh38
rs773976595 Chr.16: 88714209-88714210 on Build GRCh38
rs73262673 Chr.16: 88714195-88714195 on Build GRCh38
rs11278302 Chr.16: 88714227-88714232 on Build GRCh38
rs769555461 Chr.16: 88714087-88714087 on Build GRCh38
rs775809303 Chr.16: 88714469-88714469 on Build GRCh38
rs142838333 Chr.16: 88714396-88714396 on Build GRCh38
rs750285064 Chr.16: 88714477-88714477 on Build GRCh38
rs546642809 Chr.16: 88714260-88714260 on Build GRCh38
rs756385186 Chr.16: 88714438-88714438 on Build GRCh38
rs374364742 Chr.16: 88714453-88714453 on Build GRCh38
rs748037506 Chr.16: 88714258-88714258 on Build GRCh38
rs760304237 Chr.16: 88714271-88714280 on Build GRCh38
rs1039982227 Chr.16: 88714238-88714238 on Build GRCh38
rs569260103 Chr.16: 88714117-88714117 on Build GRCh38
rs993333216 Chr.16: 88714483-88714483 on Build GRCh38
rs369947041 Chr.16: 88714342-88714342 on Build GRCh38
rs374770342 Chr.16: 88714250-88714250 on Build GRCh38
rs771422328 Chr.16: 88714458-88714458 on Build GRCh38
rs775477017 Chr.16: 88714430-88714430 on Build GRCh38
rs376466131 Chr.16: 88714144-88714144 on Build GRCh38
rs775024882 Chr.16: 88714278-88714278 on Build GRCh38
rs753392867 Chr.16: 88714444-88714444 on Build GRCh38
rs774928478 Chr.16: 88714265-88714266 on Build GRCh38
rs779727077 Chr.16: 88714441-88714441 on Build GRCh38

View Full Product Details