Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88822505-88823006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNS
Entrez Gene ID
2588
Gene Name
galactosamine (N-acetyl)-6-sulfatase
Gene Aliases
GALNAC6S, GAS, GalN6S, MPS4A
UniGene
Hs.271383
Gene Chromosome Location
Chr.16: 88813734-88856970 on Build GRCh38
Transcripts
NM_000512.4 NM_001323543.1 NM_001323544.1 XM_005256301.3 XM_011522982.2 XM_017023111.1 XM_017023112.1 XM_017023113.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3957923 c.1356C>A p.F452L Chr.16: 88822597-88822597 on Build GRCh38
4448844 c.1360delC p.L454fs*41 Chr.16: 88822593-88822593 on Build GRCh38
6668955 c.1259C>A p.P420H Chr.16: 88822694-88822694 on Build GRCh38
3513220 c.1327C>T p.H443Y Chr.16: 88822626-88822626 on Build GRCh38
974781 c.1308G>A p.T436T Chr.16: 88822645-88822645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548331088 Chr.16: 88822852-88822852 on Build GRCh38
rs778286877 Chr.16: 88822634-88822635 on Build GRCh38
rs192600348 Chr.16: 88822886-88822886 on Build GRCh38
rs747836325 Chr.16: 88822760-88822760 on Build GRCh38
rs981584784 Chr.16: 88822976-88822976 on Build GRCh38
rs771530305 Chr.16: 88822526-88822526 on Build GRCh38
rs552690945 Chr.16: 88822814-88822814 on Build GRCh38
rs557217245 Chr.16: 88822617-88822617 on Build GRCh38
rs566315025 Chr.16: 88822887-88822887 on Build GRCh38
rs907277565 Chr.16: 88822756-88822756 on Build GRCh38
rs759760397 Chr.16: 88822606-88822606 on Build GRCh38
rs148709901 Chr.16: 88822684-88822684 on Build GRCh38
rs1019407568 Chr.16: 88822822-88822822 on Build GRCh38
rs747618641 Chr.16: 88822759-88822759 on Build GRCh38
rs781053828 Chr.16: 88822573-88822573 on Build GRCh38
rs576471416 Chr.16: 88822932-88822932 on Build GRCh38
rs560592708 Chr.16: 88822795-88822795 on Build GRCh38
rs564502999 Chr.16: 88822816-88822816 on Build GRCh38
rs779612747 Chr.16: 88822752-88822752 on Build GRCh38
rs550608526 Chr.16: 88822825-88822825 on Build GRCh38
rs946252826 Chr.16: 88822962-88822962 on Build GRCh38
rs367969896 Chr.16: 88822568-88822568 on Build GRCh38
rs761184988 Chr.16: 88822669-88822669 on Build GRCh38
rs924991010 Chr.16: 88822928-88822928 on Build GRCh38
rs757171965 Chr.16: 88822632-88822632 on Build GRCh38
rs768180795 Chr.16: 88822667-88822667 on Build GRCh38
rs75439331 Chr.16: 88822532-88822532 on Build GRCh38
rs773746427 Chr.16: 88822588-88822588 on Build GRCh38
rs751628798 Chr.16: 88822726-88822726 on Build GRCh38
rs758302397 Chr.16: 88822564-88822564 on Build GRCh38
rs375196202 Chr.16: 88822650-88822650 on Build GRCh38
rs771363009 Chr.16: 88822671-88822671 on Build GRCh38
rs774864952 Chr.16: 88822578-88822578 on Build GRCh38
rs769360669 Chr.16: 88822646-88822646 on Build GRCh38
rs368160993 Chr.16: 88822630-88822630 on Build GRCh38
rs886052455 Chr.16: 88822720-88822720 on Build GRCh38
rs769510297 Chr.16: 88822577-88822577 on Build GRCh38
rs184845441 Chr.16: 88822914-88822914 on Build GRCh38
rs752937387 Chr.16: 88822694-88822694 on Build GRCh38
rs766717526 Chr.16: 88822594-88822594 on Build GRCh38
rs971610814 Chr.16: 88822560-88822560 on Build GRCh38
rs527755420 Chr.16: 88822806-88822806 on Build GRCh38
rs142822371 Chr.16: 88822706-88822706 on Build GRCh38
rs368383143 Chr.16: 88822941-88822941 on Build GRCh38
rs568995509 Chr.16: 88822623-88822623 on Build GRCh38
rs398123432 Chr.16: 88822599-88822599 on Build GRCh38
rs371943842 Chr.16: 88822582-88822582 on Build GRCh38
rs538971843 Chr.16: 88822565-88822565 on Build GRCh38
rs571582625 Chr.16: 88822549-88822549 on Build GRCh38
rs542363475 Chr.16: 88822771-88822771 on Build GRCh38
rs554367594 Chr.16: 88822668-88822668 on Build GRCh38
rs569084669 Chr.16: 88822842-88822842 on Build GRCh38
rs757344618 Chr.16: 88822733-88822779 on Build GRCh38
rs757261806 Chr.16: 88822733-88822733 on Build GRCh38
rs778924788 Chr.16: 88822752-88822797 on Build GRCh38
rs913414045 Chr.16: 88822728-88822728 on Build GRCh38
rs536075935 Chr.16: 88822843-88822843 on Build GRCh38
rs772757609 Chr.16: 88822805-88822805 on Build GRCh38
rs748194485 Chr.16: 88822553-88822553 on Build GRCh38
rs779517946 Chr.16: 88822601-88822601 on Build GRCh38
rs928757137 Chr.16: 88822987-88822987 on Build GRCh38
rs762410752 Chr.16: 88822900-88822900 on Build GRCh38
rs974253456 Chr.16: 88822798-88822798 on Build GRCh38
rs778873962 Chr.16: 88822550-88822550 on Build GRCh38
rs766637780 Chr.16: 88822544-88822544 on Build GRCh38
rs749320235 Chr.16: 88822694-88822699 on Build GRCh38
rs531720908 Chr.16: 88822817-88822817 on Build GRCh38
rs765907150 Chr.16: 88822939-88822939 on Build GRCh38
rs751534350 Chr.16: 88822627-88822627 on Build GRCh38
rs750001517 Chr.16: 88822746-88822746 on Build GRCh38
rs929697450 Chr.16: 88822837-88822837 on Build GRCh38
rs7189744 Chr.16: 88822938-88822938 on Build GRCh38
rs76651187 Chr.16: 88822675-88822675 on Build GRCh38
rs189322865 Chr.16: 88822925-88822925 on Build GRCh38
rs368171136 Chr.16: 88822581-88822581 on Build GRCh38
rs1034716081 Chr.16: 88822547-88822547 on Build GRCh38
rs898468839 Chr.16: 88822772-88822772 on Build GRCh38
rs200110619 Chr.16: 88822757-88822757 on Build GRCh38
rs79025743 Chr.16: 88822755-88822755 on Build GRCh38
rs115109426 Chr.16: 88822953-88822953 on Build GRCh38
rs937093836 Chr.16: 88822587-88822587 on Build GRCh38
rs995451658 Chr.16: 88822780-88822780 on Build GRCh38
rs769395257 Chr.16: 88822764-88822764 on Build GRCh38
rs777720497 Chr.16: 88822567-88822567 on Build GRCh38
rs764216131 Chr.16: 88822713-88822713 on Build GRCh38
rs912355122 Chr.16: 88822880-88822880 on Build GRCh38
rs759848160 Chr.16: 88822677-88822677 on Build GRCh38
rs761068381 Chr.16: 88822593-88822593 on Build GRCh38
rs372784373 Chr.16: 88822740-88822740 on Build GRCh38
rs536402921 Chr.16: 88822645-88822645 on Build GRCh38
rs745498854 Chr.16: 88822574-88822574 on Build GRCh38
rs981654643 Chr.16: 88822768-88822768 on Build GRCh38
rs377120480 Chr.16: 88822583-88822583 on Build GRCh38
rs200956302 Chr.16: 88822735-88822735 on Build GRCh38
rs754055321 Chr.16: 88822545-88822545 on Build GRCh38
rs201676700 Chr.16: 88822543-88822543 on Build GRCh38
rs958396555 Chr.16: 88822794-88822795 on Build GRCh38
rs943993057 Chr.16: 88822881-88822881 on Build GRCh38
rs1054835249 Chr.16: 88822958-88822960 on Build GRCh38
rs777615878 Chr.16: 88822625-88822625 on Build GRCh38
rs748796648 Chr.16: 88822662-88822662 on Build GRCh38
rs574943680 Chr.16: 88822767-88822767 on Build GRCh38
rs557580871 Chr.16: 88822525-88822525 on Build GRCh38

View Full Product Details