Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88884926-88885416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBFA2T3
Entrez Gene ID
863
Gene Name
CBFA2/RUNX1 translocation partner 3
Gene Aliases
ETO2, MTG16, MTGR2, RUNX1T3, ZMYND4
UniGene
Hs.513811
Gene Chromosome Location
Chr.16: 88874855-88977198 on Build GRCh38
Transcripts
NM_005187.5 NM_175931.2 XM_005256323.4 XM_011523419.2 XM_017023811.1 NR_135194.1

Gene Symbol
LOC101927793
Entrez Gene ID
101927793
Gene Name
uncharacterized LOC101927793
Gene Aliases
-
UniGene
Hs.690953
Gene Chromosome Location
Chr.16: 88883515-88887136 on Build GRCh38
Transcripts
NM_005187.5 NM_175931.2 XM_005256323.4 XM_011523419.2 XM_017023811.1 NR_135194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
32425 c.917G>A p.R306H Chr.16: 88885246-88885246 on Build GRCh38
1247395 c.1061G>A p.R354Q Chr.16: 88885102-88885102 on Build GRCh38
974797 c.1028G>A p.R343H Chr.16: 88885135-88885135 on Build GRCh38
7423683 c.1060C>T p.R354* Chr.16: 88885103-88885103 on Build GRCh38
1237731 c.964A>G p.N322D Chr.16: 88885199-88885199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577033414 Chr.16: 88885071-88885071 on Build GRCh38
rs900342897 Chr.16: 88885005-88885005 on Build GRCh38
rs772958174 Chr.16: 88885274-88885274 on Build GRCh38
rs375093204 Chr.16: 88885080-88885080 on Build GRCh38
rs780369396 Chr.16: 88885068-88885068 on Build GRCh38
rs747925300 Chr.16: 88885315-88885315 on Build GRCh38
rs1050208810 Chr.16: 88885170-88885170 on Build GRCh38
rs776682768 Chr.16: 88885136-88885136 on Build GRCh38
rs903080518 Chr.16: 88885385-88885385 on Build GRCh38
rs767127378 Chr.16: 88885213-88885213 on Build GRCh38
rs568625819 Chr.16: 88885245-88885245 on Build GRCh38
rs775449176 Chr.16: 88885252-88885252 on Build GRCh38
rs556767136 Chr.16: 88885030-88885030 on Build GRCh38
rs141390218 Chr.16: 88885257-88885257 on Build GRCh38
rs761363984 Chr.16: 88885212-88885212 on Build GRCh38
rs529742127 Chr.16: 88885135-88885135 on Build GRCh38
rs544130326 Chr.16: 88885307-88885307 on Build GRCh38
rs536003077 Chr.16: 88885254-88885254 on Build GRCh38
rs967479156 Chr.16: 88885002-88885002 on Build GRCh38
rs957469989 Chr.16: 88885111-88885111 on Build GRCh38
rs756838631 Chr.16: 88885153-88885153 on Build GRCh38
rs376612728 Chr.16: 88885020-88885020 on Build GRCh38
rs763161275 Chr.16: 88885027-88885027 on Build GRCh38
rs758648207 Chr.16: 88885165-88885165 on Build GRCh38
rs372751882 Chr.16: 88885066-88885066 on Build GRCh38
rs945863791 Chr.16: 88885391-88885391 on Build GRCh38
rs181515376 Chr.16: 88885091-88885091 on Build GRCh38
rs1040467990 Chr.16: 88885174-88885174 on Build GRCh38
rs940310677 Chr.16: 88885300-88885300 on Build GRCh38
rs144606503 Chr.16: 88885054-88885054 on Build GRCh38
rs766921779 Chr.16: 88885156-88885156 on Build GRCh38
rs764337817 Chr.16: 88884998-88884998 on Build GRCh38
rs763830719 Chr.16: 88885147-88885147 on Build GRCh38
rs745881689 Chr.16: 88885187-88885187 on Build GRCh38
rs769740338 Chr.16: 88885195-88885195 on Build GRCh38
rs753677492 Chr.16: 88885043-88885043 on Build GRCh38
rs61757659 Chr.16: 88885186-88885186 on Build GRCh38
rs747324207 Chr.16: 88885244-88885244 on Build GRCh38
rs544407834 Chr.16: 88885075-88885075 on Build GRCh38
rs898903047 Chr.16: 88885154-88885154 on Build GRCh38
rs758046079 Chr.16: 88885114-88885114 on Build GRCh38
rs140507946 Chr.16: 88885121-88885121 on Build GRCh38
rs767591258 Chr.16: 88885011-88885011 on Build GRCh38
rs754459357 Chr.16: 88885159-88885159 on Build GRCh38
rs527370233 Chr.16: 88885371-88885371 on Build GRCh38
rs146630443 Chr.16: 88885240-88885240 on Build GRCh38
rs147184929 Chr.16: 88885061-88885061 on Build GRCh38
rs368682995 Chr.16: 88885021-88885021 on Build GRCh38
rs778476777 Chr.16: 88885015-88885015 on Build GRCh38
rs959623042 Chr.16: 88885267-88885267 on Build GRCh38
rs749788456 Chr.16: 88885125-88885125 on Build GRCh38
rs370969451 Chr.16: 88885309-88885309 on Build GRCh38
rs564040614 Chr.16: 88885203-88885203 on Build GRCh38
rs753222808 Chr.16: 88885233-88885233 on Build GRCh38
rs369844657 Chr.16: 88885034-88885034 on Build GRCh38
rs761606756 Chr.16: 88885263-88885263 on Build GRCh38
rs143983197 Chr.16: 88885292-88885292 on Build GRCh38
rs200768874 Chr.16: 88885033-88885033 on Build GRCh38
rs113270439 Chr.16: 88884985-88884985 on Build GRCh38
rs944549314 Chr.16: 88885379-88885379 on Build GRCh38
rs527263791 Chr.16: 88885162-88885162 on Build GRCh38
rs117264176 Chr.16: 88884990-88884990 on Build GRCh38
rs147179125 Chr.16: 88885321-88885321 on Build GRCh38
rs142092792 Chr.16: 88885143-88885143 on Build GRCh38
rs531010385 Chr.16: 88885302-88885302 on Build GRCh38
rs773342006 Chr.16: 88885088-88885088 on Build GRCh38
rs377342150 Chr.16: 88885040-88885040 on Build GRCh38
rs921612876 Chr.16: 88885336-88885336 on Build GRCh38
rs151180006 Chr.16: 88885144-88885144 on Build GRCh38
rs748453376 Chr.16: 88885128-88885128 on Build GRCh38
rs751133483 Chr.16: 88885149-88885149 on Build GRCh38
rs375044493 Chr.16: 88885031-88885031 on Build GRCh38
rs888923563 Chr.16: 88885374-88885374 on Build GRCh38
rs762057993 Chr.16: 88885095-88885095 on Build GRCh38
rs931883414 Chr.16: 88885345-88885345 on Build GRCh38
rs541558294 Chr.16: 88885328-88885328 on Build GRCh38
rs772340059 Chr.16: 88885230-88885230 on Build GRCh38
rs573721979 Chr.16: 88885239-88885239 on Build GRCh38
rs367560755 Chr.16: 88885247-88885247 on Build GRCh38
rs190053493 Chr.16: 88885012-88885012 on Build GRCh38
rs185715659 Chr.16: 88885229-88885229 on Build GRCh38
rs751884253 Chr.16: 88885289-88885289 on Build GRCh38
rs200106739 Chr.16: 88885035-88885035 on Build GRCh38
rs559413604 Chr.16: 88884951-88884951 on Build GRCh38
rs74764702 Chr.16: 88885276-88885276 on Build GRCh38
rs570992759 Chr.16: 88885014-88885014 on Build GRCh38
rs753676354 Chr.16: 88885093-88885093 on Build GRCh38
rs1040285764 Chr.16: 88885083-88885083 on Build GRCh38
rs574124916 Chr.16: 88885026-88885026 on Build GRCh38
rs770118907 Chr.16: 88885025-88885025 on Build GRCh38
rs763767844 Chr.16: 88885207-88885207 on Build GRCh38
rs769199782 Chr.16: 88885126-88885126 on Build GRCh38
rs745972870 Chr.16: 88885246-88885246 on Build GRCh38
rs759369853 Chr.16: 88885100-88885100 on Build GRCh38
rs373798911 Chr.16: 88885161-88885161 on Build GRCh38
rs760610315 Chr.16: 88885090-88885090 on Build GRCh38
rs777224110 Chr.16: 88885115-88885115 on Build GRCh38
rs139030498 Chr.16: 88885164-88885164 on Build GRCh38
rs750615296 Chr.16: 88885293-88885293 on Build GRCh38
rs866781288 Chr.16: 88885109-88885109 on Build GRCh38
rs529704180 Chr.16: 88884952-88884952 on Build GRCh38
rs953292167 Chr.16: 88885069-88885069 on Build GRCh38
rs555987218 Chr.16: 88885318-88885318 on Build GRCh38
rs763366195 Chr.16: 88885280-88885280 on Build GRCh38
rs940365095 Chr.16: 88885150-88885150 on Build GRCh38
rs545799996 Chr.16: 88885381-88885381 on Build GRCh38
rs779415412 Chr.16: 88885127-88885127 on Build GRCh38
rs534269815 Chr.16: 88884995-88884995 on Build GRCh38
rs767300643 Chr.16: 88885273-88885273 on Build GRCh38
rs953326394 Chr.16: 88884988-88884988 on Build GRCh38
rs201822501 Chr.16: 88885301-88885301 on Build GRCh38
rs775361242 Chr.16: 88885198-88885198 on Build GRCh38
rs1040235752 Chr.16: 88885383-88885383 on Build GRCh38
rs757357010 Chr.16: 88885003-88885003 on Build GRCh38
rs377182425 Chr.16: 88885146-88885146 on Build GRCh38
rs764998500 Chr.16: 88885102-88885102 on Build GRCh38
rs559794551 Chr.16: 88885330-88885330 on Build GRCh38
rs773255128 Chr.16: 88885132-88885132 on Build GRCh38
rs765875911 Chr.16: 88885232-88885232 on Build GRCh38
rs143614023 Chr.16: 88885058-88885058 on Build GRCh38
rs377163386 Chr.16: 88885076-88885076 on Build GRCh38
rs1008773759 Chr.16: 88885191-88885191 on Build GRCh38
rs751095715 Chr.16: 88885119-88885119 on Build GRCh38
rs994690990 Chr.16: 88884957-88884957 on Build GRCh38
rs150879448 Chr.16: 88885055-88885055 on Build GRCh38
rs368765045 Chr.16: 88885133-88885133 on Build GRCh38
rs774109097 Chr.16: 88885208-88885208 on Build GRCh38
rs914528701 Chr.16: 88885390-88885390 on Build GRCh38
rs77867180 Chr.16: 88885036-88885036 on Build GRCh38
rs752408681 Chr.16: 88885108-88885108 on Build GRCh38
rs370722363 Chr.16: 88885050-88885050 on Build GRCh38
rs772349219 Chr.16: 88885131-88885131 on Build GRCh38
rs926101740 Chr.16: 88885006-88885006 on Build GRCh38
rs367992838 Chr.16: 88885298-88885298 on Build GRCh38
rs1035443089 Chr.16: 88885262-88885262 on Build GRCh38
rs753316173 Chr.16: 88885279-88885279 on Build GRCh38
rs746367268 Chr.16: 88885022-88885022 on Build GRCh38
rs778860303 Chr.16: 88885314-88885314 on Build GRCh38
rs374540349 Chr.16: 88884958-88884958 on Build GRCh38
rs747177846 Chr.16: 88885182-88885182 on Build GRCh38
rs755909799 Chr.16: 88885013-88885013 on Build GRCh38
rs201266682 Chr.16: 88885256-88885256 on Build GRCh38
rs778095408 Chr.16: 88885169-88885169 on Build GRCh38
rs757597690 Chr.16: 88885290-88885290 on Build GRCh38
rs764596018 Chr.16: 88885286-88885286 on Build GRCh38
rs911917927 Chr.16: 88884992-88884992 on Build GRCh38
rs751694574 Chr.16: 88885000-88885000 on Build GRCh38
rs772345679 Chr.16: 88885086-88885086 on Build GRCh38

View Full Product Details