Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89738254-89738766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCA
Entrez Gene ID
2175
Gene Name
Fanconi anemia complementation group A
Gene Aliases
FA, FA-H, FA1, FAA, FACA, FAH, FANCH
UniGene
Hs.744083
Gene Chromosome Location
Chr.16: 89737551-89816658 on Build GRCh38
Transcripts
NM_000135.2 NM_001286167.1 XM_005256294.4 XM_011522945.2 XM_011522946.2 XM_011522947.2 XM_017023044.1 NM_001113525.1 NM_152287.3 NR_110122.1 NR_110126.1 NR_110128.1 NR_110129.1 XM_005256324.3 XM_005256328.3 XM_017023890.1

Gene Symbol
ZNF276
Entrez Gene ID
92822
Gene Name
zinc finger protein 276
Gene Aliases
CENP-Z, CENPZ, ZADT, ZFP276, ZNF477
UniGene
Hs.290154
Gene Chromosome Location
Chr.16: 89720368-89740925 on Build GRCh38
Transcripts
NM_000135.2 NM_001286167.1 XM_005256294.4 XM_011522945.2 XM_011522946.2 XM_011522947.2 XM_017023044.1 NM_001113525.1 NM_152287.3 NR_110122.1 NR_110126.1 NR_110128.1 NR_110129.1 XM_005256324.3 XM_005256328.3 XM_017023890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4657336 c.4284C>T p.C1428C Chr.16: 89738685-89738685 on Build GRCh38
6934824 c.4303G>A p.A1435T Chr.16: 89738666-89738666 on Build GRCh38
4593520 c.4307T>C p.L1436P Chr.16: 89738662-89738662 on Build GRCh38
4444562 c.4274G>A p.R1425H Chr.16: 89738695-89738695 on Build GRCh38
6021484 c.4357C>T p.H1453Y Chr.16: 89738612-89738612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991918629 Chr.16: 89738380-89738380 on Build GRCh38
rs587778322 Chr.16: 89738653-89738653 on Build GRCh38
rs186497529 Chr.16: 89738459-89738459 on Build GRCh38
rs147195004 Chr.16: 89738486-89738486 on Build GRCh38
rs746069040 Chr.16: 89738498-89738500 on Build GRCh38
rs908909494 Chr.16: 89738285-89738285 on Build GRCh38
rs767234774 Chr.16: 89738651-89738651 on Build GRCh38
rs897557830 Chr.16: 89738614-89738614 on Build GRCh38
rs749315540 Chr.16: 89738619-89738619 on Build GRCh38
rs751400000 Chr.16: 89738597-89738597 on Build GRCh38
rs529748804 Chr.16: 89738559-89738559 on Build GRCh38
rs575606938 Chr.16: 89738457-89738457 on Build GRCh38
rs765925445 Chr.16: 89738742-89738743 on Build GRCh38
rs915983602 Chr.16: 89738710-89738710 on Build GRCh38
rs887428503 Chr.16: 89738448-89738448 on Build GRCh38
rs752576350 Chr.16: 89738601-89738601 on Build GRCh38
rs144171225 Chr.16: 89738644-89738644 on Build GRCh38
rs1014701946 Chr.16: 89738323-89738323 on Build GRCh38
rs141761812 Chr.16: 89738454-89738454 on Build GRCh38
rs374830555 Chr.16: 89738634-89738634 on Build GRCh38
rs533812797 Chr.16: 89738446-89738446 on Build GRCh38
rs190645652 Chr.16: 89738567-89738567 on Build GRCh38
rs757549007 Chr.16: 89738620-89738620 on Build GRCh38
rs923174934 Chr.16: 89738327-89738327 on Build GRCh38
rs948918065 Chr.16: 89738548-89738548 on Build GRCh38
rs772736966 Chr.16: 89738568-89738568 on Build GRCh38
rs371859584 Chr.16: 89738731-89738731 on Build GRCh38
rs772589360 Chr.16: 89738595-89738595 on Build GRCh38
rs748269284 Chr.16: 89738288-89738288 on Build GRCh38
rs368506826 Chr.16: 89738717-89738717 on Build GRCh38
rs544435184 Chr.16: 89738547-89738547 on Build GRCh38
rs1047662111 Chr.16: 89738668-89738668 on Build GRCh38
rs914493960 Chr.16: 89738478-89738478 on Build GRCh38
rs773999989 Chr.16: 89738690-89738690 on Build GRCh38
rs528787665 Chr.16: 89738324-89738324 on Build GRCh38
rs761241462 Chr.16: 89738581-89738581 on Build GRCh38
rs1057480360 Chr.16: 89738516-89738516 on Build GRCh38
rs147406377 Chr.16: 89738695-89738695 on Build GRCh38
rs776090306 Chr.16: 89738578-89738578 on Build GRCh38
rs1030186205 Chr.16: 89738462-89738462 on Build GRCh38
rs111698147 Chr.16: 89738714-89738714 on Build GRCh38
rs17227417 Chr.16: 89738314-89738314 on Build GRCh38
rs1023324062 Chr.16: 89738299-89738299 on Build GRCh38
rs754732247 Chr.16: 89738599-89738599 on Build GRCh38
rs765478990 Chr.16: 89738711-89738711 on Build GRCh38
rs371228419 Chr.16: 89738640-89738640 on Build GRCh38
rs753660236 Chr.16: 89738705-89738705 on Build GRCh38
rs761987751 Chr.16: 89738707-89738707 on Build GRCh38
rs142784426 Chr.16: 89738667-89738667 on Build GRCh38
rs56346328 Chr.16: 89738292-89738292 on Build GRCh38
rs372268907 Chr.16: 89738716-89738716 on Build GRCh38
rs984981073 Chr.16: 89738427-89738427 on Build GRCh38
rs964506692 Chr.16: 89738527-89738527 on Build GRCh38
rs557319516 Chr.16: 89738351-89738351 on Build GRCh38
rs781731674 Chr.16: 89738726-89738726 on Build GRCh38
rs769025633 Chr.16: 89738636-89738636 on Build GRCh38
rs577149121 Chr.16: 89738429-89738429 on Build GRCh38
rs17233783 Chr.16: 89738639-89738639 on Build GRCh38
rs916052051 Chr.16: 89738545-89738545 on Build GRCh38
rs17233804 Chr.16: 89738315-89738315 on Build GRCh38
rs748856769 Chr.16: 89738684-89738684 on Build GRCh38
rs760722877 Chr.16: 89738660-89738660 on Build GRCh38
rs374984587 Chr.16: 89738669-89738669 on Build GRCh38
rs747310276 Chr.16: 89738671-89738671 on Build GRCh38
rs1030509993 Chr.16: 89738637-89738638 on Build GRCh38
rs938807623 Chr.16: 89738501-89738501 on Build GRCh38
rs760732305 Chr.16: 89738522-89738522 on Build GRCh38
rs112177258 Chr.16: 89738293-89738293 on Build GRCh38
rs750442764 Chr.16: 89738713-89738716 on Build GRCh38
rs1005953047 Chr.16: 89738500-89738500 on Build GRCh38
rs199937973 Chr.16: 89738661-89738661 on Build GRCh38
rs903277788 Chr.16: 89738382-89738382 on Build GRCh38
rs181842476 Chr.16: 89738436-89738436 on Build GRCh38
rs541302571 Chr.16: 89738741-89738741 on Build GRCh38
rs554999842 Chr.16: 89738465-89738465 on Build GRCh38
rs950493395 Chr.16: 89738418-89738418 on Build GRCh38
rs55679217 Chr.16: 89738492-89738492 on Build GRCh38
rs779236651 Chr.16: 89738623-89738623 on Build GRCh38
rs765072800 Chr.16: 89738668-89738669 on Build GRCh38
rs370357192 Chr.16: 89738600-89738600 on Build GRCh38
rs1041759070 Chr.16: 89738367-89738367 on Build GRCh38
rs370868774 Chr.16: 89738297-89738297 on Build GRCh38
rs74977201 Chr.16: 89738666-89738666 on Build GRCh38
rs556325258 Chr.16: 89738455-89738455 on Build GRCh38
rs201817705 Chr.16: 89738284-89738284 on Build GRCh38
rs145495509 Chr.16: 89738551-89738551 on Build GRCh38
rs922937790 Chr.16: 89738553-89738553 on Build GRCh38
rs368670194 Chr.16: 89738558-89738558 on Build GRCh38
rs367630825 Chr.16: 89738685-89738685 on Build GRCh38
rs905886601 Chr.16: 89738338-89738338 on Build GRCh38
rs568833102 Chr.16: 89738428-89738428 on Build GRCh38
rs17233797 Chr.16: 89738349-89738349 on Build GRCh38
rs749463081 Chr.16: 89738295-89738295 on Build GRCh38
rs3743863 Chr.16: 89738694-89738694 on Build GRCh38
rs764113707 Chr.16: 89738700-89738700 on Build GRCh38
rs568293451 Chr.16: 89738670-89738670 on Build GRCh38
rs746081933 Chr.16: 89738713-89738714 on Build GRCh38
rs56049032 Chr.16: 89738352-89738352 on Build GRCh38
rs751989527 Chr.16: 89738720-89738720 on Build GRCh38
rs1043241541 Chr.16: 89738618-89738618 on Build GRCh38
rs746256466 Chr.16: 89738631-89738631 on Build GRCh38
rs536287114 Chr.16: 89738498-89738498 on Build GRCh38
rs566175597 Chr.16: 89738437-89738437 on Build GRCh38
rs764735826 Chr.16: 89738587-89738587 on Build GRCh38
rs1021626760 Chr.16: 89738333-89738333 on Build GRCh38
rs886052479 Chr.16: 89738435-89738435 on Build GRCh38
rs551173588 Chr.16: 89738383-89738383 on Build GRCh38
rs975923448 Chr.16: 89738326-89738326 on Build GRCh38
rs573302337 Chr.16: 89738470-89738470 on Build GRCh38
rs775549018 Chr.16: 89738463-89738463 on Build GRCh38
rs149531696 Chr.16: 89738637-89738637 on Build GRCh38
rs554525485 Chr.16: 89738560-89738560 on Build GRCh38
rs756173179 Chr.16: 89738290-89738290 on Build GRCh38
rs1057067682 Chr.16: 89738461-89738461 on Build GRCh38
rs777977318 Chr.16: 89738610-89738610 on Build GRCh38
rs375487632 Chr.16: 89738742-89738742 on Build GRCh38
rs199675569 Chr.16: 89738596-89738596 on Build GRCh38
rs772505725 Chr.16: 89738633-89738633 on Build GRCh38
rs139532591 Chr.16: 89738698-89738698 on Build GRCh38
rs761743531 Chr.16: 89738628-89738633 on Build GRCh38
rs1026096277 Chr.16: 89738494-89738494 on Build GRCh38
rs778452514 Chr.16: 89738740-89738740 on Build GRCh38
rs1230 Chr.16: 89738447-89738447 on Build GRCh38
rs587778321 Chr.16: 89738696-89738696 on Build GRCh38
rs776321328 Chr.16: 89738578-89738578 on Build GRCh38
rs190762834 Chr.16: 89738370-89738370 on Build GRCh38
rs770282073 Chr.16: 89738737-89738737 on Build GRCh38
rs769243354 Chr.16: 89738675-89738675 on Build GRCh38
rs968652405 Chr.16: 89738483-89738483 on Build GRCh38
rs759343346 Chr.16: 89738647-89738647 on Build GRCh38
rs931959948 Chr.16: 89738332-89738333 on Build GRCh38
rs55859244 Chr.16: 89738497-89738497 on Build GRCh38
rs947517919 Chr.16: 89738617-89738617 on Build GRCh38
rs1012078925 Chr.16: 89738325-89738325 on Build GRCh38
rs923490846 Chr.16: 89738440-89738440 on Build GRCh38
rs570739902 Chr.16: 89738453-89738453 on Build GRCh38
rs937443966 Chr.16: 89738648-89738648 on Build GRCh38
rs974454406 Chr.16: 89738439-89738439 on Build GRCh38
rs892010151 Chr.16: 89738448-89738448 on Build GRCh38
rs1013197020 Chr.16: 89738445-89738445 on Build GRCh38
rs200757877 Chr.16: 89738287-89738287 on Build GRCh38

View Full Product Details