Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89738594-89739086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCA
Entrez Gene ID
2175
Gene Name
Fanconi anemia complementation group A
Gene Aliases
FA, FA-H, FA1, FAA, FACA, FAH, FANCH
UniGene
Hs.744083
Gene Chromosome Location
Chr.16: 89737551-89816658 on Build GRCh38
Transcripts
NM_000135.2 NM_001286167.1 XM_005256294.4 XM_011522945.2 XM_011522946.2 XM_011522947.2 XM_017023044.1 NM_001113525.1 NM_152287.3 NR_110122.1 NR_110126.1 NR_110128.1 NR_110129.1 XM_017023890.1

Gene Symbol
ZNF276
Entrez Gene ID
92822
Gene Name
zinc finger protein 276
Gene Aliases
CENP-Z, CENPZ, ZADT, ZFP276, ZNF477
UniGene
Hs.290154
Gene Chromosome Location
Chr.16: 89720368-89740925 on Build GRCh38
Transcripts
NM_000135.2 NM_001286167.1 XM_005256294.4 XM_011522945.2 XM_011522946.2 XM_011522947.2 XM_017023044.1 NM_001113525.1 NM_152287.3 NR_110122.1 NR_110126.1 NR_110128.1 NR_110129.1 XM_017023890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4593520 c.4307T>C p.L1436P Chr.16: 89738662-89738662 on Build GRCh38
6987547 c.4258G>T p.E1420* Chr.16: 89738884-89738884 on Build GRCh38
6021484 c.4357C>T p.H1453Y Chr.16: 89738612-89738612 on Build GRCh38
6958638 c.4331_4332delCT p.P1444fs*3 Chr.16: 89738637-89738638 on Build GRCh38
2923602 c.4299C>T p.S1433S Chr.16: 89738670-89738670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763863651 Chr.16: 89738981-89738981 on Build GRCh38
rs187792766 Chr.16: 89739060-89739060 on Build GRCh38
rs587778322 Chr.16: 89738653-89738653 on Build GRCh38
rs767234774 Chr.16: 89738651-89738651 on Build GRCh38
rs754274108 Chr.16: 89738843-89738843 on Build GRCh38
rs776860862 Chr.16: 89738966-89738966 on Build GRCh38
rs749315540 Chr.16: 89738619-89738619 on Build GRCh38
rs149851163 Chr.16: 89738943-89738943 on Build GRCh38
rs756853923 Chr.16: 89738986-89738986 on Build GRCh38
rs1010028646 Chr.16: 89738995-89738995 on Build GRCh38
rs559541393 Chr.16: 89738784-89738784 on Build GRCh38
rs775643194 Chr.16: 89738867-89738867 on Build GRCh38
rs1800359 Chr.16: 89738853-89738853 on Build GRCh38
rs765925445 Chr.16: 89738742-89738743 on Build GRCh38
rs915983602 Chr.16: 89738710-89738710 on Build GRCh38
rs144171225 Chr.16: 89738644-89738644 on Build GRCh38
rs754622518 Chr.16: 89738848-89738848 on Build GRCh38
rs756798736 Chr.16: 89738771-89738771 on Build GRCh38
rs750161523 Chr.16: 89738993-89738993 on Build GRCh38
rs374830555 Chr.16: 89738634-89738634 on Build GRCh38
rs757549007 Chr.16: 89738620-89738620 on Build GRCh38
rs760223551 Chr.16: 89738980-89738980 on Build GRCh38
rs774344839 Chr.16: 89738855-89738855 on Build GRCh38
rs747917354 Chr.16: 89738819-89738819 on Build GRCh38
rs765239590 Chr.16: 89738774-89738774 on Build GRCh38
rs371859584 Chr.16: 89738731-89738731 on Build GRCh38
rs984178125 Chr.16: 89739052-89739052 on Build GRCh38
rs745958661 Chr.16: 89738900-89738900 on Build GRCh38
rs573170442 Chr.16: 89739009-89739009 on Build GRCh38
rs749574677 Chr.16: 89738947-89738947 on Build GRCh38
rs773238151 Chr.16: 89738823-89738823 on Build GRCh38
rs781273405 Chr.16: 89739015-89739015 on Build GRCh38
rs776480113 Chr.16: 89738759-89738759 on Build GRCh38
rs368506826 Chr.16: 89738717-89738717 on Build GRCh38
rs1047662111 Chr.16: 89738668-89738668 on Build GRCh38
rs755883570 Chr.16: 89738933-89738933 on Build GRCh38
rs753103430 Chr.16: 89738839-89738839 on Build GRCh38
rs773999989 Chr.16: 89738690-89738690 on Build GRCh38
rs769897335 Chr.16: 89739018-89739018 on Build GRCh38
rs763083176 Chr.16: 89738783-89738783 on Build GRCh38
rs761896034 Chr.16: 89738760-89738760 on Build GRCh38
rs201494304 Chr.16: 89738910-89738910 on Build GRCh38
rs370507983 Chr.16: 89738916-89738916 on Build GRCh38
rs558916721 Chr.16: 89738748-89738748 on Build GRCh38
rs768842673 Chr.16: 89738907-89738907 on Build GRCh38
rs780156946 Chr.16: 89738959-89738959 on Build GRCh38
rs375508301 Chr.16: 89739059-89739059 on Build GRCh38
rs147406377 Chr.16: 89738695-89738695 on Build GRCh38
rs574812539 Chr.16: 89739058-89739058 on Build GRCh38
rs768340256 Chr.16: 89738810-89738810 on Build GRCh38
rs111698147 Chr.16: 89738714-89738714 on Build GRCh38
rs772122405 Chr.16: 89738901-89738901 on Build GRCh38
rs763142032 Chr.16: 89738824-89738824 on Build GRCh38
rs765478990 Chr.16: 89738711-89738711 on Build GRCh38
rs747975371 Chr.16: 89738852-89738852 on Build GRCh38
rs371228419 Chr.16: 89738640-89738640 on Build GRCh38
rs577194734 Chr.16: 89738974-89738974 on Build GRCh38
rs753660236 Chr.16: 89738705-89738705 on Build GRCh38
rs556778941 Chr.16: 89739040-89739040 on Build GRCh38
rs142784426 Chr.16: 89738667-89738667 on Build GRCh38
rs761987751 Chr.16: 89738707-89738707 on Build GRCh38
rs753048298 Chr.16: 89738790-89738790 on Build GRCh38
rs754990688 Chr.16: 89739011-89739011 on Build GRCh38
rs751059431 Chr.16: 89738886-89738886 on Build GRCh38
rs765136708 Chr.16: 89738987-89738987 on Build GRCh38
rs544934389 Chr.16: 89738785-89738785 on Build GRCh38
rs774951303 Chr.16: 89739037-89739037 on Build GRCh38
rs372268907 Chr.16: 89738716-89738716 on Build GRCh38
rs750141209 Chr.16: 89738789-89738789 on Build GRCh38
rs200284171 Chr.16: 89738888-89738888 on Build GRCh38
rs753475025 Chr.16: 89738985-89738985 on Build GRCh38
rs984285795 Chr.16: 89738884-89738884 on Build GRCh38
rs781731674 Chr.16: 89738726-89738726 on Build GRCh38
rs566927763 Chr.16: 89738941-89738941 on Build GRCh38
rs769025633 Chr.16: 89738636-89738636 on Build GRCh38
rs770892107 Chr.16: 89738854-89738854 on Build GRCh38
rs759735114 Chr.16: 89738833-89738833 on Build GRCh38
rs768269080 Chr.16: 89738753-89738753 on Build GRCh38
rs17233783 Chr.16: 89738639-89738639 on Build GRCh38
rs748856769 Chr.16: 89738684-89738684 on Build GRCh38
rs969373089 Chr.16: 89738874-89738874 on Build GRCh38
rs754170870 Chr.16: 89738935-89738936 on Build GRCh38
rs761574058 Chr.16: 89739045-89739045 on Build GRCh38
rs773679037 Chr.16: 89739020-89739020 on Build GRCh38
rs779280925 Chr.16: 89739014-89739017 on Build GRCh38
rs760722877 Chr.16: 89738660-89738660 on Build GRCh38
rs374984587 Chr.16: 89738669-89738669 on Build GRCh38
rs1030509993 Chr.16: 89738637-89738638 on Build GRCh38
rs747310276 Chr.16: 89738671-89738671 on Build GRCh38
rs747111023 Chr.16: 89739010-89739010 on Build GRCh38
rs374793201 Chr.16: 89738876-89738876 on Build GRCh38
rs750442764 Chr.16: 89738713-89738716 on Build GRCh38
rs199937973 Chr.16: 89738661-89738661 on Build GRCh38
rs767206558 Chr.16: 89738920-89738920 on Build GRCh38
rs913443728 Chr.16: 89738787-89738787 on Build GRCh38
rs541302571 Chr.16: 89738741-89738741 on Build GRCh38
rs746415530 Chr.16: 89739051-89739051 on Build GRCh38
rs1020362407 Chr.16: 89739002-89739002 on Build GRCh38
rs775180524 Chr.16: 89738978-89738978 on Build GRCh38
rs779236651 Chr.16: 89738623-89738623 on Build GRCh38
rs371280585 Chr.16: 89739025-89739025 on Build GRCh38
rs765072800 Chr.16: 89738668-89738669 on Build GRCh38
rs770127493 Chr.16: 89738967-89738967 on Build GRCh38
rs759630635 Chr.16: 89738857-89738857 on Build GRCh38
rs752333498 Chr.16: 89738891-89738891 on Build GRCh38
rs748472421 Chr.16: 89739017-89739017 on Build GRCh38
rs74977201 Chr.16: 89738666-89738666 on Build GRCh38
rs367630825 Chr.16: 89738685-89738685 on Build GRCh38
rs746832960 Chr.16: 89738751-89738751 on Build GRCh38
rs111846044 Chr.16: 89738842-89738842 on Build GRCh38
rs562917320 Chr.16: 89738794-89738794 on Build GRCh38
rs3743863 Chr.16: 89738694-89738694 on Build GRCh38
rs764113707 Chr.16: 89738700-89738700 on Build GRCh38
rs568293451 Chr.16: 89738670-89738670 on Build GRCh38
rs769835904 Chr.16: 89738927-89738927 on Build GRCh38
rs778136548 Chr.16: 89738923-89738923 on Build GRCh38
rs751166862 Chr.16: 89738768-89738783 on Build GRCh38
rs34218545 Chr.16: 89739059-89739059 on Build GRCh38
rs755531081 Chr.16: 89738850-89738850 on Build GRCh38
rs775541226 Chr.16: 89738903-89738903 on Build GRCh38
rs55873203 Chr.16: 89739013-89739014 on Build GRCh38
rs145148206 Chr.16: 89738890-89738890 on Build GRCh38
rs746081933 Chr.16: 89738713-89738714 on Build GRCh38
rs751989527 Chr.16: 89738720-89738720 on Build GRCh38
rs1043241541 Chr.16: 89738618-89738618 on Build GRCh38
rs368985041 Chr.16: 89738909-89738909 on Build GRCh38
rs1019949093 Chr.16: 89738862-89738862 on Build GRCh38
rs745882980 Chr.16: 89738944-89738944 on Build GRCh38
rs746256466 Chr.16: 89738631-89738631 on Build GRCh38
rs139478274 Chr.16: 89738917-89738917 on Build GRCh38
rs940187828 Chr.16: 89738895-89738895 on Build GRCh38
rs995915107 Chr.16: 89738847-89738847 on Build GRCh38
rs553145380 Chr.16: 89739064-89739064 on Build GRCh38
rs199652831 Chr.16: 89738965-89738965 on Build GRCh38
rs772216317 Chr.16: 89738866-89738866 on Build GRCh38
rs773463465 Chr.16: 89738775-89738775 on Build GRCh38
rs754660451 Chr.16: 89738806-89738809 on Build GRCh38
rs149531696 Chr.16: 89738637-89738637 on Build GRCh38
rs757777876 Chr.16: 89738846-89738846 on Build GRCh38
rs1023409042 Chr.16: 89739029-89739029 on Build GRCh38
rs201658945 Chr.16: 89738898-89738898 on Build GRCh38
rs375487632 Chr.16: 89738742-89738742 on Build GRCh38
rs775989654 Chr.16: 89739043-89739043 on Build GRCh38
rs772505725 Chr.16: 89738633-89738633 on Build GRCh38
rs558672797 Chr.16: 89738957-89738957 on Build GRCh38
rs752245802 Chr.16: 89738924-89738924 on Build GRCh38
rs139532591 Chr.16: 89738698-89738698 on Build GRCh38
rs17227403 Chr.16: 89738893-89738893 on Build GRCh38
rs376361490 Chr.16: 89739007-89739007 on Build GRCh38
rs201205004 Chr.16: 89738746-89738746 on Build GRCh38
rs747475688 Chr.16: 89738828-89738828 on Build GRCh38
rs761743531 Chr.16: 89738628-89738633 on Build GRCh38
rs767757264 Chr.16: 89738835-89738835 on Build GRCh38
rs778452514 Chr.16: 89738740-89738740 on Build GRCh38
rs559788532 Chr.16: 89738807-89738807 on Build GRCh38
rs587778321 Chr.16: 89738696-89738696 on Build GRCh38
rs760945805 Chr.16: 89738906-89738906 on Build GRCh38
rs770282073 Chr.16: 89738737-89738737 on Build GRCh38
rs776335845 Chr.16: 89738816-89738816 on Build GRCh38
rs769243354 Chr.16: 89738675-89738675 on Build GRCh38
rs574723272 Chr.16: 89738798-89738798 on Build GRCh38
rs538139633 Chr.16: 89739031-89739031 on Build GRCh38
rs765683250 Chr.16: 89738913-89738913 on Build GRCh38
rs780438035 Chr.16: 89738820-89738820 on Build GRCh38
rs763809014 Chr.16: 89738935-89738935 on Build GRCh38
rs34602467 Chr.16: 89738802-89738802 on Build GRCh38
rs759343346 Chr.16: 89738647-89738647 on Build GRCh38
rs564148401 Chr.16: 89738892-89738892 on Build GRCh38
rs947517919 Chr.16: 89738617-89738617 on Build GRCh38
rs1047071335 Chr.16: 89738831-89738831 on Build GRCh38
rs771957724 Chr.16: 89739013-89739013 on Build GRCh38
rs937443966 Chr.16: 89738648-89738648 on Build GRCh38
rs756431845 Chr.16: 89738841-89738841 on Build GRCh38
rs774488518 Chr.16: 89738829-89738829 on Build GRCh38
rs760276584 Chr.16: 89739039-89739039 on Build GRCh38

View Full Product Details