Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89738988-89739499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCA
Entrez Gene ID
2175
Gene Name
Fanconi anemia complementation group A
Gene Aliases
FA, FA-H, FA1, FAA, FACA, FAH, FANCH
UniGene
Hs.744083
Gene Chromosome Location
Chr.16: 89737551-89816658 on Build GRCh38
Transcripts
NM_000135.2 NM_001286167.1 XM_005256294.4 XM_011522945.2 XM_011522946.2 XM_011522947.2 XM_017023044.1 NM_001113525.1 NM_152287.3 NR_110122.1 NR_110126.1 NR_110128.1 NR_110129.1 XM_017023890.1

Gene Symbol
ZNF276
Entrez Gene ID
92822
Gene Name
zinc finger protein 276
Gene Aliases
CENP-Z, CENPZ, ZADT, ZFP276, ZNF477
UniGene
Hs.290154
Gene Chromosome Location
Chr.16: 89720368-89740925 on Build GRCh38
Transcripts
NM_000135.2 NM_001286167.1 XM_005256294.4 XM_011522945.2 XM_011522946.2 XM_011522947.2 XM_017023044.1 NM_001113525.1 NM_152287.3 NR_110122.1 NR_110126.1 NR_110128.1 NR_110129.1 XM_017023890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6953813 c.4114G>A p.D1372N Chr.16: 89739186-89739186 on Build GRCh38
6566705 c.4000G>A p.A1334T Chr.16: 89739488-89739488 on Build GRCh38
4129524 c.4115A>G p.D1372G Chr.16: 89739185-89739185 on Build GRCh38
5575680 c.4097A>G p.Q1366R Chr.16: 89739203-89739203 on Build GRCh38
6924997 c.4037C>T p.A1346V Chr.16: 89739263-89739263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187792766 Chr.16: 89739060-89739060 on Build GRCh38
rs372493612 Chr.16: 89739165-89739165 on Build GRCh38
rs1014447679 Chr.16: 89739471-89739471 on Build GRCh38
rs201886956 Chr.16: 89739263-89739263 on Build GRCh38
rs367970303 Chr.16: 89739155-89739155 on Build GRCh38
rs774175580 Chr.16: 89739152-89739152 on Build GRCh38
rs775186188 Chr.16: 89739274-89739274 on Build GRCh38
rs74033848 Chr.16: 89739311-89739311 on Build GRCh38
rs764090579 Chr.16: 89739229-89739229 on Build GRCh38
rs773113065 Chr.16: 89739279-89739283 on Build GRCh38
rs1005560269 Chr.16: 89739191-89739191 on Build GRCh38
rs372364590 Chr.16: 89739293-89739293 on Build GRCh38
rs775351812 Chr.16: 89739216-89739216 on Build GRCh38
rs746789359 Chr.16: 89739339-89739339 on Build GRCh38
rs371366647 Chr.16: 89739106-89739106 on Build GRCh38
rs553955430 Chr.16: 89739078-89739078 on Build GRCh38
rs749929686 Chr.16: 89739336-89739336 on Build GRCh38
rs767060427 Chr.16: 89739458-89739463 on Build GRCh38
rs984178125 Chr.16: 89739052-89739052 on Build GRCh38
rs777315528 Chr.16: 89739199-89739199 on Build GRCh38
rs781273405 Chr.16: 89739015-89739015 on Build GRCh38
rs773947543 Chr.16: 89739212-89739212 on Build GRCh38
rs745688750 Chr.16: 89739215-89739215 on Build GRCh38
rs751344774 Chr.16: 89739094-89739094 on Build GRCh38
rs769672682 Chr.16: 89739276-89739278 on Build GRCh38
rs764710767 Chr.16: 89739369-89739369 on Build GRCh38
rs769897335 Chr.16: 89739018-89739018 on Build GRCh38
rs750839091 Chr.16: 89739192-89739192 on Build GRCh38
rs1018933292 Chr.16: 89739444-89739444 on Build GRCh38
rs751928369 Chr.16: 89739261-89739261 on Build GRCh38
rs369398999 Chr.16: 89739323-89739323 on Build GRCh38
rs565924938 Chr.16: 89739258-89739258 on Build GRCh38
rs911481295 Chr.16: 89739291-89739292 on Build GRCh38
rs756403983 Chr.16: 89739086-89739086 on Build GRCh38
rs545928949 Chr.16: 89739104-89739104 on Build GRCh38
rs375508301 Chr.16: 89739059-89739059 on Build GRCh38
rs574812539 Chr.16: 89739058-89739058 on Build GRCh38
rs769830774 Chr.16: 89739445-89739445 on Build GRCh38
rs897226700 Chr.16: 89739395-89739395 on Build GRCh38
rs529210242 Chr.16: 89739207-89739207 on Build GRCh38
rs925692168 Chr.16: 89739405-89739405 on Build GRCh38
rs961489297 Chr.16: 89739205-89739205 on Build GRCh38
rs761988162 Chr.16: 89739290-89739290 on Build GRCh38
rs375383533 Chr.16: 89739307-89739307 on Build GRCh38
rs377636161 Chr.16: 89739090-89739090 on Build GRCh38
rs56010756 Chr.16: 89739434-89739434 on Build GRCh38
rs556778941 Chr.16: 89739040-89739040 on Build GRCh38
rs557800770 Chr.16: 89739455-89739455 on Build GRCh38
rs754990688 Chr.16: 89739011-89739011 on Build GRCh38
rs746203082 Chr.16: 89739112-89739112 on Build GRCh38
rs759174496 Chr.16: 89739330-89739331 on Build GRCh38
rs769115368 Chr.16: 89739137-89739137 on Build GRCh38
rs757592625 Chr.16: 89739189-89739189 on Build GRCh38
rs769505201 Chr.16: 89739092-89739092 on Build GRCh38
rs774951303 Chr.16: 89739037-89739037 on Build GRCh38
rs766314322 Chr.16: 89739067-89739067 on Build GRCh38
rs775234845 Chr.16: 89739162-89739162 on Build GRCh38
rs201225325 Chr.16: 89739195-89739195 on Build GRCh38
rs9282682 Chr.16: 89739386-89739386 on Build GRCh38
rs761934804 Chr.16: 89739249-89739249 on Build GRCh38
rs375237681 Chr.16: 89739196-89739196 on Build GRCh38
rs747194256 Chr.16: 89739090-89739091 on Build GRCh38
rs781393563 Chr.16: 89739081-89739081 on Build GRCh38
rs778544563 Chr.16: 89739269-89739269 on Build GRCh38
rs373682746 Chr.16: 89739337-89739337 on Build GRCh38
rs191404781 Chr.16: 89739087-89739087 on Build GRCh38
rs763049986 Chr.16: 89739470-89739470 on Build GRCh38
rs761574058 Chr.16: 89739045-89739045 on Build GRCh38
rs772665586 Chr.16: 89739125-89739125 on Build GRCh38
rs773679037 Chr.16: 89739020-89739020 on Build GRCh38
rs779280925 Chr.16: 89739014-89739017 on Build GRCh38
rs778348461 Chr.16: 89739332-89739332 on Build GRCh38
rs921303215 Chr.16: 89739360-89739360 on Build GRCh38
rs771283997 Chr.16: 89739306-89739306 on Build GRCh38
rs535784995 Chr.16: 89739379-89739379 on Build GRCh38
rs780301799 Chr.16: 89739086-89739087 on Build GRCh38
rs760814763 Chr.16: 89739167-89739167 on Build GRCh38
rs572335110 Chr.16: 89739428-89739428 on Build GRCh38
rs762625682 Chr.16: 89739400-89739400 on Build GRCh38
rs779568987 Chr.16: 89739089-89739089 on Build GRCh38
rs746415530 Chr.16: 89739051-89739051 on Build GRCh38
rs17227389 Chr.16: 89739265-89739265 on Build GRCh38
rs776244734 Chr.16: 89739091-89739091 on Build GRCh38
rs768733593 Chr.16: 89739278-89739278 on Build GRCh38
rs371280585 Chr.16: 89739025-89739025 on Build GRCh38
rs762902309 Chr.16: 89739285-89739285 on Build GRCh38
rs17227396 Chr.16: 89739264-89739264 on Build GRCh38
rs747892390 Chr.16: 89739218-89739231 on Build GRCh38
rs748472421 Chr.16: 89739017-89739017 on Build GRCh38
rs759521427 Chr.16: 89739096-89739096 on Build GRCh38
rs911709601 Chr.16: 89739385-89739385 on Build GRCh38
rs145886270 Chr.16: 89739236-89739236 on Build GRCh38
rs531372521 Chr.16: 89739143-89739143 on Build GRCh38
rs199599393 Chr.16: 89739170-89739170 on Build GRCh38
rs999835890 Chr.16: 89739239-89739239 on Build GRCh38
rs1041491550 Chr.16: 89739202-89739202 on Build GRCh38
rs768026295 Chr.16: 89739316-89739316 on Build GRCh38
rs34218545 Chr.16: 89739059-89739059 on Build GRCh38
rs766226560 Chr.16: 89739093-89739093 on Build GRCh38
rs55873203 Chr.16: 89739013-89739014 on Build GRCh38
rs759216137 Chr.16: 89739154-89739154 on Build GRCh38
rs149775657 Chr.16: 89739285-89739285 on Build GRCh38
rs779466859 Chr.16: 89739111-89739111 on Build GRCh38
rs754048546 Chr.16: 89739109-89739109 on Build GRCh38
rs553145380 Chr.16: 89739064-89739064 on Build GRCh38
rs773859464 Chr.16: 89739329-89739330 on Build GRCh38
rs753944130 Chr.16: 89739183-89739183 on Build GRCh38
rs200304203 Chr.16: 89739107-89739107 on Build GRCh38
rs755479861 Chr.16: 89739262-89739262 on Build GRCh38
rs765515655 Chr.16: 89739190-89739190 on Build GRCh38
rs372257628 Chr.16: 89739415-89739415 on Build GRCh38
rs112734327 Chr.16: 89739281-89739281 on Build GRCh38
rs750543279 Chr.16: 89739255-89739255 on Build GRCh38
rs752700058 Chr.16: 89739083-89739083 on Build GRCh38
rs1023409042 Chr.16: 89739029-89739029 on Build GRCh38
rs757550124 Chr.16: 89739110-89739110 on Build GRCh38
rs955687843 Chr.16: 89739416-89739416 on Build GRCh38
rs775989654 Chr.16: 89739043-89739043 on Build GRCh38
rs994282443 Chr.16: 89739402-89739402 on Build GRCh38
rs766710139 Chr.16: 89739260-89739260 on Build GRCh38
rs766594215 Chr.16: 89739309-89739309 on Build GRCh38
rs773327440 Chr.16: 89739304-89739304 on Build GRCh38
rs768765651 Chr.16: 89739098-89739099 on Build GRCh38
rs978377623 Chr.16: 89739459-89739459 on Build GRCh38
rs768494844 Chr.16: 89739341-89739341 on Build GRCh38
rs530872988 Chr.16: 89739315-89739316 on Build GRCh38
rs538139633 Chr.16: 89739031-89739031 on Build GRCh38
rs374765708 Chr.16: 89739131-89739131 on Build GRCh38
rs763132382 Chr.16: 89739305-89739305 on Build GRCh38
rs1041955692 Chr.16: 89739404-89739404 on Build GRCh38
rs752457319 Chr.16: 89739460-89739477 on Build GRCh38
rs376189323 Chr.16: 89739313-89739313 on Build GRCh38
rs201740587 Chr.16: 89739277-89739277 on Build GRCh38
rs771957724 Chr.16: 89739013-89739013 on Build GRCh38
rs772701240 Chr.16: 89739138-89739138 on Build GRCh38
rs776969626 Chr.16: 89739175-89739176 on Build GRCh38
rs760589739 Chr.16: 89739224-89739224 on Build GRCh38
rs370080063 Chr.16: 89739129-89739129 on Build GRCh38
rs56336895 Chr.16: 89739364-89739364 on Build GRCh38
rs756611935 Chr.16: 89739326-89739326 on Build GRCh38
rs745577614 Chr.16: 89739271-89739271 on Build GRCh38
rs770551622 Chr.16: 89739145-89739145 on Build GRCh38
rs760276584 Chr.16: 89739039-89739039 on Build GRCh38
rs1055386152 Chr.16: 89739430-89739430 on Build GRCh38

View Full Product Details