Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 180983-181487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOC2B
Entrez Gene ID
8447
Gene Name
double C2 domain beta
Gene Aliases
DOC2BL
UniGene
Hs.551705
Gene Chromosome Location
Chr.17: 146748-181630 on Build GRCh38
Transcripts
NM_003585.4 XM_011535363.2 XM_017025227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3755384 c.63C>T p.D21D Chr.17: 181417-181417 on Build GRCh38
6021485 c.349G>A p.D117N Chr.17: 181131-181131 on Build GRCh38
6021486 c.244G>A p.D82N Chr.17: 181236-181236 on Build GRCh38
6356908 c.93G>A p.K31K Chr.17: 181387-181387 on Build GRCh38
3691478 c.124C>T p.R42W Chr.17: 181356-181356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966329790 Chr.17: 181049-181049 on Build GRCh38
rs780749144 Chr.17: 181160-181160 on Build GRCh38
rs1043049837 Chr.17: 181123-181123 on Build GRCh38
rs975027358 Chr.17: 181161-181161 on Build GRCh38
rs931463480 Chr.17: 181268-181268 on Build GRCh38
rs983799846 Chr.17: 181041-181041 on Build GRCh38
rs937323498 Chr.17: 181440-181440 on Build GRCh38
rs913302950 Chr.17: 181180-181180 on Build GRCh38
rs907471731 Chr.17: 181083-181083 on Build GRCh38
rs747513578 Chr.17: 181343-181343 on Build GRCh38
rs1028901711 Chr.17: 181369-181369 on Build GRCh38
rs1005004998 Chr.17: 181317-181317 on Build GRCh38
rs866842886 Chr.17: 181190-181190 on Build GRCh38
rs866873676 Chr.17: 181208-181208 on Build GRCh38
rs1004570228 Chr.17: 181144-181144 on Build GRCh38
rs937645368 Chr.17: 181237-181237 on Build GRCh38
rs961529091 Chr.17: 181175-181186 on Build GRCh38
rs775461472 Chr.17: 181420-181420 on Build GRCh38
rs931535608 Chr.17: 181446-181446 on Build GRCh38
rs957217850 Chr.17: 181378-181378 on Build GRCh38
rs950379783 Chr.17: 181033-181033 on Build GRCh38
rs1018615956 Chr.17: 181332-181332 on Build GRCh38
rs920018945 Chr.17: 181054-181054 on Build GRCh38
rs548884378 Chr.17: 181469-181469 on Build GRCh38
rs144982616 Chr.17: 181021-181021 on Build GRCh38
rs188684711 Chr.17: 181370-181370 on Build GRCh38
rs1047194093 Chr.17: 181284-181284 on Build GRCh38
rs920196084 Chr.17: 181445-181445 on Build GRCh38
rs868547346 Chr.17: 181191-181191 on Build GRCh38
rs868700467 Chr.17: 181205-181205 on Build GRCh38
rs758814753 Chr.17: 181088-181088 on Build GRCh38
rs769487824 Chr.17: 181345-181345 on Build GRCh38
rs770636834 Chr.17: 181410-181410 on Build GRCh38
rs200059495 Chr.17: 181351-181351 on Build GRCh38
rs544143652 Chr.17: 181025-181025 on Build GRCh38
rs886202075 Chr.17: 181308-181308 on Build GRCh38
rs977248255 Chr.17: 181184-181195 on Build GRCh38
rs551529924 Chr.17: 181408-181408 on Build GRCh38
rs911792466 Chr.17: 181048-181049 on Build GRCh38
rs912749818 Chr.17: 181072-181072 on Build GRCh38
rs1046429571 Chr.17: 181075-181075 on Build GRCh38
rs1040597801 Chr.17: 181165-181165 on Build GRCh38
rs908742535 Chr.17: 181174-181175 on Build GRCh38
rs1055969214 Chr.17: 181094-181094 on Build GRCh38
rs753987588 Chr.17: 181451-181451 on Build GRCh38
rs763956550 Chr.17: 181444-181444 on Build GRCh38
rs913270056 Chr.17: 181066-181066 on Build GRCh38
rs563369420 Chr.17: 181411-181411 on Build GRCh38
rs760616738 Chr.17: 181423-181423 on Build GRCh38
rs994577978 Chr.17: 181113-181113 on Build GRCh38
rs961390102 Chr.17: 181050-181050 on Build GRCh38
rs541499941 Chr.17: 181294-181294 on Build GRCh38
rs147645420 Chr.17: 181417-181417 on Build GRCh38
rs574089430 Chr.17: 181058-181058 on Build GRCh38
rs990003322 Chr.17: 181392-181392 on Build GRCh38
rs74169589 Chr.17: 181035-181035 on Build GRCh38
rs757239970 Chr.17: 181456-181456 on Build GRCh38
rs996279800 Chr.17: 181318-181318 on Build GRCh38
rs1055962026 Chr.17: 181239-181239 on Build GRCh38
rs1018996888 Chr.17: 181151-181151 on Build GRCh38
rs900610732 Chr.17: 181183-181183 on Build GRCh38
rs865814753 Chr.17: 181206-181206 on Build GRCh38
rs559791077 Chr.17: 181307-181307 on Build GRCh38
rs886129621 Chr.17: 181141-181141 on Build GRCh38
rs994505691 Chr.17: 181024-181024 on Build GRCh38
rs1001693091 Chr.17: 181015-181015 on Build GRCh38
rs759362130 Chr.17: 181414-181414 on Build GRCh38
rs901504290 Chr.17: 181334-181334 on Build GRCh38
rs985552674 Chr.17: 181060-181060 on Build GRCh38
rs898664072 Chr.17: 181099-181100 on Build GRCh38
rs1028870671 Chr.17: 181166-181166 on Build GRCh38
rs928685174 Chr.17: 181438-181438 on Build GRCh38
rs1027347335 Chr.17: 181132-181132 on Build GRCh38
rs898693816 Chr.17: 181256-181256 on Build GRCh38
rs184036955 Chr.17: 181051-181051 on Build GRCh38

View Full Product Details