Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 181379-181870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOC2B
Entrez Gene ID
8447
Gene Name
double C2 domain beta
Gene Aliases
DOC2BL
UniGene
Hs.551705
Gene Chromosome Location
Chr.17: 146748-181630 on Build GRCh38
Transcripts
NM_003585.4 XM_011535363.2 XM_017025227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3755384 c.63C>T p.D21D Chr.17: 181417-181417 on Build GRCh38
6356908 c.93G>A p.K31K Chr.17: 181387-181387 on Build GRCh38
6391151 c.23A>G p.E8G Chr.17: 181457-181457 on Build GRCh38
6391856 c.37A>G p.S13G Chr.17: 181443-181443 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs892802801 Chr.17: 181748-181748 on Build GRCh38
rs1020167841 Chr.17: 181593-181593 on Build GRCh38
rs923035552 Chr.17: 181730-181730 on Build GRCh38
rs937323498 Chr.17: 181440-181440 on Build GRCh38
rs1003136949 Chr.17: 181805-181805 on Build GRCh38
rs1050097806 Chr.17: 181746-181746 on Build GRCh38
rs967607210 Chr.17: 181595-181595 on Build GRCh38
rs1011531950 Chr.17: 181755-181755 on Build GRCh38
rs534325308 Chr.17: 181772-181772 on Build GRCh38
rs1040519820 Chr.17: 181493-181493 on Build GRCh38
rs931535608 Chr.17: 181446-181446 on Build GRCh38
rs566875759 Chr.17: 181514-181514 on Build GRCh38
rs775461472 Chr.17: 181420-181420 on Build GRCh38
rs1011467961 Chr.17: 181587-181587 on Build GRCh38
rs1036163169 Chr.17: 181610-181610 on Build GRCh38
rs548884378 Chr.17: 181469-181469 on Build GRCh38
rs920196084 Chr.17: 181445-181445 on Build GRCh38
rs770636834 Chr.17: 181410-181410 on Build GRCh38
rs868378465 Chr.17: 181692-181692 on Build GRCh38
rs551529924 Chr.17: 181408-181408 on Build GRCh38
rs34151622 Chr.17: 181648-181648 on Build GRCh38
rs908744636 Chr.17: 181666-181666 on Build GRCh38
rs893073645 Chr.17: 181554-181554 on Build GRCh38
rs753987588 Chr.17: 181451-181451 on Build GRCh38
rs760616738 Chr.17: 181423-181423 on Build GRCh38
rs763956550 Chr.17: 181444-181444 on Build GRCh38
rs563369420 Chr.17: 181411-181411 on Build GRCh38
rs952966460 Chr.17: 181660-181660 on Build GRCh38
rs771813392 Chr.17: 181490-181515 on Build GRCh38
rs991462884 Chr.17: 181618-181618 on Build GRCh38
rs919892847 Chr.17: 181652-181652 on Build GRCh38
rs1040148454 Chr.17: 181679-181679 on Build GRCh38
rs147645420 Chr.17: 181417-181417 on Build GRCh38
rs747309146 Chr.17: 181518-181518 on Build GRCh38
rs758901207 Chr.17: 181490-181490 on Build GRCh38
rs982994312 Chr.17: 181661-181661 on Build GRCh38
rs571314931 Chr.17: 181838-181838 on Build GRCh38
rs1050067021 Chr.17: 181520-181520 on Build GRCh38
rs757239970 Chr.17: 181456-181456 on Build GRCh38
rs1035499697 Chr.17: 181825-181825 on Build GRCh38
rs958794863 Chr.17: 181613-181613 on Build GRCh38
rs780554915 Chr.17: 181496-181496 on Build GRCh38
rs944191575 Chr.17: 181673-181673 on Build GRCh38
rs546317936 Chr.17: 181774-181774 on Build GRCh38
rs1002985875 Chr.17: 181605-181605 on Build GRCh38
rs958788912 Chr.17: 181847-181847 on Build GRCh38
rs759362130 Chr.17: 181414-181414 on Build GRCh38
rs901538283 Chr.17: 181506-181506 on Build GRCh38
rs931693127 Chr.17: 181742-181742 on Build GRCh38
rs928685174 Chr.17: 181438-181438 on Build GRCh38

View Full Product Details