Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 410237-410738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf97
Entrez Gene ID
400566
Gene Name
chromosome 17 open reading frame 97
Gene Aliases
CK20, LIAT1
UniGene
Hs.499607
Gene Chromosome Location
Chr.17: 410327-414666 on Build GRCh38
Transcripts
NM_001013672.4 NR_136407.1

Gene Symbol
LOC105371430
Entrez Gene ID
105371430
Gene Name
uncharacterized LOC105371430
Gene Aliases
-
UniGene
Hs.616427
Gene Chromosome Location
Chr.17: 404535-414023 on Build GRCh38
Transcripts
NM_001013672.4 NR_136407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5694457 c.128C>G p.A43G Chr.17: 410470-410470 on Build GRCh38
3749393 c.9G>T p.T3T Chr.17: 410351-410351 on Build GRCh38
3749394 c.49T>C p.L17L Chr.17: 410391-410391 on Build GRCh38
6317456 c.61G>T p.G21W Chr.17: 410403-410403 on Build GRCh38
4617741 c.118C>T p.R40C Chr.17: 410460-410460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7503725 Chr.17: 410351-410351 on Build GRCh38
rs1013592988 Chr.17: 410468-410468 on Build GRCh38
rs782785887 Chr.17: 410325-410325 on Build GRCh38
rs782726428 Chr.17: 410384-410384 on Build GRCh38
rs782341943 Chr.17: 410609-410609 on Build GRCh38
rs200863451 Chr.17: 410289-410290 on Build GRCh38
rs782005776 Chr.17: 410363-410363 on Build GRCh38
rs782166778 Chr.17: 410330-410330 on Build GRCh38
rs547983708 Chr.17: 410471-410471 on Build GRCh38
rs782813580 Chr.17: 410332-410332 on Build GRCh38
rs7502594 Chr.17: 410391-410391 on Build GRCh38
rs782431874 Chr.17: 410302-410302 on Build GRCh38
rs532664861 Chr.17: 410419-410419 on Build GRCh38
rs782051805 Chr.17: 410445-410445 on Build GRCh38
rs782502424 Chr.17: 410352-410352 on Build GRCh38
rs961012049 Chr.17: 410406-410406 on Build GRCh38
rs782723895 Chr.17: 410348-410348 on Build GRCh38
rs934734670 Chr.17: 410623-410623 on Build GRCh38
rs924686157 Chr.17: 410620-410620 on Build GRCh38
rs1027071207 Chr.17: 410547-410547 on Build GRCh38
rs781860740 Chr.17: 410326-410326 on Build GRCh38
rs558440796 Chr.17: 410587-410587 on Build GRCh38
rs782635683 Chr.17: 410501-410501 on Build GRCh38
rs782560901 Chr.17: 410472-410472 on Build GRCh38
rs782719705 Chr.17: 410288-410288 on Build GRCh38
rs907383259 Chr.17: 410478-410478 on Build GRCh38
rs181148788 Chr.17: 410407-410407 on Build GRCh38
rs782680431 Chr.17: 410354-410354 on Build GRCh38
rs4581766 Chr.17: 410508-410508 on Build GRCh38
rs782535189 Chr.17: 410377-410377 on Build GRCh38
rs371014858 Chr.17: 410515-410515 on Build GRCh38
rs985796891 Chr.17: 410577-410577 on Build GRCh38
rs188378590 Chr.17: 410310-410310 on Build GRCh38
rs782164120 Chr.17: 410431-410431 on Build GRCh38
rs781783260 Chr.17: 410624-410624 on Build GRCh38
rs782272301 Chr.17: 410356-410356 on Build GRCh38
rs534036714 Chr.17: 410567-410567 on Build GRCh38
rs781936716 Chr.17: 410438-410438 on Build GRCh38
rs782473527 Chr.17: 410497-410497 on Build GRCh38
rs782061032 Chr.17: 410648-410648 on Build GRCh38
rs782412409 Chr.17: 410513-410513 on Build GRCh38
rs782345354 Chr.17: 410359-410359 on Build GRCh38
rs782268580 Chr.17: 410425-410425 on Build GRCh38
rs782214675 Chr.17: 410436-410437 on Build GRCh38
rs919868294 Chr.17: 410265-410265 on Build GRCh38
rs781967744 Chr.17: 410321-410321 on Build GRCh38
rs529529303 Chr.17: 410458-410458 on Build GRCh38
rs912501877 Chr.17: 410580-410580 on Build GRCh38
rs1050059859 Chr.17: 410292-410292 on Build GRCh38
rs564797333 Chr.17: 410268-410268 on Build GRCh38
rs781948972 Chr.17: 410369-410369 on Build GRCh38
rs202232939 Chr.17: 410448-410448 on Build GRCh38
rs560133543 Chr.17: 410385-410385 on Build GRCh38
rs782397718 Chr.17: 410714-410714 on Build GRCh38
rs750745401 Chr.17: 410427-410427 on Build GRCh38
rs533783222 Chr.17: 410261-410261 on Build GRCh38
rs782482289 Chr.17: 410346-410346 on Build GRCh38
rs782643546 Chr.17: 410410-410410 on Build GRCh38
rs186460174 Chr.17: 410436-410436 on Build GRCh38
rs782225930 Chr.17: 410358-410358 on Build GRCh38
rs782333549 Chr.17: 410404-410404 on Build GRCh38
rs782689597 Chr.17: 410399-410399 on Build GRCh38
rs782201134 Chr.17: 410415-410415 on Build GRCh38
rs139629895 Chr.17: 410362-410362 on Build GRCh38
rs781910500 Chr.17: 410463-410463 on Build GRCh38
rs782286822 Chr.17: 410403-410403 on Build GRCh38
rs781783421 Chr.17: 410342-410342 on Build GRCh38
rs948976057 Chr.17: 410667-410667 on Build GRCh38
rs961146523 Chr.17: 410529-410529 on Build GRCh38
rs782781299 Chr.17: 410460-410460 on Build GRCh38
rs376563819 Chr.17: 410295-410295 on Build GRCh38
rs782395760 Chr.17: 410566-410566 on Build GRCh38
rs544517665 Chr.17: 410340-410340 on Build GRCh38
rs782086328 Chr.17: 410563-410563 on Build GRCh38
rs537734547 Chr.17: 410666-410666 on Build GRCh38
rs191309282 Chr.17: 410682-410682 on Build GRCh38
rs782311624 Chr.17: 410315-410315 on Build GRCh38
rs781797945 Chr.17: 410483-410483 on Build GRCh38
rs767872907 Chr.17: 410400-410401 on Build GRCh38
rs566579133 Chr.17: 410548-410548 on Build GRCh38
rs781979287 Chr.17: 410420-410420 on Build GRCh38
rs782542776 Chr.17: 410470-410470 on Build GRCh38
rs782088302 Chr.17: 410324-410324 on Build GRCh38
rs558718845 Chr.17: 410298-410298 on Build GRCh38
rs76528291 Chr.17: 410554-410554 on Build GRCh38
rs1040460983 Chr.17: 410444-410444 on Build GRCh38
rs374291590 Chr.17: 410457-410457 on Build GRCh38
rs782425524 Chr.17: 410364-410364 on Build GRCh38
rs1003020281 Chr.17: 410486-410486 on Build GRCh38
rs782755412 Chr.17: 410680-410680 on Build GRCh38
rs782596035 Chr.17: 410386-410386 on Build GRCh38
rs574519890 Chr.17: 410360-410360 on Build GRCh38
rs782236519 Chr.17: 410296-410296 on Build GRCh38
rs781900579 Chr.17: 410376-410376 on Build GRCh38
rs943838316 Chr.17: 410603-410603 on Build GRCh38
rs889206764 Chr.17: 410307-410307 on Build GRCh38
rs973709423 Chr.17: 410543-410543 on Build GRCh38
rs782022629 Chr.17: 410367-410367 on Build GRCh38
rs570654883 Chr.17: 410636-410636 on Build GRCh38
rs566321247 Chr.17: 410260-410260 on Build GRCh38
rs556464286 Chr.17: 410673-410673 on Build GRCh38

View Full Product Details