Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 745892-746387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEMIN4
Entrez Gene ID
50628
Gene Name
gem nuclear organelle associated protein 4
Gene Aliases
HC56, HCAP1, HHRF-1, p97
UniGene
Hs.499620
Gene Chromosome Location
Chr.17: 744421-752868 on Build GRCh38
Transcripts
NM_015721.2 XM_005256667.4 XM_005256670.4 XM_011523910.2 XM_011523911.2 XM_011523912.2 XM_011523913.2 XM_017024709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4473856 c.1879C>T p.L627F Chr.17: 746164-746164 on Build GRCh38
1385279 c.1778A>T p.E593V Chr.17: 746265-746265 on Build GRCh38
3521131 c.2004C>T p.D668D Chr.17: 746039-746039 on Build GRCh38
148143 c.1996G>C p.E666Q Chr.17: 746047-746047 on Build GRCh38
1610644 c.1796C>G p.S599C Chr.17: 746247-746247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759630942 Chr.17: 746326-746326 on Build GRCh38
rs199702345 Chr.17: 746272-746272 on Build GRCh38
rs562143356 Chr.17: 746139-746139 on Build GRCh38
rs780061624 Chr.17: 746079-746079 on Build GRCh38
rs770601467 Chr.17: 746321-746321 on Build GRCh38
rs776242615 Chr.17: 746244-746244 on Build GRCh38
rs747763804 Chr.17: 746293-746293 on Build GRCh38
rs533913356 Chr.17: 746140-746140 on Build GRCh38
rs775968214 Chr.17: 745923-745923 on Build GRCh38
rs143008523 Chr.17: 746304-746304 on Build GRCh38
rs773931767 Chr.17: 745941-745941 on Build GRCh38
rs767196443 Chr.17: 745951-745951 on Build GRCh38
rs371459371 Chr.17: 746186-746186 on Build GRCh38
rs775642910 Chr.17: 746328-746328 on Build GRCh38
rs754371280 Chr.17: 746336-746336 on Build GRCh38
rs764982624 Chr.17: 746153-746153 on Build GRCh38
rs199571890 Chr.17: 745999-745999 on Build GRCh38
rs765070381 Chr.17: 746061-746061 on Build GRCh38
rs753601096 Chr.17: 746038-746038 on Build GRCh38
rs754463282 Chr.17: 746287-746287 on Build GRCh38
rs776310179 Chr.17: 746105-746107 on Build GRCh38
rs3744741 Chr.17: 745992-745992 on Build GRCh38
rs771679352 Chr.17: 746012-746012 on Build GRCh38
rs757544507 Chr.17: 746295-746295 on Build GRCh38
rs374700498 Chr.17: 746142-746142 on Build GRCh38
rs759844591 Chr.17: 746266-746266 on Build GRCh38
rs34519093 Chr.17: 745924-745924 on Build GRCh38
rs770262994 Chr.17: 746112-746112 on Build GRCh38
rs1009180395 Chr.17: 746289-746289 on Build GRCh38
rs750243511 Chr.17: 746155-746155 on Build GRCh38
rs748035562 Chr.17: 746245-746245 on Build GRCh38
rs368959543 Chr.17: 746005-746005 on Build GRCh38
rs372394762 Chr.17: 746299-746299 on Build GRCh38
rs750619292 Chr.17: 746062-746062 on Build GRCh38
rs759112263 Chr.17: 745943-745943 on Build GRCh38
rs774893520 Chr.17: 746013-746013 on Build GRCh38
rs779567946 Chr.17: 746222-746222 on Build GRCh38
rs776892989 Chr.17: 745935-745935 on Build GRCh38
rs776483020 Chr.17: 745993-745993 on Build GRCh38
rs758056773 Chr.17: 746213-746213 on Build GRCh38
rs1007005570 Chr.17: 746029-746029 on Build GRCh38
rs749939964 Chr.17: 746205-746205 on Build GRCh38
rs774233710 Chr.17: 746324-746324 on Build GRCh38
rs780153129 Chr.17: 745978-745978 on Build GRCh38
rs953196599 Chr.17: 746019-746019 on Build GRCh38
rs765476343 Chr.17: 746364-746364 on Build GRCh38
rs762622001 Chr.17: 745937-745937 on Build GRCh38
rs771274075 Chr.17: 746327-746328 on Build GRCh38
rs769037135 Chr.17: 745933-745933 on Build GRCh38
rs779108447 Chr.17: 745966-745966 on Build GRCh38
rs542942969 Chr.17: 746319-746319 on Build GRCh38
rs766254914 Chr.17: 746256-746256 on Build GRCh38
rs4968104 Chr.17: 746265-746265 on Build GRCh38
rs761587959 Chr.17: 746055-746055 on Build GRCh38
rs758457318 Chr.17: 745975-745975 on Build GRCh38
rs375214416 Chr.17: 746126-746126 on Build GRCh38
rs763952079 Chr.17: 746032-746032 on Build GRCh38
rs1046242682 Chr.17: 746191-746191 on Build GRCh38
rs916642992 Chr.17: 746173-746173 on Build GRCh38
rs746151136 Chr.17: 745973-745973 on Build GRCh38
rs368058625 Chr.17: 746212-746212 on Build GRCh38
rs762712933 Chr.17: 746233-746233 on Build GRCh38
rs550280037 Chr.17: 745998-745998 on Build GRCh38
rs759313307 Chr.17: 746010-746010 on Build GRCh38
rs534078852 Chr.17: 746364-746365 on Build GRCh38
rs767791810 Chr.17: 746327-746327 on Build GRCh38
rs763908038 Chr.17: 745961-745961 on Build GRCh38
rs762736188 Chr.17: 746254-746254 on Build GRCh38
rs377606651 Chr.17: 746095-746095 on Build GRCh38
rs764724631 Chr.17: 746110-746110 on Build GRCh38
rs760611110 Chr.17: 746015-746015 on Build GRCh38
rs778323201 Chr.17: 746124-746124 on Build GRCh38
rs761785821 Chr.17: 746152-746152 on Build GRCh38
rs756630533 Chr.17: 746276-746276 on Build GRCh38
rs1026532007 Chr.17: 746006-746006 on Build GRCh38
rs772928799 Chr.17: 746253-746253 on Build GRCh38
rs906433403 Chr.17: 745985-745985 on Build GRCh38
rs376416115 Chr.17: 746316-746316 on Build GRCh38
rs753597687 Chr.17: 745962-745962 on Build GRCh38
rs200492563 Chr.17: 746141-746141 on Build GRCh38
rs756721979 Chr.17: 746210-746210 on Build GRCh38
rs373700715 Chr.17: 746193-746193 on Build GRCh38
rs746728929 Chr.17: 746236-746236 on Build GRCh38
rs200818121 Chr.17: 746077-746077 on Build GRCh38
rs766146852 Chr.17: 745940-745940 on Build GRCh38
rs200443736 Chr.17: 746161-746161 on Build GRCh38
rs898838600 Chr.17: 745996-745996 on Build GRCh38
rs370625526 Chr.17: 746096-746096 on Build GRCh38
rs764701907 Chr.17: 746281-746281 on Build GRCh38
rs768076749 Chr.17: 746021-746021 on Build GRCh38
rs548208529 Chr.17: 745964-745964 on Build GRCh38
rs1008625124 Chr.17: 746275-746275 on Build GRCh38
rs890703589 Chr.17: 746070-746070 on Build GRCh38
rs111822058 Chr.17: 746331-746331 on Build GRCh38
rs368565942 Chr.17: 746156-746156 on Build GRCh38
rs898791411 Chr.17: 746241-746241 on Build GRCh38
rs377022947 Chr.17: 746002-746002 on Build GRCh38
rs768025861 Chr.17: 746239-746239 on Build GRCh38
rs576980675 Chr.17: 746163-746163 on Build GRCh38
rs374674049 Chr.17: 746103-746103 on Build GRCh38
rs573134834 Chr.17: 746341-746341 on Build GRCh38
rs369912401 Chr.17: 746310-746310 on Build GRCh38
rs754999536 Chr.17: 746181-746181 on Build GRCh38
rs769363855 Chr.17: 746315-746315 on Build GRCh38
rs763106997 Chr.17: 746160-746160 on Build GRCh38
rs752659494 Chr.17: 745952-745952 on Build GRCh38
rs756044477 Chr.17: 745960-745960 on Build GRCh38
rs910925 Chr.17: 746307-746307 on Build GRCh38
rs373348504 Chr.17: 746000-746000 on Build GRCh38

View Full Product Details