Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 769777-770275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLOD4
Entrez Gene ID
51031
Gene Name
glyoxalase domain containing 4
Gene Aliases
C17orf25, CGI-150, HC71
UniGene
Hs.279061
Gene Chromosome Location
Chr.17: 759309-782331 on Build GRCh38
Transcripts
NM_016080.3 XM_006721540.2 XM_017024711.1 XM_017024712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6262026 c.693C>A p.S231R Chr.17: 770095-770095 on Build GRCh38
4429177 c.784C>T p.R262* Chr.17: 769916-769916 on Build GRCh38
344589 c.631T>C p.L211L Chr.17: 770157-770157 on Build GRCh38
6671679 c.861C>A p.S287R Chr.17: 769884-769884 on Build GRCh38
4270509 c.784G>A p.D262N Chr.17: 770049-770049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953243593 Chr.17: 769997-769997 on Build GRCh38
rs376633659 Chr.17: 769977-769977 on Build GRCh38
rs746796656 Chr.17: 770197-770197 on Build GRCh38
rs750667835 Chr.17: 770002-770003 on Build GRCh38
rs776883388 Chr.17: 770210-770210 on Build GRCh38
rs762699207 Chr.17: 770025-770026 on Build GRCh38
rs754861743 Chr.17: 770192-770193 on Build GRCh38
rs746970515 Chr.17: 769991-769991 on Build GRCh38
rs368221029 Chr.17: 770183-770183 on Build GRCh38
rs775647255 Chr.17: 770045-770045 on Build GRCh38
rs753988099 Chr.17: 769822-769822 on Build GRCh38
rs368905215 Chr.17: 770010-770010 on Build GRCh38
rs542245318 Chr.17: 769892-769892 on Build GRCh38
rs764027687 Chr.17: 770049-770049 on Build GRCh38
rs780970288 Chr.17: 769989-769989 on Build GRCh38
rs374317025 Chr.17: 770181-770181 on Build GRCh38
rs1046655436 Chr.17: 769835-769835 on Build GRCh38
rs767800783 Chr.17: 770239-770239 on Build GRCh38
rs763577506 Chr.17: 770155-770155 on Build GRCh38
rs763191316 Chr.17: 770217-770217 on Build GRCh38
rs754254427 Chr.17: 769955-769955 on Build GRCh38
rs762634820 Chr.17: 769821-769821 on Build GRCh38
rs148206481 Chr.17: 769920-769920 on Build GRCh38
rs755355039 Chr.17: 770171-770171 on Build GRCh38
rs113617081 Chr.17: 770056-770056 on Build GRCh38
rs771800790 Chr.17: 770129-770129 on Build GRCh38
rs17851349 Chr.17: 769899-769899 on Build GRCh38
rs766169780 Chr.17: 770037-770040 on Build GRCh38
rs747843878 Chr.17: 769998-769998 on Build GRCh38
rs753477278 Chr.17: 770243-770243 on Build GRCh38
rs767711047 Chr.17: 769938-769938 on Build GRCh38
rs770896118 Chr.17: 770013-770013 on Build GRCh38
rs946621897 Chr.17: 770153-770153 on Build GRCh38
rs769389516 Chr.17: 770005-770005 on Build GRCh38
rs758340343 Chr.17: 770196-770196 on Build GRCh38
rs114814106 Chr.17: 769915-769915 on Build GRCh38
rs373166347 Chr.17: 769804-769805 on Build GRCh38
rs773439162 Chr.17: 770215-770215 on Build GRCh38
rs757800101 Chr.17: 769968-769968 on Build GRCh38
rs759302550 Chr.17: 770030-770030 on Build GRCh38
rs765424101 Chr.17: 769971-769972 on Build GRCh38
rs112396332 Chr.17: 770050-770050 on Build GRCh38
rs115211226 Chr.17: 769928-769928 on Build GRCh38
rs756748678 Chr.17: 770184-770184 on Build GRCh38
rs773342760 Chr.17: 769820-769820 on Build GRCh38
rs145702773 Chr.17: 770143-770143 on Build GRCh38
rs745744569 Chr.17: 770195-770195 on Build GRCh38
rs747617679 Chr.17: 770098-770098 on Build GRCh38
rs765274994 Chr.17: 770149-770149 on Build GRCh38
rs766782753 Chr.17: 770159-770159 on Build GRCh38
rs751444806 Chr.17: 770186-770186 on Build GRCh38
rs999212353 Chr.17: 769906-769906 on Build GRCh38
rs769479560 Chr.17: 769909-769909 on Build GRCh38
rs11663 Chr.17: 769950-769950 on Build GRCh38
rs774211950 Chr.17: 770019-770019 on Build GRCh38
rs144364801 Chr.17: 770043-770043 on Build GRCh38
rs779082623 Chr.17: 769971-769971 on Build GRCh38
rs115327234 Chr.17: 770122-770122 on Build GRCh38
rs760823386 Chr.17: 770137-770137 on Build GRCh38
rs778011488 Chr.17: 769807-769807 on Build GRCh38
rs780597980 Chr.17: 770077-770077 on Build GRCh38
rs902341073 Chr.17: 770179-770179 on Build GRCh38
rs960455532 Chr.17: 769907-769907 on Build GRCh38
rs761994898 Chr.17: 769864-769865 on Build GRCh38
rs749838246 Chr.17: 769886-769886 on Build GRCh38
rs775281534 Chr.17: 770136-770136 on Build GRCh38
rs751683450 Chr.17: 769832-769832 on Build GRCh38
rs372308114 Chr.17: 770199-770199 on Build GRCh38
rs199729596 Chr.17: 770067-770067 on Build GRCh38
rs756241718 Chr.17: 769880-769880 on Build GRCh38
rs770394682 Chr.17: 770103-770103 on Build GRCh38
rs116072465 Chr.17: 770090-770090 on Build GRCh38
rs751967625 Chr.17: 770169-770169 on Build GRCh38
rs758892735 Chr.17: 769979-769979 on Build GRCh38
rs776496105 Chr.17: 769902-769902 on Build GRCh38
rs977124484 Chr.17: 770060-770060 on Build GRCh38
rs771333749 Chr.17: 770247-770247 on Build GRCh38
rs896198799 Chr.17: 770203-770203 on Build GRCh38
rs759910427 Chr.17: 770234-770234 on Build GRCh38
rs746588800 Chr.17: 769898-769898 on Build GRCh38
rs760905206 Chr.17: 770046-770046 on Build GRCh38
rs769869885 Chr.17: 770211-770211 on Build GRCh38
rs751018018 Chr.17: 770065-770065 on Build GRCh38
rs370605102 Chr.17: 770145-770145 on Build GRCh38
rs758806415 Chr.17: 770070-770070 on Build GRCh38
rs925646363 Chr.17: 770083-770083 on Build GRCh38
rs766236555 Chr.17: 769829-769829 on Build GRCh38
rs184466050 Chr.17: 769916-769916 on Build GRCh38
rs143261573 Chr.17: 769843-769843 on Build GRCh38
rs150194401 Chr.17: 770231-770233 on Build GRCh38
rs376309803 Chr.17: 769841-769841 on Build GRCh38
rs778303878 Chr.17: 770193-770193 on Build GRCh38
rs139770786 Chr.17: 769953-769953 on Build GRCh38
rs750920820 Chr.17: 770154-770154 on Build GRCh38
rs777355323 Chr.17: 770009-770009 on Build GRCh38
rs777458707 Chr.17: 770178-770178 on Build GRCh38
rs752349761 Chr.17: 770084-770084 on Build GRCh38
rs889678468 Chr.17: 769814-769814 on Build GRCh38
rs778227509 Chr.17: 769883-769883 on Build GRCh38
rs932072660 Chr.17: 770106-770106 on Build GRCh38
rs879164893 Chr.17: 770026-770026 on Build GRCh38
rs141571460 Chr.17: 769932-769932 on Build GRCh38
rs567463779 Chr.17: 770140-770140 on Build GRCh38
rs752768207 Chr.17: 769869-769869 on Build GRCh38
rs757518800 Chr.17: 769890-769890 on Build GRCh38

View Full Product Details