Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 775636-776153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLOD4
Entrez Gene ID
51031
Gene Name
glyoxalase domain containing 4
Gene Aliases
C17orf25, CGI-150, HC71
UniGene
Hs.279061
Gene Chromosome Location
Chr.17: 759309-782331 on Build GRCh38
Transcripts
NM_016080.3 XM_006721540.2 XM_017024711.1 XM_017024712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
347714 c.334G>A p.E112K Chr.17: 775847-775847 on Build GRCh38
3370878 c.388C>T p.R130C Chr.17: 775793-775793 on Build GRCh38
4069090 c.314C>T p.T105M Chr.17: 775912-775912 on Build GRCh38
4270519 c.340A>T p.N114Y Chr.17: 775886-775886 on Build GRCh38
3726678 c.395T>C p.L132P Chr.17: 775786-775786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762427534 Chr.17: 775968-775968 on Build GRCh38
rs759388702 Chr.17: 775940-775940 on Build GRCh38
rs956715833 Chr.17: 776026-776026 on Build GRCh38
rs777149421 Chr.17: 775963-775963 on Build GRCh38
rs1012490621 Chr.17: 775666-775666 on Build GRCh38
rs763178751 Chr.17: 775912-775913 on Build GRCh38
rs747682880 Chr.17: 775912-775912 on Build GRCh38
rs977201513 Chr.17: 776021-776021 on Build GRCh38
rs534554788 Chr.17: 775959-775959 on Build GRCh38
rs560691640 Chr.17: 775693-775693 on Build GRCh38
rs755109464 Chr.17: 775731-775731 on Build GRCh38
rs948072197 Chr.17: 775719-775719 on Build GRCh38
rs759803686 Chr.17: 775945-775945 on Build GRCh38
rs912375989 Chr.17: 775805-775805 on Build GRCh38
rs560934260 Chr.17: 775786-775786 on Build GRCh38
rs778212178 Chr.17: 775736-775736 on Build GRCh38
rs769238015 Chr.17: 775922-775922 on Build GRCh38
rs147622792 Chr.17: 775825-775825 on Build GRCh38
rs375635480 Chr.17: 775817-775817 on Build GRCh38
rs770288916 Chr.17: 775768-775768 on Build GRCh38
rs770654132 Chr.17: 775938-775938 on Build GRCh38
rs967935229 Chr.17: 775978-775983 on Build GRCh38
rs1006355070 Chr.17: 775687-775687 on Build GRCh38
rs753144521 Chr.17: 775806-775806 on Build GRCh38
rs370083059 Chr.17: 775971-775971 on Build GRCh38
rs762724986 Chr.17: 775776-775776 on Build GRCh38
rs374873979 Chr.17: 775746-775746 on Build GRCh38
rs149237575 Chr.17: 775884-775884 on Build GRCh38
rs142062507 Chr.17: 775830-775830 on Build GRCh38
rs139627395 Chr.17: 775810-775810 on Build GRCh38
rs150125013 Chr.17: 775857-775857 on Build GRCh38
rs998049745 Chr.17: 775694-775694 on Build GRCh38
rs2295476 Chr.17: 775858-775858 on Build GRCh38
rs781713902 Chr.17: 775692-775693 on Build GRCh38
rs773065202 Chr.17: 775772-775772 on Build GRCh38
rs547985715 Chr.17: 775800-775800 on Build GRCh38
rs138466937 Chr.17: 775871-775871 on Build GRCh38
rs560039548 Chr.17: 775808-775808 on Build GRCh38
rs757607791 Chr.17: 775819-775819 on Build GRCh38
rs1030821058 Chr.17: 776014-776014 on Build GRCh38
rs777670927 Chr.17: 775728-775728 on Build GRCh38
rs781614495 Chr.17: 775670-775670 on Build GRCh38
rs979780458 Chr.17: 775999-776005 on Build GRCh38
rs768342828 Chr.17: 775956-775956 on Build GRCh38
rs777724634 Chr.17: 775926-775926 on Build GRCh38
rs771117299 Chr.17: 775907-775907 on Build GRCh38
rs917618271 Chr.17: 776119-776119 on Build GRCh38
rs764637083 Chr.17: 775895-775895 on Build GRCh38
rs754682367 Chr.17: 775908-775908 on Build GRCh38
rs1015517225 Chr.17: 775691-775691 on Build GRCh38
rs768237197 Chr.17: 775829-775829 on Build GRCh38
rs745441340 Chr.17: 775741-775741 on Build GRCh38
rs761835834 Chr.17: 775771-775771 on Build GRCh38
rs781640466 Chr.17: 775732-775732 on Build GRCh38
rs372677368 Chr.17: 775962-775962 on Build GRCh38
rs567400465 Chr.17: 775934-775934 on Build GRCh38
rs545030100 Chr.17: 775701-775701 on Build GRCh38
rs776674660 Chr.17: 775766-775766 on Build GRCh38
rs767517153 Chr.17: 775881-775881 on Build GRCh38
rs774506220 Chr.17: 775869-775869 on Build GRCh38
rs888049733 Chr.17: 775676-775676 on Build GRCh38
rs768325163 Chr.17: 775757-775757 on Build GRCh38
rs368535297 Chr.17: 775849-775849 on Build GRCh38
rs779824839 Chr.17: 775822-775822 on Build GRCh38
rs752814764 Chr.17: 775883-775883 on Build GRCh38
rs766650206 Chr.17: 775928-775930 on Build GRCh38
rs753519097 Chr.17: 776114-776116 on Build GRCh38
rs766612407 Chr.17: 775784-775784 on Build GRCh38
rs529543488 Chr.17: 775793-775793 on Build GRCh38
rs563149354 Chr.17: 775734-775734 on Build GRCh38
rs992023844 Chr.17: 776056-776056 on Build GRCh38
rs924098232 Chr.17: 776043-776043 on Build GRCh38
rs754346225 Chr.17: 775898-775898 on Build GRCh38
rs748683487 Chr.17: 775733-775733 on Build GRCh38
rs747970458 Chr.17: 775845-775845 on Build GRCh38
rs184202479 Chr.17: 775671-775671 on Build GRCh38
rs772264589 Chr.17: 775944-775944 on Build GRCh38
rs185292592 Chr.17: 775753-775753 on Build GRCh38
rs773083275 Chr.17: 775856-775856 on Build GRCh38
rs779446417 Chr.17: 775901-775901 on Build GRCh38
rs143453373 Chr.17: 775911-775911 on Build GRCh38
rs1030807481 Chr.17: 775951-775951 on Build GRCh38

View Full Product Details