Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1422481-1423025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRK
Entrez Gene ID
1398
Gene Name
CRK proto-oncogene, adaptor protein
Gene Aliases
CRKII, p38
UniGene
Hs.461896
Gene Chromosome Location
Chr.17: 1421353-1456267 on Build GRCh38
Transcripts
NM_005206.4 NM_016823.3

SNP Detail

SNP ID Chromosome Location
rs533245864 Chr.17: 1422974-1422974 on Build GRCh38
rs147372165 Chr.17: 1422893-1422893 on Build GRCh38
rs540777039 Chr.17: 1422789-1422789 on Build GRCh38
rs189061840 Chr.17: 1422815-1422815 on Build GRCh38
rs552774877 Chr.17: 1422864-1422864 on Build GRCh38
rs1021438190 Chr.17: 1422529-1422529 on Build GRCh38
rs769067758 Chr.17: 1422766-1422766 on Build GRCh38
rs534402802 Chr.17: 1422854-1422854 on Build GRCh38
rs573408956 Chr.17: 1422769-1422769 on Build GRCh38
rs34970553 Chr.17: 1422792-1422792 on Build GRCh38
rs35409499 Chr.17: 1422843-1422844 on Build GRCh38
rs549397331 Chr.17: 1422730-1422730 on Build GRCh38
rs757982646 Chr.17: 1422947-1422947 on Build GRCh38
rs749555955 Chr.17: 1422676-1422676 on Build GRCh38
rs553789646 Chr.17: 1422925-1422925 on Build GRCh38
rs545551544 Chr.17: 1422521-1422521 on Build GRCh38
rs563856294 Chr.17: 1422545-1422545 on Build GRCh38
rs757222327 Chr.17: 1422851-1422851 on Build GRCh38
rs968092968 Chr.17: 1422624-1422624 on Build GRCh38
rs980920754 Chr.17: 1422813-1422814 on Build GRCh38
rs909413913 Chr.17: 1422823-1422823 on Build GRCh38
rs149908866 Chr.17: 1422964-1422964 on Build GRCh38
rs988944771 Chr.17: 1422855-1422855 on Build GRCh38
rs1026555241 Chr.17: 1422740-1422740 on Build GRCh38
rs193240256 Chr.17: 1422682-1422682 on Build GRCh38
rs763255570 Chr.17: 1422597-1422597 on Build GRCh38
rs932952106 Chr.17: 1422870-1422870 on Build GRCh38
rs1026064199 Chr.17: 1422972-1422972 on Build GRCh38
rs1044708391 Chr.17: 1422543-1422543 on Build GRCh38
rs879545008 Chr.17: 1422594-1422594 on Build GRCh38
rs567108669 Chr.17: 1422822-1422822 on Build GRCh38
rs889014687 Chr.17: 1422896-1422896 on Build GRCh38
rs61762272 Chr.17: 1422897-1422897 on Build GRCh38
rs1021407302 Chr.17: 1422919-1422919 on Build GRCh38
rs58414824 Chr.17: 1422901-1422901 on Build GRCh38
rs979959950 Chr.17: 1422642-1422642 on Build GRCh38
rs1034740012 Chr.17: 1422687-1422687 on Build GRCh38
rs914722352 Chr.17: 1422858-1422858 on Build GRCh38
rs192161161 Chr.17: 1422907-1422907 on Build GRCh38
rs139110929 Chr.17: 1422927-1422927 on Build GRCh38
rs530466005 Chr.17: 1422808-1422808 on Build GRCh38
rs185435497 Chr.17: 1422759-1422759 on Build GRCh38
rs535791681 Chr.17: 1422922-1422922 on Build GRCh38
rs766776261 Chr.17: 1422697-1422697 on Build GRCh38
rs556560428 Chr.17: 1422902-1422902 on Build GRCh38
rs1056127030 Chr.17: 1422910-1422910 on Build GRCh38

View Full Product Details