Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1436942-1437454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRK
Entrez Gene ID
1398
Gene Name
CRK proto-oncogene, adaptor protein
Gene Aliases
CRKII, p38
UniGene
Hs.461896
Gene Chromosome Location
Chr.17: 1421353-1456267 on Build GRCh38
Transcripts
NM_005206.4 NM_016823.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6043023 c.382A>G p.I128V Chr.17: 1437015-1437015 on Build GRCh38
2738048 c.444G>A p.E148E Chr.17: 1436953-1436953 on Build GRCh38
1640590 c.242-4_242-3insT p.? Chr.17: 1437158-1437159 on Build GRCh38
6359133 c.248G>A p.S83N Chr.17: 1437149-1437149 on Build GRCh38
95819 c.420C>G p.L140L Chr.17: 1436977-1436977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965485586 Chr.17: 1437226-1437226 on Build GRCh38
rs370128419 Chr.17: 1437388-1437388 on Build GRCh38
rs544674176 Chr.17: 1437404-1437405 on Build GRCh38
rs947045345 Chr.17: 1437003-1437003 on Build GRCh38
rs936818635 Chr.17: 1437389-1437389 on Build GRCh38
rs1041967 Chr.17: 1437071-1437071 on Build GRCh38
rs938425892 Chr.17: 1437395-1437395 on Build GRCh38
rs1026873856 Chr.17: 1437187-1437187 on Build GRCh38
rs570938088 Chr.17: 1437381-1437381 on Build GRCh38
rs748384553 Chr.17: 1437020-1437020 on Build GRCh38
rs985436541 Chr.17: 1437190-1437190 on Build GRCh38
rs749892558 Chr.17: 1437081-1437081 on Build GRCh38
rs750129488 Chr.17: 1437200-1437200 on Build GRCh38
rs759517234 Chr.17: 1437396-1437396 on Build GRCh38
rs897014160 Chr.17: 1437153-1437153 on Build GRCh38
rs924095760 Chr.17: 1437278-1437278 on Build GRCh38
rs765166114 Chr.17: 1437414-1437414 on Build GRCh38
rs753041568 Chr.17: 1437053-1437053 on Build GRCh38
rs986781248 Chr.17: 1437279-1437279 on Build GRCh38
rs974170958 Chr.17: 1437236-1437236 on Build GRCh38
rs763079680 Chr.17: 1437023-1437023 on Build GRCh38
rs761736955 Chr.17: 1437109-1437109 on Build GRCh38
rs547133034 Chr.17: 1437431-1437431 on Build GRCh38
rs764912543 Chr.17: 1437196-1437196 on Build GRCh38
rs796296786 Chr.17: 1437219-1437219 on Build GRCh38
rs375463605 Chr.17: 1436997-1436997 on Build GRCh38
rs544154068 Chr.17: 1437320-1437320 on Build GRCh38
rs904025018 Chr.17: 1437008-1437008 on Build GRCh38
rs376047184 Chr.17: 1437049-1437049 on Build GRCh38
rs778455201 Chr.17: 1436984-1436984 on Build GRCh38
rs368288593 Chr.17: 1437252-1437252 on Build GRCh38
rs771019679 Chr.17: 1437171-1437171 on Build GRCh38
rs948308127 Chr.17: 1437351-1437351 on Build GRCh38
rs202210551 Chr.17: 1437177-1437177 on Build GRCh38
rs969370941 Chr.17: 1437162-1437162 on Build GRCh38
rs369347817 Chr.17: 1437183-1437183 on Build GRCh38
rs745434190 Chr.17: 1436988-1436988 on Build GRCh38
rs781014005 Chr.17: 1437144-1437144 on Build GRCh38
rs905572417 Chr.17: 1437054-1437054 on Build GRCh38
rs776671383 Chr.17: 1437014-1437014 on Build GRCh38
rs989620723 Chr.17: 1437303-1437303 on Build GRCh38
rs774845596 Chr.17: 1437031-1437031 on Build GRCh38
rs541131894 Chr.17: 1436995-1436995 on Build GRCh38
rs963186832 Chr.17: 1437204-1437204 on Build GRCh38
rs765178772 Chr.17: 1437418-1437419 on Build GRCh38
rs542938170 Chr.17: 1437344-1437344 on Build GRCh38
rs768530327 Chr.17: 1437004-1437004 on Build GRCh38
rs767900527 Chr.17: 1437050-1437050 on Build GRCh38
rs201648872 Chr.17: 1437058-1437058 on Build GRCh38
rs769953070 Chr.17: 1437021-1437021 on Build GRCh38
rs896919392 Chr.17: 1437418-1437418 on Build GRCh38
rs977272840 Chr.17: 1437256-1437256 on Build GRCh38
rs760956850 Chr.17: 1437192-1437192 on Build GRCh38
rs146930761 Chr.17: 1437022-1437022 on Build GRCh38
rs368739765 Chr.17: 1437001-1437001 on Build GRCh38
rs777985840 Chr.17: 1437160-1437160 on Build GRCh38
rs1035514589 Chr.17: 1437143-1437143 on Build GRCh38
rs747960532 Chr.17: 1437146-1437146 on Build GRCh38
rs766815891 Chr.17: 1437029-1437029 on Build GRCh38
rs935479174 Chr.17: 1437294-1437294 on Build GRCh38
rs765037033 Chr.17: 1437061-1437061 on Build GRCh38
rs772107356 Chr.17: 1436996-1436996 on Build GRCh38
rs527292310 Chr.17: 1437405-1437405 on Build GRCh38
rs200440102 Chr.17: 1437141-1437141 on Build GRCh38
rs770045198 Chr.17: 1437152-1437152 on Build GRCh38
rs749301729 Chr.17: 1437164-1437164 on Build GRCh38
rs552359472 Chr.17: 1437251-1437252 on Build GRCh38
rs562854243 Chr.17: 1437417-1437417 on Build GRCh38
rs925478336 Chr.17: 1437364-1437364 on Build GRCh38
rs529524747 Chr.17: 1437275-1437275 on Build GRCh38
rs780103155 Chr.17: 1437134-1437134 on Build GRCh38
rs951367396 Chr.17: 1437272-1437272 on Build GRCh38
rs563578598 Chr.17: 1437188-1437188 on Build GRCh38
rs11544140 Chr.17: 1437077-1437077 on Build GRCh38
rs757985067 Chr.17: 1437126-1437126 on Build GRCh38
rs1044058587 Chr.17: 1437352-1437352 on Build GRCh38
rs915459612 Chr.17: 1437327-1437327 on Build GRCh38
rs762505287 Chr.17: 1437205-1437205 on Build GRCh38
rs777881954 Chr.17: 1437187-1437188 on Build GRCh38
rs201254340 Chr.17: 1437136-1437136 on Build GRCh38

View Full Product Details