Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 2030032-2030522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH1
Entrez Gene ID
1801
Gene Name
diphthamide biosynthesis 1
Gene Aliases
DEDSSH, DPH2L, DPH2L1, OVCA1
UniGene
Hs.513856
Gene Chromosome Location
Chr.17: 2030137-2043431 on Build GRCh38
Transcripts
NM_001383.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7365356 c.35C>G p.S12C Chr.17: 2030189-2030189 on Build GRCh38
84363 c.35C>T p.S12F Chr.17: 2030189-2030189 on Build GRCh38
976413 c.14T>C p.V5A Chr.17: 2030168-2030168 on Build GRCh38
4495256 c.44C>T p.A15V Chr.17: 2030198-2030198 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764674601 Chr.17: 2030139-2030139 on Build GRCh38
rs759161613 Chr.17: 2030093-2030093 on Build GRCh38
rs767067132 Chr.17: 2030101-2030101 on Build GRCh38
rs371007597 Chr.17: 2030159-2030159 on Build GRCh38
rs376971506 Chr.17: 2030217-2030217 on Build GRCh38
rs757167361 Chr.17: 2030171-2030171 on Build GRCh38
rs757220221 Chr.17: 2030261-2030261 on Build GRCh38
rs915015540 Chr.17: 2030488-2030488 on Build GRCh38
rs969706846 Chr.17: 2030158-2030158 on Build GRCh38
rs1018109185 Chr.17: 2030295-2030295 on Build GRCh38
rs777260398 Chr.17: 2030169-2030169 on Build GRCh38
rs368768635 Chr.17: 2030058-2030058 on Build GRCh38
rs969759196 Chr.17: 2030450-2030450 on Build GRCh38
rs750242772 Chr.17: 2030272-2030272 on Build GRCh38
rs544426347 Chr.17: 2030492-2030492 on Build GRCh38
rs993979986 Chr.17: 2030080-2030080 on Build GRCh38
rs772138366 Chr.17: 2030117-2030117 on Build GRCh38
rs148835478 Chr.17: 2030441-2030441 on Build GRCh38
rs534743578 Chr.17: 2030155-2030155 on Build GRCh38
rs763646344 Chr.17: 2030212-2030212 on Build GRCh38
rs1014134595 Chr.17: 2030400-2030400 on Build GRCh38
rs969522520 Chr.17: 2030405-2030405 on Build GRCh38
rs930717263 Chr.17: 2030225-2030225 on Build GRCh38
rs753299811 Chr.17: 2030249-2030249 on Build GRCh38
rs754680419 Chr.17: 2030156-2030156 on Build GRCh38
rs565523696 Chr.17: 2030136-2030136 on Build GRCh38
rs897906177 Chr.17: 2030072-2030072 on Build GRCh38
rs895632140 Chr.17: 2030377-2030377 on Build GRCh38
rs766753554 Chr.17: 2030199-2030199 on Build GRCh38
rs553434451 Chr.17: 2030371-2030371 on Build GRCh38
rs1036777488 Chr.17: 2030205-2030205 on Build GRCh38
rs762458101 Chr.17: 2030192-2030192 on Build GRCh38
rs757704030 Chr.17: 2030213-2030213 on Build GRCh38
rs747238195 Chr.17: 2030184-2030184 on Build GRCh38
rs972741323 Chr.17: 2030196-2030196 on Build GRCh38
rs758182036 Chr.17: 2030274-2030274 on Build GRCh38
rs772341134 Chr.17: 2030177-2030177 on Build GRCh38
rs1006751826 Chr.17: 2030119-2030119 on Build GRCh38
rs573155253 Chr.17: 2030378-2030378 on Build GRCh38
rs751537348 Chr.17: 2030071-2030072 on Build GRCh38
rs199608890 Chr.17: 2030362-2030363 on Build GRCh38
rs746138840 Chr.17: 2030105-2030105 on Build GRCh38
rs377506869 Chr.17: 2030133-2030133 on Build GRCh38
rs1051795860 Chr.17: 2030235-2030235 on Build GRCh38
rs773741408 Chr.17: 2030194-2030194 on Build GRCh38
rs375040102 Chr.17: 2030204-2030204 on Build GRCh38
rs574002137 Chr.17: 2030330-2030330 on Build GRCh38
rs776821196 Chr.17: 2030191-2030191 on Build GRCh38
rs980938566 Chr.17: 2030482-2030482 on Build GRCh38
rs1036423573 Chr.17: 2030065-2030065 on Build GRCh38
rs754283257 Chr.17: 2030198-2030198 on Build GRCh38
rs764601941 Chr.17: 2030175-2030175 on Build GRCh38
rs1006699574 Chr.17: 2030284-2030284 on Build GRCh38
rs575648592 Chr.17: 2030472-2030472 on Build GRCh38
rs762400906 Chr.17: 2030143-2030143 on Build GRCh38
rs571457784 Chr.17: 2030188-2030188 on Build GRCh38
rs943465744 Chr.17: 2030290-2030290 on Build GRCh38
rs923680458 Chr.17: 2030152-2030152 on Build GRCh38
rs998093399 Chr.17: 2030333-2030333 on Build GRCh38
rs868253226 Chr.17: 2030363-2030363 on Build GRCh38
rs901107269 Chr.17: 2030300-2030300 on Build GRCh38
rs145623725 Chr.17: 2030342-2030342 on Build GRCh38
rs767013444 Chr.17: 2030157-2030157 on Build GRCh38
rs752358862 Chr.17: 2030202-2030202 on Build GRCh38
rs752205738 Chr.17: 2030112-2030112 on Build GRCh38
rs779890763 Chr.17: 2030282-2030282 on Build GRCh38
rs2273981 Chr.17: 2030087-2030087 on Build GRCh38
rs765788770 Chr.17: 2030193-2030193 on Build GRCh38
rs1005326446 Chr.17: 2030483-2030483 on Build GRCh38
rs545525992 Chr.17: 2030404-2030404 on Build GRCh38
rs557144639 Chr.17: 2030270-2030270 on Build GRCh38
rs749267261 Chr.17: 2030170-2030170 on Build GRCh38
rs541900220 Chr.17: 2030164-2030164 on Build GRCh38
rs775513728 Chr.17: 2030182-2030182 on Build GRCh38
rs765733518 Chr.17: 2030149-2030149 on Build GRCh38
rs778705666 Chr.17: 2030174-2030174 on Build GRCh38
rs879092660 Chr.17: 2030411-2030411 on Build GRCh38
rs777334847 Chr.17: 2030141-2030141 on Build GRCh38
rs2273982 Chr.17: 2030070-2030070 on Build GRCh38

View Full Product Details