Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3489102-3489525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPA
Entrez Gene ID
443
Gene Name
aspartoacylase
Gene Aliases
ACY2, ASP
UniGene
Hs.171142
Gene Chromosome Location
Chr.17: 3474106-3502534 on Build GRCh38
Transcripts
NM_000049.2 NM_001128085.1 XM_017024661.1 NM_001321336.1 NM_001321337.1

Gene Symbol
SPATA22
Entrez Gene ID
84690
Gene Name
spermatogenesis associated 22
Gene Aliases
NYD-SP20, NYDSP20
UniGene
Hs.351068
Gene Chromosome Location
Chr.17: 3440011-3513852 on Build GRCh38
Transcripts
NM_000049.2 NM_001128085.1 XM_017024661.1 NM_001321336.1 NM_001321337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5758340 c.580C>G p.Q194E Chr.17: 3489288-3489288 on Build GRCh38
6333192 c.634+1G>T p.? Chr.17: 3489343-3489343 on Build GRCh38
6362582 c.560T>C p.L187P Chr.17: 3489268-3489268 on Build GRCh38
344509 c.587G>A p.R196K Chr.17: 3489295-3489295 on Build GRCh38
6404964 c.556G>A p.V186I Chr.17: 3489264-3489264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772264936 Chr.17: 3489261-3489261 on Build GRCh38
rs544964870 Chr.17: 3489400-3489400 on Build GRCh38
rs763251910 Chr.17: 3489315-3489315 on Build GRCh38
rs747825306 Chr.17: 3489156-3489156 on Build GRCh38
rs753389782 Chr.17: 3489215-3489215 on Build GRCh38
rs769136884 Chr.17: 3489277-3489277 on Build GRCh38
rs375663977 Chr.17: 3489212-3489212 on Build GRCh38
rs745553536 Chr.17: 3489244-3489244 on Build GRCh38
rs778385612 Chr.17: 3489233-3489233 on Build GRCh38
rs778693280 Chr.17: 3489343-3489346 on Build GRCh38
rs772023643 Chr.17: 3489365-3489365 on Build GRCh38
rs573646986 Chr.17: 3489168-3489168 on Build GRCh38
rs767152579 Chr.17: 3489320-3489320 on Build GRCh38
rs373381764 Chr.17: 3489385-3489385 on Build GRCh38
rs780468376 Chr.17: 3489262-3489262 on Build GRCh38
rs765354678 Chr.17: 3489188-3489188 on Build GRCh38
rs868350167 Chr.17: 3489426-3489426 on Build GRCh38
rs761849556 Chr.17: 3489310-3489310 on Build GRCh38
rs567140525 Chr.17: 3489152-3489152 on Build GRCh38
rs762720755 Chr.17: 3489403-3489403 on Build GRCh38
rs534647503 Chr.17: 3489158-3489158 on Build GRCh38
rs786204572 Chr.17: 3489249-3489249 on Build GRCh38
rs372085439 Chr.17: 3489202-3489202 on Build GRCh38
rs769947909 Chr.17: 3489311-3489311 on Build GRCh38
rs147763700 Chr.17: 3489312-3489312 on Build GRCh38
rs911346550 Chr.17: 3489414-3489414 on Build GRCh38
rs777170306 Chr.17: 3489299-3489299 on Build GRCh38
rs73311007 Chr.17: 3489169-3489169 on Build GRCh38
rs976795599 Chr.17: 3489457-3489457 on Build GRCh38
rs756844626 Chr.17: 3489221-3489221 on Build GRCh38
rs369805590 Chr.17: 3489329-3489329 on Build GRCh38
rs769048944 Chr.17: 3489373-3489373 on Build GRCh38
rs1012357349 Chr.17: 3489178-3489178 on Build GRCh38
rs749974010 Chr.17: 3489363-3489363 on Build GRCh38
rs756669857 Chr.17: 3489345-3489345 on Build GRCh38
rs372987490 Chr.17: 3489376-3489376 on Build GRCh38
rs745752959 Chr.17: 3489360-3489360 on Build GRCh38
rs944114693 Chr.17: 3489425-3489425 on Build GRCh38
rs1023365853 Chr.17: 3489200-3489200 on Build GRCh38
rs764104886 Chr.17: 3489341-3489341 on Build GRCh38
rs750698855 Chr.17: 3489193-3489193 on Build GRCh38
rs536855227 Chr.17: 3489128-3489128 on Build GRCh38
rs1014551540 Chr.17: 3489247-3489247 on Build GRCh38
rs752538129 Chr.17: 3489325-3489325 on Build GRCh38
rs747438350 Chr.17: 3489265-3489265 on Build GRCh38
rs754689698 Chr.17: 3489198-3489198 on Build GRCh38
rs779738539 Chr.17: 3489367-3489367 on Build GRCh38
rs973361661 Chr.17: 3489349-3489349 on Build GRCh38
rs902481026 Chr.17: 3489138-3489138 on Build GRCh38
rs374512344 Chr.17: 3489366-3489366 on Build GRCh38
rs778496275 Chr.17: 3489350-3489350 on Build GRCh38
rs368144957 Chr.17: 3489201-3489201 on Build GRCh38
rs753871454 Chr.17: 3489343-3489343 on Build GRCh38

View Full Product Details