Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3876101-3876604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMKK1
Entrez Gene ID
84254
Gene Name
calcium/calmodulin dependent protein kinase kinase 1
Gene Aliases
CAMKKA
UniGene
Hs.8417
Gene Chromosome Location
Chr.17: 3860315-3894891 on Build GRCh38
Transcripts
NM_032294.2 NM_172206.1 NM_172207.2 XM_005256817.1 XM_006721588.2 XM_011524031.2 XM_011524033.1 XM_011524035.2 XM_017025219.1 XM_017025220.1 XM_017025221.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6146687 c.805C>T p.Q269* Chr.17: 3876414-3876414 on Build GRCh38
6146686 c.919C>T p.Q307* Chr.17: 3876414-3876414 on Build GRCh38
7086517 c.810G>C p.K270N Chr.17: 3876409-3876409 on Build GRCh38
7122706 c.933G>T p.A311A Chr.17: 3876286-3876286 on Build GRCh38
4930252 c.1089C>A p.S363S Chr.17: 3876244-3876244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560406629 Chr.17: 3876244-3876244 on Build GRCh38
rs976859547 Chr.17: 3876566-3876566 on Build GRCh38
rs79056067 Chr.17: 3876198-3876198 on Build GRCh38
rs767490064 Chr.17: 3876306-3876306 on Build GRCh38
rs915865133 Chr.17: 3876456-3876456 on Build GRCh38
rs754154039 Chr.17: 3876178-3876178 on Build GRCh38
rs548648072 Chr.17: 3876354-3876354 on Build GRCh38
rs371028799 Chr.17: 3876303-3876303 on Build GRCh38
rs547772096 Chr.17: 3876483-3876483 on Build GRCh38
rs915750810 Chr.17: 3876179-3876179 on Build GRCh38
rs148844492 Chr.17: 3876289-3876289 on Build GRCh38
rs756267627 Chr.17: 3876310-3876310 on Build GRCh38
rs763372597 Chr.17: 3876286-3876286 on Build GRCh38
rs960048193 Chr.17: 3876346-3876346 on Build GRCh38
rs919595430 Chr.17: 3876506-3876506 on Build GRCh38
rs764163624 Chr.17: 3876458-3876458 on Build GRCh38
rs969969307 Chr.17: 3876579-3876579 on Build GRCh38
rs554642672 Chr.17: 3876545-3876545 on Build GRCh38
rs558906016 Chr.17: 3876509-3876509 on Build GRCh38
rs752862171 Chr.17: 3876307-3876307 on Build GRCh38
rs534613302 Chr.17: 3876466-3876466 on Build GRCh38
rs905031623 Chr.17: 3876484-3876484 on Build GRCh38
rs377322922 Chr.17: 3876472-3876472 on Build GRCh38
rs752145624 Chr.17: 3876182-3876182 on Build GRCh38
rs370912763 Chr.17: 3876262-3876262 on Build GRCh38
rs931006520 Chr.17: 3876507-3876507 on Build GRCh38
rs893134898 Chr.17: 3876538-3876538 on Build GRCh38
rs753165786 Chr.17: 3876439-3876439 on Build GRCh38
rs377491155 Chr.17: 3876355-3876355 on Build GRCh38
rs867486831 Chr.17: 3876322-3876322 on Build GRCh38
rs752055786 Chr.17: 3876290-3876290 on Build GRCh38
rs755567907 Chr.17: 3876294-3876294 on Build GRCh38
rs754950220 Chr.17: 3876184-3876184 on Build GRCh38
rs200680352 Chr.17: 3876216-3876216 on Build GRCh38
rs777922449 Chr.17: 3876327-3876327 on Build GRCh38
rs766837051 Chr.17: 3876403-3876403 on Build GRCh38
rs185443583 Chr.17: 3876214-3876214 on Build GRCh38
rs901893777 Chr.17: 3876525-3876525 on Build GRCh38
rs567177203 Chr.17: 3876305-3876305 on Build GRCh38
rs1026104071 Chr.17: 3876513-3876513 on Build GRCh38
rs999106892 Chr.17: 3876205-3876205 on Build GRCh38
rs377123035 Chr.17: 3876243-3876243 on Build GRCh38
rs762711219 Chr.17: 3876388-3876388 on Build GRCh38
rs1032048751 Chr.17: 3876531-3876531 on Build GRCh38
rs189227649 Chr.17: 3876560-3876560 on Build GRCh38
rs766276238 Chr.17: 3876449-3876465 on Build GRCh38
rs145599553 Chr.17: 3876510-3876510 on Build GRCh38
rs756426484 Chr.17: 3876204-3876204 on Build GRCh38
rs761363870 Chr.17: 3876247-3876247 on Build GRCh38
rs781146888 Chr.17: 3876471-3876471 on Build GRCh38
rs957249192 Chr.17: 3876409-3876409 on Build GRCh38
rs568489467 Chr.17: 3876406-3876406 on Build GRCh38
rs370732140 Chr.17: 3876487-3876487 on Build GRCh38
rs575530601 Chr.17: 3876157-3876157 on Build GRCh38
rs775365245 Chr.17: 3876218-3876218 on Build GRCh38
rs1018275342 Chr.17: 3876550-3876550 on Build GRCh38
rs370319027 Chr.17: 3876389-3876389 on Build GRCh38
rs543364320 Chr.17: 3876481-3876482 on Build GRCh38
rs1022820633 Chr.17: 3876567-3876567 on Build GRCh38
rs990027389 Chr.17: 3876410-3876410 on Build GRCh38
rs762978384 Chr.17: 3876200-3876237 on Build GRCh38
rs772937558 Chr.17: 3876366-3876366 on Build GRCh38
rs541466139 Chr.17: 3876165-3876165 on Build GRCh38
rs756274484 Chr.17: 3876141-3876141 on Build GRCh38
rs537670949 Chr.17: 3876514-3876514 on Build GRCh38
rs144606226 Chr.17: 3876405-3876405 on Build GRCh38
rs140330846 Chr.17: 3876353-3876353 on Build GRCh38
rs373496993 Chr.17: 3876334-3876334 on Build GRCh38
rs1041273605 Chr.17: 3876190-3876190 on Build GRCh38
rs137879236 Chr.17: 3876343-3876343 on Build GRCh38
rs768119619 Chr.17: 3876433-3876433 on Build GRCh38
rs1049220861 Chr.17: 3876287-3876287 on Build GRCh38
rs757529098 Chr.17: 3876465-3876465 on Build GRCh38
rs374879530 Chr.17: 3876469-3876469 on Build GRCh38
rs751408563 Chr.17: 3876468-3876468 on Build GRCh38
rs1023929150 Chr.17: 3876265-3876265 on Build GRCh38
rs1040565194 Chr.17: 3876523-3876523 on Build GRCh38
rs558736763 Chr.17: 3876140-3876140 on Build GRCh38
rs779254375 Chr.17: 3876445-3876445 on Build GRCh38
rs570861109 Chr.17: 3876486-3876486 on Build GRCh38
rs200152858 Chr.17: 3876333-3876333 on Build GRCh38
rs552857978 Chr.17: 3876527-3876527 on Build GRCh38
rs538943427 Chr.17: 3876134-3876134 on Build GRCh38
rs758954787 Chr.17: 3876172-3876172 on Build GRCh38
rs746903648 Chr.17: 3876345-3876345 on Build GRCh38
rs181847866 Chr.17: 3876185-3876185 on Build GRCh38
rs1007764338 Chr.17: 3876313-3876313 on Build GRCh38
rs935025423 Chr.17: 3876451-3876451 on Build GRCh38
rs554992311 Chr.17: 3876226-3876226 on Build GRCh38
rs779113525 Chr.17: 3876179-3876180 on Build GRCh38
rs745711852 Chr.17: 3876215-3876215 on Build GRCh38
rs764993423 Chr.17: 3876261-3876261 on Build GRCh38
rs145381314 Chr.17: 3876342-3876342 on Build GRCh38

View Full Product Details