Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4142667-4143159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5D2
Entrez Gene ID
124936
Gene Name
cytochrome b5 domain containing 2
Gene Aliases
-
UniGene
Hs.513871
Gene Chromosome Location
Chr.17: 4143117-4157701 on Build GRCh38
Transcripts
XM_011523649.2 XM_017024180.1 XM_017024181.1 NM_015113.3 XM_005256561.2 XM_011523759.2 XM_017024382.1 XM_017024383.1

Gene Symbol
ZZEF1
Entrez Gene ID
23140
Gene Name
zinc finger ZZ-type and EF-hand domain containing 1
Gene Aliases
ZZZ4
UniGene
Hs.277624
Gene Chromosome Location
Chr.17: 4004445-4143020 on Build GRCh38
Transcripts
XM_011523649.2 XM_017024180.1 XM_017024181.1 NM_015113.3 XM_005256561.2 XM_011523759.2 XM_017024382.1 XM_017024383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4745513 c.185C>T p.A62V Chr.17: 4142711-4142711 on Build GRCh38
6421707 c.73C>T p.Q25* Chr.17: 4142823-4142823 on Build GRCh38
4745512 c.204delC p.C69fs*84 Chr.17: 4142692-4142692 on Build GRCh38
6433541 c.85G>A p.A29T Chr.17: 4142811-4142811 on Build GRCh38
3755556 c.89T>C p.V30A Chr.17: 4142807-4142807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050221828 Chr.17: 4142956-4142956 on Build GRCh38
rs907877295 Chr.17: 4142887-4142887 on Build GRCh38
rs962371059 Chr.17: 4143013-4143013 on Build GRCh38
rs751858422 Chr.17: 4142740-4142740 on Build GRCh38
rs562000574 Chr.17: 4142962-4142962 on Build GRCh38
rs999426774 Chr.17: 4142991-4142991 on Build GRCh38
rs1454121 Chr.17: 4142807-4142807 on Build GRCh38
rs957899497 Chr.17: 4143002-4143002 on Build GRCh38
rs529354624 Chr.17: 4142824-4142824 on Build GRCh38
rs756254957 Chr.17: 4142764-4142764 on Build GRCh38
rs190125447 Chr.17: 4142977-4142977 on Build GRCh38
rs970187606 Chr.17: 4143061-4143061 on Build GRCh38
rs780599502 Chr.17: 4142924-4142925 on Build GRCh38
rs931881415 Chr.17: 4143078-4143078 on Build GRCh38
rs781709310 Chr.17: 4142758-4142758 on Build GRCh38
rs940649185 Chr.17: 4142898-4142898 on Build GRCh38
rs920500639 Chr.17: 4142918-4142918 on Build GRCh38
rs986003705 Chr.17: 4143079-4143079 on Build GRCh38
rs749543043 Chr.17: 4142768-4142768 on Build GRCh38
rs755387517 Chr.17: 4142870-4142870 on Build GRCh38
rs753713228 Chr.17: 4142703-4142703 on Build GRCh38
rs746867323 Chr.17: 4142785-4142785 on Build GRCh38
rs761443139 Chr.17: 4142710-4142710 on Build GRCh38
rs115973899 Chr.17: 4142983-4142983 on Build GRCh38
rs970824902 Chr.17: 4142993-4142993 on Build GRCh38
rs558390947 Chr.17: 4143138-4143138 on Build GRCh38
rs918748835 Chr.17: 4143103-4143103 on Build GRCh38
rs549391713 Chr.17: 4142936-4142936 on Build GRCh38
rs1019913594 Chr.17: 4142820-4142820 on Build GRCh38
rs535023610 Chr.17: 4143116-4143116 on Build GRCh38
rs565642303 Chr.17: 4143108-4143108 on Build GRCh38
rs376323975 Chr.17: 4142780-4142780 on Build GRCh38
rs757938913 Chr.17: 4142716-4142716 on Build GRCh38
rs535119753 Chr.17: 4143018-4143018 on Build GRCh38
rs111724159 Chr.17: 4142692-4142692 on Build GRCh38
rs574053683 Chr.17: 4143102-4143102 on Build GRCh38
rs1014902950 Chr.17: 4143007-4143007 on Build GRCh38
rs980130009 Chr.17: 4142927-4142929 on Build GRCh38
rs764623154 Chr.17: 4142712-4142712 on Build GRCh38
rs982152462 Chr.17: 4143004-4143004 on Build GRCh38
rs965266912 Chr.17: 4142830-4142830 on Build GRCh38
rs768555831 Chr.17: 4142800-4142800 on Build GRCh38
rs767834166 Chr.17: 4142884-4142884 on Build GRCh38
rs554927926 Chr.17: 4143066-4143066 on Build GRCh38
rs766091266 Chr.17: 4142829-4142829 on Build GRCh38
rs920515798 Chr.17: 4143073-4143073 on Build GRCh38
rs1041439677 Chr.17: 4142975-4142975 on Build GRCh38
rs1030805028 Chr.17: 4142895-4142896 on Build GRCh38
rs558588815 Chr.17: 4142926-4142927 on Build GRCh38
rs1010748498 Chr.17: 4143015-4143021 on Build GRCh38
rs907460324 Chr.17: 4142702-4142702 on Build GRCh38
rs569407442 Chr.17: 4143015-4143015 on Build GRCh38
rs775428409 Chr.17: 4142784-4142784 on Build GRCh38
rs942477166 Chr.17: 4143124-4143124 on Build GRCh38
rs990217583 Chr.17: 4142917-4142917 on Build GRCh38
rs938930177 Chr.17: 4142944-4142944 on Build GRCh38
rs1037223434 Chr.17: 4142756-4142756 on Build GRCh38
rs758883508 Chr.17: 4142691-4142692 on Build GRCh38
rs760701992 Chr.17: 4142694-4142694 on Build GRCh38
rs918784911 Chr.17: 4142969-4142969 on Build GRCh38
rs572396045 Chr.17: 4142693-4142693 on Build GRCh38
rs1454122 Chr.17: 4142976-4142976 on Build GRCh38
rs777728798 Chr.17: 4142799-4142799 on Build GRCh38
rs578098174 Chr.17: 4142772-4142772 on Build GRCh38
rs931821432 Chr.17: 4142946-4142946 on Build GRCh38
rs890214749 Chr.17: 4142963-4142963 on Build GRCh38
rs1037191075 Chr.17: 4142952-4142952 on Build GRCh38
rs928651155 Chr.17: 4143083-4143083 on Build GRCh38
rs1048556000 Chr.17: 4143115-4143115 on Build GRCh38
rs563727364 Chr.17: 4142813-4142813 on Build GRCh38
rs973712358 Chr.17: 4142909-4142909 on Build GRCh38
rs973400692 Chr.17: 4143050-4143050 on Build GRCh38
rs776678546 Chr.17: 4142801-4142801 on Build GRCh38
rs751201927 Chr.17: 4142837-4142837 on Build GRCh38
rs114582898 Chr.17: 4142974-4142974 on Build GRCh38
rs747088691 Chr.17: 4142944-4142945 on Build GRCh38
rs762602976 Chr.17: 4142814-4142814 on Build GRCh38
rs1033024158 Chr.17: 4142978-4142978 on Build GRCh38
rs960261855 Chr.17: 4143096-4143096 on Build GRCh38
rs777672885 Chr.17: 4142767-4142767 on Build GRCh38
rs944466553 Chr.17: 4143126-4143126 on Build GRCh38
rs766744303 Chr.17: 4142721-4142721 on Build GRCh38
rs549467104 Chr.17: 4142992-4142992 on Build GRCh38
rs769751993 Chr.17: 4142688-4142688 on Build GRCh38
rs768190777 Chr.17: 4143109-4143109 on Build GRCh38
rs928858595 Chr.17: 4142927-4142927 on Build GRCh38
rs1012377000 Chr.17: 4142987-4142987 on Build GRCh38
rs1022089604 Chr.17: 4143041-4143041 on Build GRCh38
rs771215770 Chr.17: 4142776-4142776 on Build GRCh38
rs199847329 Chr.17: 4142696-4142696 on Build GRCh38
rs199896183 Chr.17: 4142760-4142760 on Build GRCh38
rs755336147 Chr.17: 4142741-4142741 on Build GRCh38

View Full Product Details