Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4157162-4157654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5D2
Entrez Gene ID
124936
Gene Name
cytochrome b5 domain containing 2
Gene Aliases
-
UniGene
Hs.513871
Gene Chromosome Location
Chr.17: 4143117-4157701 on Build GRCh38
Transcripts
NM_001254755.1 NM_001254756.1 NM_144611.3 XM_006721447.2 XM_011523649.2 XM_017024180.1 XM_017024181.1 XM_017024182.1

SNP Detail

SNP ID Chromosome Location
rs1009503651 Chr.17: 4157300-4157300 on Build GRCh38
rs1046113905 Chr.17: 4157255-4157255 on Build GRCh38
rs778070040 Chr.17: 4157628-4157628 on Build GRCh38
rs780773520 Chr.17: 4157402-4157402 on Build GRCh38
rs146738774 Chr.17: 4157375-4157375 on Build GRCh38
rs183702678 Chr.17: 4157356-4157356 on Build GRCh38
rs985511474 Chr.17: 4157226-4157226 on Build GRCh38
rs562508546 Chr.17: 4157550-4157550 on Build GRCh38
rs779444606 Chr.17: 4157252-4157254 on Build GRCh38
rs965532455 Chr.17: 4157486-4157486 on Build GRCh38
rs3087605 Chr.17: 4157285-4157285 on Build GRCh38
rs895291683 Chr.17: 4157249-4157249 on Build GRCh38
rs573746057 Chr.17: 4157245-4157245 on Build GRCh38
rs923932918 Chr.17: 4157497-4157497 on Build GRCh38
rs1028790416 Chr.17: 4157199-4157199 on Build GRCh38
rs936682486 Chr.17: 4157506-4157506 on Build GRCh38
rs917175315 Chr.17: 4157436-4157436 on Build GRCh38
rs188179717 Chr.17: 4157439-4157439 on Build GRCh38
rs748804658 Chr.17: 4157631-4157631 on Build GRCh38
rs755366728 Chr.17: 4157378-4157378 on Build GRCh38
rs772771865 Chr.17: 4157604-4157604 on Build GRCh38
rs193272839 Chr.17: 4157492-4157492 on Build GRCh38
rs560463906 Chr.17: 4157442-4157442 on Build GRCh38
rs527824353 Chr.17: 4157441-4157441 on Build GRCh38
rs953166772 Chr.17: 4157399-4157399 on Build GRCh38
rs554562942 Chr.17: 4157187-4157187 on Build GRCh38
rs1015243846 Chr.17: 4157181-4157181 on Build GRCh38
rs554038718 Chr.17: 4157231-4157231 on Build GRCh38
rs958736605 Chr.17: 4157432-4157432 on Build GRCh38
rs948242885 Chr.17: 4157249-4157249 on Build GRCh38
rs936901029 Chr.17: 4157246-4157246 on Build GRCh38
rs149718656 Chr.17: 4157239-4157239 on Build GRCh38
rs899509383 Chr.17: 4157243-4157243 on Build GRCh38
rs991481675 Chr.17: 4157435-4157435 on Build GRCh38
rs906321790 Chr.17: 4157259-4157259 on Build GRCh38
rs747342124 Chr.17: 4157348-4157348 on Build GRCh38
rs139545488 Chr.17: 4157190-4157190 on Build GRCh38
rs758933068 Chr.17: 4157248-4157248 on Build GRCh38
rs1049523 Chr.17: 4157188-4157188 on Build GRCh38
rs190648638 Chr.17: 4157308-4157308 on Build GRCh38
rs186920254 Chr.17: 4157549-4157549 on Build GRCh38
rs577062772 Chr.17: 4157238-4157238 on Build GRCh38
rs994729580 Chr.17: 4157320-4157320 on Build GRCh38
rs185418014 Chr.17: 4157540-4157540 on Build GRCh38
rs953199003 Chr.17: 4157202-4157202 on Build GRCh38
rs540874651 Chr.17: 4157272-4157272 on Build GRCh38

View Full Product Details