Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4631939-4632429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX15
Entrez Gene ID
246
Gene Name
arachidonate 15-lipoxygenase
Gene Aliases
12-LOX, 15-LOX-1, 15LOX-1
UniGene
Hs.73809
Gene Chromosome Location
Chr.17: 4630919-4641665 on Build GRCh38
Transcripts
NM_001140.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1479677 c.1621G>A p.A541T Chr.17: 4632201-4632201 on Build GRCh38
6598688 c.1716G>A p.T572T Chr.17: 4631982-4631982 on Build GRCh38
4067221 c.1750C>T p.R584W Chr.17: 4632014-4632014 on Build GRCh38
2696324 c.1746G>A p.T582T Chr.17: 4632018-4632018 on Build GRCh38
6851226 c.1745C>T p.T582M Chr.17: 4632019-4632019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376763252 Chr.17: 4632101-4632101 on Build GRCh38
rs748459113 Chr.17: 4632276-4632276 on Build GRCh38
rs543478770 Chr.17: 4632013-4632013 on Build GRCh38
rs763008416 Chr.17: 4632195-4632195 on Build GRCh38
rs774250671 Chr.17: 4632299-4632299 on Build GRCh38
rs577136020 Chr.17: 4632053-4632053 on Build GRCh38
rs773348641 Chr.17: 4632187-4632187 on Build GRCh38
rs771034178 Chr.17: 4632290-4632290 on Build GRCh38
rs2856415 Chr.17: 4632102-4632102 on Build GRCh38
rs985307514 Chr.17: 4631962-4631962 on Build GRCh38
rs758371475 Chr.17: 4632325-4632325 on Build GRCh38
rs533297249 Chr.17: 4632173-4632173 on Build GRCh38
rs1006300026 Chr.17: 4632138-4632138 on Build GRCh38
rs370211925 Chr.17: 4631964-4631964 on Build GRCh38
rs373939688 Chr.17: 4631992-4631992 on Build GRCh38
rs774183330 Chr.17: 4632193-4632194 on Build GRCh38
rs778987195 Chr.17: 4632229-4632229 on Build GRCh38
rs979177620 Chr.17: 4632015-4632015 on Build GRCh38
rs372580352 Chr.17: 4631986-4631986 on Build GRCh38
rs11568129 Chr.17: 4632006-4632006 on Build GRCh38
rs377406445 Chr.17: 4632208-4632208 on Build GRCh38
rs534093111 Chr.17: 4632377-4632377 on Build GRCh38
rs35176831 Chr.17: 4632159-4632159 on Build GRCh38
rs775264698 Chr.17: 4632107-4632107 on Build GRCh38
rs201836513 Chr.17: 4632230-4632230 on Build GRCh38
rs372526745 Chr.17: 4632346-4632346 on Build GRCh38
rs139519764 Chr.17: 4632211-4632211 on Build GRCh38
rs201278134 Chr.17: 4631999-4631999 on Build GRCh38
rs149366613 Chr.17: 4631965-4631965 on Build GRCh38
rs374001013 Chr.17: 4632018-4632018 on Build GRCh38
rs371864202 Chr.17: 4632331-4632331 on Build GRCh38
rs2856416 Chr.17: 4632077-4632077 on Build GRCh38
rs771790003 Chr.17: 4632026-4632026 on Build GRCh38
rs746392710 Chr.17: 4632349-4632349 on Build GRCh38
rs778379310 Chr.17: 4632038-4632038 on Build GRCh38
rs767571474 Chr.17: 4632309-4632309 on Build GRCh38
rs1016936159 Chr.17: 4631981-4631981 on Build GRCh38
rs950225925 Chr.17: 4632353-4632353 on Build GRCh38
rs779638646 Chr.17: 4632328-4632328 on Build GRCh38
rs750350521 Chr.17: 4632320-4632320 on Build GRCh38
rs777173779 Chr.17: 4632040-4632040 on Build GRCh38
rs199808667 Chr.17: 4631994-4631994 on Build GRCh38
rs770801037 Chr.17: 4631997-4632003 on Build GRCh38
rs34210653 Chr.17: 4632019-4632019 on Build GRCh38
rs200102401 Chr.17: 4632100-4632100 on Build GRCh38
rs770345235 Chr.17: 4632280-4632280 on Build GRCh38
rs2619112 Chr.17: 4632090-4632090 on Build GRCh38
rs753065107 Chr.17: 4631982-4631982 on Build GRCh38
rs139746240 Chr.17: 4632048-4632048 on Build GRCh38
rs769033136 Chr.17: 4632160-4632160 on Build GRCh38
rs761685166 Chr.17: 4632312-4632312 on Build GRCh38
rs761093526 Chr.17: 4632034-4632034 on Build GRCh38
rs763061394 Chr.17: 4632288-4632288 on Build GRCh38
rs553967871 Chr.17: 4632390-4632390 on Build GRCh38
rs773687745 Chr.17: 4632281-4632281 on Build GRCh38
rs753730814 Chr.17: 4632311-4632311 on Build GRCh38
rs986753287 Chr.17: 4632225-4632225 on Build GRCh38
rs770448397 Chr.17: 4632014-4632014 on Build GRCh38
rs6502804 Chr.17: 4632118-4632118 on Build GRCh38
rs759820341 Chr.17: 4632302-4632302 on Build GRCh38
rs759553239 Chr.17: 4632205-4632205 on Build GRCh38
rs201784601 Chr.17: 4632170-4632170 on Build GRCh38
rs149679697 Chr.17: 4632258-4632258 on Build GRCh38
rs962046079 Chr.17: 4632197-4632197 on Build GRCh38
rs150552792 Chr.17: 4632190-4632190 on Build GRCh38
rs41414251 Chr.17: 4632164-4632164 on Build GRCh38
rs764593975 Chr.17: 4631984-4631984 on Build GRCh38
rs868055472 Chr.17: 4632233-4632233 on Build GRCh38
rs761098208 Chr.17: 4631983-4631983 on Build GRCh38
rs766911588 Chr.17: 4632212-4632213 on Build GRCh38
rs917942648 Chr.17: 4632201-4632201 on Build GRCh38
rs2515886 Chr.17: 4632075-4632075 on Build GRCh38
rs751774986 Chr.17: 4632204-4632204 on Build GRCh38
rs564865192 Chr.17: 4632210-4632210 on Build GRCh38
rs568551669 Chr.17: 4632376-4632376 on Build GRCh38
rs768026830 Chr.17: 4631972-4631972 on Build GRCh38
rs1041613338 Chr.17: 4632049-4632049 on Build GRCh38
rs979084359 Chr.17: 4632248-4632248 on Build GRCh38
rs147458306 Chr.17: 4632003-4632003 on Build GRCh38
rs138264088 Chr.17: 4632092-4632092 on Build GRCh38
rs199690416 Chr.17: 4631985-4631988 on Build GRCh38
rs928425664 Chr.17: 4632384-4632384 on Build GRCh38
rs11568128 Chr.17: 4632108-4632108 on Build GRCh38
rs56310657 Chr.17: 4632244-4632244 on Build GRCh38
rs918185169 Chr.17: 4631990-4631990 on Build GRCh38
rs775998644 Chr.17: 4632027-4632027 on Build GRCh38
rs972486094 Chr.17: 4632200-4632200 on Build GRCh38
rs552591899 Chr.17: 4632202-4632202 on Build GRCh38
rs1045991344 Chr.17: 4632367-4632367 on Build GRCh38
rs757263469 Chr.17: 4632228-4632228 on Build GRCh38
rs765311000 Chr.17: 4632319-4632319 on Build GRCh38
rs747873506 Chr.17: 4632008-4632008 on Build GRCh38
rs749282547 Chr.17: 4632098-4632098 on Build GRCh38
rs577106801 Chr.17: 4632408-4632408 on Build GRCh38
rs750452049 Chr.17: 4632051-4632051 on Build GRCh38
rs1055954918 Chr.17: 4632078-4632078 on Build GRCh38
rs779612806 Chr.17: 4632257-4632257 on Build GRCh38
rs887351772 Chr.17: 4632374-4632374 on Build GRCh38
rs909215074 Chr.17: 4631963-4631963 on Build GRCh38
rs41398648 Chr.17: 4632169-4632169 on Build GRCh38
rs1017010920 Chr.17: 4632137-4632137 on Build GRCh38
rs761838242 Chr.17: 4632045-4632045 on Build GRCh38
rs41391948 Chr.17: 4632099-4632099 on Build GRCh38
rs754662144 Chr.17: 4632332-4632332 on Build GRCh38
rs888922355 Chr.17: 4632378-4632378 on Build GRCh38
rs142629035 Chr.17: 4632058-4632058 on Build GRCh38
rs138342123 Chr.17: 4632004-4632004 on Build GRCh38
rs777753372 Chr.17: 4632009-4632009 on Build GRCh38

View Full Product Details