Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4717578-4718068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRB2
Entrez Gene ID
409
Gene Name
arrestin beta 2
Gene Aliases
ARB2, ARR2, BARR2
UniGene
Hs.435811
Gene Chromosome Location
Chr.17: 4710494-4721500 on Build GRCh38
Transcripts
NM_001257328.1 NM_001257329.1 NM_001257330.1 NM_001257331.1 NM_004313.3 NM_199004.1 NR_047516.1 XM_006721520.1 XM_011523858.2 XM_017024645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7356491 c.556G>A p.E186K Chr.17: 4717958-4717958 on Build GRCh38
5594223 c.461C>T p.S154L Chr.17: 4717728-4717728 on Build GRCh38
243064 c.607T>G p.S203A Chr.17: 4718009-4718009 on Build GRCh38
7316047 c.450C>T p.F150F Chr.17: 4717717-4717717 on Build GRCh38
5554630 c.425G>A p.G142D Chr.17: 4717692-4717692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761032273 Chr.17: 4717693-4717693 on Build GRCh38
rs572320777 Chr.17: 4717858-4717858 on Build GRCh38
rs1010440696 Chr.17: 4717744-4717744 on Build GRCh38
rs267604947 Chr.17: 4717944-4717944 on Build GRCh38
rs757893765 Chr.17: 4717791-4717791 on Build GRCh38
rs746460867 Chr.17: 4718047-4718047 on Build GRCh38
rs143327202 Chr.17: 4717643-4717643 on Build GRCh38
rs532538037 Chr.17: 4717967-4717967 on Build GRCh38
rs763090389 Chr.17: 4717997-4717997 on Build GRCh38
rs200283276 Chr.17: 4717694-4717694 on Build GRCh38
rs771693958 Chr.17: 4717958-4717958 on Build GRCh38
rs773698196 Chr.17: 4717992-4717992 on Build GRCh38
rs1036719289 Chr.17: 4717890-4717890 on Build GRCh38
rs376881992 Chr.17: 4717999-4717999 on Build GRCh38
rs765279006 Chr.17: 4717711-4717711 on Build GRCh38
rs1049979407 Chr.17: 4717817-4717817 on Build GRCh38
rs1051915298 Chr.17: 4717684-4717684 on Build GRCh38
rs35741745 Chr.17: 4717602-4717602 on Build GRCh38
rs373701235 Chr.17: 4717887-4717887 on Build GRCh38
rs553305263 Chr.17: 4717653-4717653 on Build GRCh38
rs199982675 Chr.17: 4717918-4717918 on Build GRCh38
rs542009374 Chr.17: 4717826-4717826 on Build GRCh38
rs750382915 Chr.17: 4718038-4718038 on Build GRCh38
rs531914581 Chr.17: 4717620-4717620 on Build GRCh38
rs267604946 Chr.17: 4717943-4717943 on Build GRCh38
rs767592053 Chr.17: 4718003-4718003 on Build GRCh38
rs531803906 Chr.17: 4718002-4718002 on Build GRCh38
rs556200186 Chr.17: 4717788-4717788 on Build GRCh38
rs1045523592 Chr.17: 4717949-4717949 on Build GRCh38
rs762432944 Chr.17: 4717652-4717652 on Build GRCh38
rs373133862 Chr.17: 4717968-4717968 on Build GRCh38
rs750455960 Chr.17: 4717730-4717730 on Build GRCh38
rs755598221 Chr.17: 4717780-4717780 on Build GRCh38
rs34438956 Chr.17: 4717685-4717685 on Build GRCh38
rs570417917 Chr.17: 4717681-4717681 on Build GRCh38
rs548347966 Chr.17: 4718033-4718033 on Build GRCh38
rs375238507 Chr.17: 4717746-4717746 on Build GRCh38
rs751862122 Chr.17: 4717778-4717778 on Build GRCh38
rs796803344 Chr.17: 4717650-4717650 on Build GRCh38
rs757321101 Chr.17: 4717710-4717710 on Build GRCh38
rs769626763 Chr.17: 4717649-4717649 on Build GRCh38
rs540637696 Chr.17: 4717866-4717866 on Build GRCh38
rs138077887 Chr.17: 4717898-4717898 on Build GRCh38
rs113104031 Chr.17: 4717800-4717800 on Build GRCh38
rs770302219 Chr.17: 4717979-4717979 on Build GRCh38
rs757288423 Chr.17: 4717939-4717939 on Build GRCh38
rs370716675 Chr.17: 4717651-4717651 on Build GRCh38
rs1033006166 Chr.17: 4717601-4717601 on Build GRCh38
rs555720035 Chr.17: 4717837-4717837 on Build GRCh38
rs369737636 Chr.17: 4717801-4717801 on Build GRCh38
rs990462579 Chr.17: 4717647-4717664 on Build GRCh38
rs753693235 Chr.17: 4718030-4718030 on Build GRCh38
rs777598057 Chr.17: 4717929-4717929 on Build GRCh38
rs768403944 Chr.17: 4717597-4717597 on Build GRCh38
rs146732997 Chr.17: 4717978-4717978 on Build GRCh38
rs758418471 Chr.17: 4717911-4717911 on Build GRCh38
rs371532220 Chr.17: 4718031-4718031 on Build GRCh38
rs768146300 Chr.17: 4717975-4717975 on Build GRCh38
rs750508814 Chr.17: 4717910-4717910 on Build GRCh38
rs754356239 Chr.17: 4717702-4717702 on Build GRCh38
rs774184306 Chr.17: 4717656-4717656 on Build GRCh38
rs755897043 Chr.17: 4717928-4717928 on Build GRCh38
rs890306747 Chr.17: 4718046-4718046 on Build GRCh38
rs765694454 Chr.17: 4717904-4717904 on Build GRCh38
rs775148853 Chr.17: 4717873-4717873 on Build GRCh38
rs28365158 Chr.17: 4717633-4717633 on Build GRCh38
rs918115994 Chr.17: 4717880-4717880 on Build GRCh38
rs376922116 Chr.17: 4717899-4717899 on Build GRCh38
rs1019927955 Chr.17: 4717777-4717777 on Build GRCh38
rs774018655 Chr.17: 4717857-4717857 on Build GRCh38
rs761002315 Chr.17: 4717883-4717883 on Build GRCh38
rs751805903 Chr.17: 4717924-4717924 on Build GRCh38
rs765225491 Chr.17: 4717678-4717678 on Build GRCh38
rs749920277 Chr.17: 4717766-4717766 on Build GRCh38
rs371066182 Chr.17: 4717930-4717930 on Build GRCh38
rs191254209 Chr.17: 4717627-4717627 on Build GRCh38
rs552634178 Chr.17: 4717600-4717600 on Build GRCh38
rs780185793 Chr.17: 4717790-4717790 on Build GRCh38
rs777086124 Chr.17: 4717976-4717976 on Build GRCh38
rs770610622 Chr.17: 4717838-4717838 on Build GRCh38
rs768104540 Chr.17: 4717783-4717783 on Build GRCh38
rs367941912 Chr.17: 4717793-4717793 on Build GRCh38
rs111935802 Chr.17: 4717957-4717957 on Build GRCh38
rs759239609 Chr.17: 4717660-4717660 on Build GRCh38
rs36056585 Chr.17: 4718045-4718045 on Build GRCh38
rs375027517 Chr.17: 4717767-4717767 on Build GRCh38
rs779250850 Chr.17: 4717648-4717648 on Build GRCh38
rs775746698 Chr.17: 4717682-4717682 on Build GRCh38

View Full Product Details