Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4948941-4949454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO3
Entrez Gene ID
2027
Gene Name
enolase 3
Gene Aliases
GSD13, MSE
UniGene
Hs.224171
Gene Chromosome Location
Chr.17: 4949182-4957131 on Build GRCh38
Transcripts
XM_011523729.1 NM_005022.3 XM_017024761.1

Gene Symbol
PFN1
Entrez Gene ID
5216
Gene Name
profilin 1
Gene Aliases
ALS18
UniGene
Hs.494691
Gene Chromosome Location
Chr.17: 4945650-4949088 on Build GRCh38
Transcripts
XM_011523729.1 NM_005022.3 XM_017024761.1

SNP Detail

SNP ID Chromosome Location
rs569004633 Chr.17: 4949251-4949251 on Build GRCh38
rs1019990881 Chr.17: 4949162-4949162 on Build GRCh38
rs1029186044 Chr.17: 4949171-4949171 on Build GRCh38
rs909275506 Chr.17: 4948963-4948963 on Build GRCh38
rs1016507758 Chr.17: 4949427-4949427 on Build GRCh38
rs940809733 Chr.17: 4948964-4948964 on Build GRCh38
rs1008785959 Chr.17: 4948970-4948970 on Build GRCh38
rs901635724 Chr.17: 4949163-4949163 on Build GRCh38
rs964779531 Chr.17: 4949149-4949149 on Build GRCh38
rs1020378136 Chr.17: 4949062-4949062 on Build GRCh38
rs949856178 Chr.17: 4949138-4949138 on Build GRCh38
rs1036837468 Chr.17: 4948967-4948967 on Build GRCh38
rs1051003283 Chr.17: 4949409-4949409 on Build GRCh38
rs1012000052 Chr.17: 4949081-4949082 on Build GRCh38
rs1047676274 Chr.17: 4949344-4949344 on Build GRCh38
rs927159761 Chr.17: 4949289-4949289 on Build GRCh38
rs974405083 Chr.17: 4949026-4949026 on Build GRCh38
rs1001593862 Chr.17: 4949021-4949021 on Build GRCh38
rs945709789 Chr.17: 4949355-4949355 on Build GRCh38
rs527589018 Chr.17: 4949078-4949078 on Build GRCh38
rs2283568 Chr.17: 4949010-4949010 on Build GRCh38
rs901583435 Chr.17: 4949385-4949385 on Build GRCh38
rs984844132 Chr.17: 4948959-4948959 on Build GRCh38
rs954339964 Chr.17: 4949164-4949164 on Build GRCh38
rs113707551 Chr.17: 4948991-4948991 on Build GRCh38
rs945522362 Chr.17: 4949046-4949046 on Build GRCh38
rs200429340 Chr.17: 4949165-4949166 on Build GRCh38
rs1051280699 Chr.17: 4949036-4949036 on Build GRCh38
rs889293458 Chr.17: 4949417-4949417 on Build GRCh38
rs917991960 Chr.17: 4949241-4949241 on Build GRCh38
rs984580954 Chr.17: 4948984-4948984 on Build GRCh38
rs4790714 Chr.17: 4949168-4949168 on Build GRCh38
rs538088963 Chr.17: 4949328-4949328 on Build GRCh38
rs934199509 Chr.17: 4949031-4949032 on Build GRCh38
rs562443624 Chr.17: 4949029-4949029 on Build GRCh38
rs953735186 Chr.17: 4949183-4949183 on Build GRCh38
rs938068124 Chr.17: 4949260-4949260 on Build GRCh38
rs949908519 Chr.17: 4948981-4948981 on Build GRCh38
rs1006058019 Chr.17: 4949381-4949381 on Build GRCh38
rs1045673243 Chr.17: 4948983-4948983 on Build GRCh38
rs995947864 Chr.17: 4949431-4949431 on Build GRCh38
rs914531673 Chr.17: 4949350-4949350 on Build GRCh38
rs1026632316 Chr.17: 4949109-4949109 on Build GRCh38
rs34447441 Chr.17: 4949351-4949352 on Build GRCh38
rs972145507 Chr.17: 4949116-4949116 on Build GRCh38
rs892852612 Chr.17: 4949049-4949049 on Build GRCh38
rs962035125 Chr.17: 4949214-4949214 on Build GRCh38
rs1009957060 Chr.17: 4949054-4949054 on Build GRCh38
rs1045619103 Chr.17: 4949146-4949146 on Build GRCh38
rs997238704 Chr.17: 4949397-4949397 on Build GRCh38
rs552278267 Chr.17: 4949243-4949243 on Build GRCh38
rs966186689 Chr.17: 4949065-4949065 on Build GRCh38
rs1041394255 Chr.17: 4949358-4949358 on Build GRCh38
rs1002162944 Chr.17: 4949095-4949095 on Build GRCh38
rs896515602 Chr.17: 4949304-4949304 on Build GRCh38
rs925397511 Chr.17: 4949221-4949221 on Build GRCh38
rs892813881 Chr.17: 4949151-4949151 on Build GRCh38
rs981655389 Chr.17: 4949253-4949253 on Build GRCh38
rs972093048 Chr.17: 4949215-4949215 on Build GRCh38
rs953683830 Chr.17: 4949104-4949104 on Build GRCh38
rs937290339 Chr.17: 4949148-4949148 on Build GRCh38
rs1054502036 Chr.17: 4949150-4949150 on Build GRCh38
rs976262860 Chr.17: 4949094-4949094 on Build GRCh38
rs896420794 Chr.17: 4949099-4949099 on Build GRCh38
rs979968096 Chr.17: 4949206-4949206 on Build GRCh38
rs896998015 Chr.17: 4948972-4948972 on Build GRCh38
rs940720964 Chr.17: 4949114-4949114 on Build GRCh38
rs950609774 Chr.17: 4949137-4949137 on Build GRCh38
rs905754425 Chr.17: 4949147-4949147 on Build GRCh38
rs1026076570 Chr.17: 4948971-4948971 on Build GRCh38
rs918044014 Chr.17: 4949135-4949135 on Build GRCh38
rs1006839530 Chr.17: 4949423-4949423 on Build GRCh38
rs985178325 Chr.17: 4949106-4949106 on Build GRCh38
rs547348951 Chr.17: 4949098-4949098 on Build GRCh38
rs1016639852 Chr.17: 4949208-4949208 on Build GRCh38
rs914130921 Chr.17: 4949200-4949204 on Build GRCh38
rs531584914 Chr.17: 4949007-4949007 on Build GRCh38
rs909233309 Chr.17: 4949113-4949113 on Build GRCh38
rs950538654 Chr.17: 4948973-4948973 on Build GRCh38
rs561826954 Chr.17: 4949042-4949042 on Build GRCh38
rs1054383648 Chr.17: 4949341-4949341 on Build GRCh38
rs566876439 Chr.17: 4949153-4949153 on Build GRCh38
rs1006481198 Chr.17: 4949204-4949204 on Build GRCh38

View Full Product Details