Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 5442325-5442827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX33
Entrez Gene ID
56919
Gene Name
DEAH-box helicase 33
Gene Aliases
DDX33
UniGene
Hs.250456
Gene Chromosome Location
Chr.17: 5440912-5469060 on Build GRCh38
Transcripts
NM_001199699.1 NM_020162.3 XM_017024877.1

SNP Detail

SNP ID Chromosome Location
rs938543612 Chr.17: 5442547-5442547 on Build GRCh38
rs189008956 Chr.17: 5442552-5442552 on Build GRCh38
rs1034389668 Chr.17: 5442737-5442737 on Build GRCh38
rs577728480 Chr.17: 5442701-5442701 on Build GRCh38
rs557859028 Chr.17: 5442556-5442556 on Build GRCh38
rs994388501 Chr.17: 5442603-5442603 on Build GRCh38
rs1031655729 Chr.17: 5442795-5442795 on Build GRCh38
rs1022136161 Chr.17: 5442464-5442464 on Build GRCh38
rs958698069 Chr.17: 5442404-5442404 on Build GRCh38
rs748652868 Chr.17: 5442540-5442540 on Build GRCh38
rs185048114 Chr.17: 5442550-5442550 on Build GRCh38
rs397963188 Chr.17: 5442373-5442374 on Build GRCh38
rs976819484 Chr.17: 5442780-5442780 on Build GRCh38
rs894572312 Chr.17: 5442776-5442776 on Build GRCh38
rs930256169 Chr.17: 5442697-5442697 on Build GRCh38
rs942309276 Chr.17: 5442588-5442588 on Build GRCh38
rs898725242 Chr.17: 5442667-5442667 on Build GRCh38
rs58645097 Chr.17: 5442368-5442369 on Build GRCh38
rs990145372 Chr.17: 5442429-5442429 on Build GRCh38
rs543403796 Chr.17: 5442710-5442710 on Build GRCh38
rs555729354 Chr.17: 5442449-5442449 on Build GRCh38
rs1047791253 Chr.17: 5442699-5442699 on Build GRCh38
rs978306301 Chr.17: 5442493-5442493 on Build GRCh38
rs907349342 Chr.17: 5442706-5442706 on Build GRCh38
rs770187858 Chr.17: 5442604-5442604 on Build GRCh38
rs1034873068 Chr.17: 5442401-5442401 on Build GRCh38
rs201794439 Chr.17: 5442374-5442377 on Build GRCh38
rs764830627 Chr.17: 5442487-5442487 on Build GRCh38
rs3088255 Chr.17: 5442368-5442368 on Build GRCh38
rs933848121 Chr.17: 5442521-5442521 on Build GRCh38
rs989690162 Chr.17: 5442430-5442430 on Build GRCh38
rs1003492768 Chr.17: 5442717-5442717 on Build GRCh38
rs967941702 Chr.17: 5442472-5442472 on Build GRCh38
rs879641388 Chr.17: 5442364-5442365 on Build GRCh38
rs923811312 Chr.17: 5442500-5442500 on Build GRCh38
rs865930449 Chr.17: 5442693-5442693 on Build GRCh38
rs1018197127 Chr.17: 5442744-5442744 on Build GRCh38
rs375808058 Chr.17: 5442367-5442373 on Build GRCh38
rs375402610 Chr.17: 5442600-5442600 on Build GRCh38
rs868577456 Chr.17: 5442460-5442460 on Build GRCh38
rs1044059407 Chr.17: 5442518-5442518 on Build GRCh38

View Full Product Details