Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 5442994-5443527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX33
Entrez Gene ID
56919
Gene Name
DEAH-box helicase 33
Gene Aliases
DDX33
UniGene
Hs.250456
Gene Chromosome Location
Chr.17: 5440912-5469060 on Build GRCh38
Transcripts
NM_001199699.1 NM_020162.3 XM_017024877.1

SNP Detail

SNP ID Chromosome Location
rs916069461 Chr.17: 5443385-5443385 on Build GRCh38
rs773813697 Chr.17: 5443069-5443069 on Build GRCh38
rs1018226974 Chr.17: 5443462-5443462 on Build GRCh38
rs564204318 Chr.17: 5443145-5443145 on Build GRCh38
rs995375381 Chr.17: 5443489-5443489 on Build GRCh38
rs901291983 Chr.17: 5443060-5443065 on Build GRCh38
rs77195858 Chr.17: 5443126-5443126 on Build GRCh38
rs1011158889 Chr.17: 5443138-5443138 on Build GRCh38
rs557896141 Chr.17: 5443306-5443306 on Build GRCh38
rs892472652 Chr.17: 5443364-5443364 on Build GRCh38
rs894542863 Chr.17: 5443049-5443049 on Build GRCh38
rs1011357264 Chr.17: 5443381-5443381 on Build GRCh38
rs3744711 Chr.17: 5443279-5443279 on Build GRCh38
rs868529429 Chr.17: 5443120-5443120 on Build GRCh38
rs1052826221 Chr.17: 5443350-5443350 on Build GRCh38
rs909104918 Chr.17: 5443129-5443129 on Build GRCh38
rs891013933 Chr.17: 5443448-5443448 on Build GRCh38
rs999294614 Chr.17: 5443128-5443128 on Build GRCh38
rs577767535 Chr.17: 5443329-5443329 on Build GRCh38
rs371080373 Chr.17: 5443356-5443356 on Build GRCh38
rs535185013 Chr.17: 5443283-5443283 on Build GRCh38
rs770618079 Chr.17: 5443041-5443041 on Build GRCh38
rs1052215507 Chr.17: 5443435-5443435 on Build GRCh38
rs373321995 Chr.17: 5443399-5443399 on Build GRCh38
rs570027757 Chr.17: 5443190-5443190 on Build GRCh38
rs549636126 Chr.17: 5443231-5443231 on Build GRCh38
rs771702167 Chr.17: 5443215-5443215 on Build GRCh38
rs992873861 Chr.17: 5443233-5443233 on Build GRCh38
rs947582219 Chr.17: 5443407-5443407 on Build GRCh38
rs903272524 Chr.17: 5443414-5443414 on Build GRCh38
rs951073330 Chr.17: 5443305-5443305 on Build GRCh38
rs748596480 Chr.17: 5443022-5443024 on Build GRCh38
rs184594316 Chr.17: 5443177-5443177 on Build GRCh38
rs998825768 Chr.17: 5443421-5443421 on Build GRCh38
rs535812107 Chr.17: 5443216-5443216 on Build GRCh38
rs1011632446 Chr.17: 5443086-5443086 on Build GRCh38
rs973798974 Chr.17: 5443263-5443263 on Build GRCh38
rs180850458 Chr.17: 5443124-5443124 on Build GRCh38
rs372857509 Chr.17: 5443034-5443037 on Build GRCh38
rs867176540 Chr.17: 5443262-5443262 on Build GRCh38
rs1008135150 Chr.17: 5443458-5443458 on Build GRCh38
rs1043121817 Chr.17: 5443408-5443408 on Build GRCh38
rs963902468 Chr.17: 5443260-5443260 on Build GRCh38
rs1048292439 Chr.17: 5443264-5443264 on Build GRCh38
rs998300727 Chr.17: 5443333-5443333 on Build GRCh38
rs955472183 Chr.17: 5443174-5443174 on Build GRCh38
rs201214796 Chr.17: 5443031-5443034 on Build GRCh38
rs1031016537 Chr.17: 5443122-5443122 on Build GRCh38
rs940392466 Chr.17: 5443133-5443133 on Build GRCh38
rs928864306 Chr.17: 5443320-5443320 on Build GRCh38
rs189380471 Chr.17: 5443484-5443484 on Build GRCh38
rs918275593 Chr.17: 5443255-5443255 on Build GRCh38
rs138715319 Chr.17: 5443156-5443156 on Build GRCh38
rs998582855 Chr.17: 5443068-5443069 on Build GRCh38
rs879781810 Chr.17: 5443128-5443129 on Build GRCh38
rs992599314 Chr.17: 5443486-5443486 on Build GRCh38
rs1018133610 Chr.17: 5443210-5443210 on Build GRCh38

View Full Product Details