Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 5443317-5443836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX33
Entrez Gene ID
56919
Gene Name
DEAH-box helicase 33
Gene Aliases
DDX33
UniGene
Hs.250456
Gene Chromosome Location
Chr.17: 5440912-5469060 on Build GRCh38
Transcripts
NM_001199699.1 NM_020162.3 XM_017024877.1

SNP Detail

SNP ID Chromosome Location
rs916069461 Chr.17: 5443385-5443385 on Build GRCh38
rs1018226974 Chr.17: 5443462-5443462 on Build GRCh38
rs572971099 Chr.17: 5443672-5443672 on Build GRCh38
rs780319626 Chr.17: 5443688-5443688 on Build GRCh38
rs995375381 Chr.17: 5443489-5443489 on Build GRCh38
rs1025373740 Chr.17: 5443780-5443780 on Build GRCh38
rs557038402 Chr.17: 5443592-5443592 on Build GRCh38
rs896645678 Chr.17: 5443769-5443769 on Build GRCh38
rs1039633764 Chr.17: 5443733-5443733 on Build GRCh38
rs1035470399 Chr.17: 5443805-5443805 on Build GRCh38
rs892472652 Chr.17: 5443364-5443364 on Build GRCh38
rs1011357264 Chr.17: 5443381-5443381 on Build GRCh38
rs959698551 Chr.17: 5443806-5443806 on Build GRCh38
rs942636626 Chr.17: 5443550-5443550 on Build GRCh38
rs1052826221 Chr.17: 5443350-5443350 on Build GRCh38
rs891013933 Chr.17: 5443448-5443448 on Build GRCh38
rs2074437 Chr.17: 5443764-5443764 on Build GRCh38
rs371080373 Chr.17: 5443356-5443356 on Build GRCh38
rs1052215507 Chr.17: 5443435-5443435 on Build GRCh38
rs373321995 Chr.17: 5443399-5443399 on Build GRCh38
rs143514102 Chr.17: 5443669-5443669 on Build GRCh38
rs1043778123 Chr.17: 5443698-5443698 on Build GRCh38
rs760666920 Chr.17: 5443523-5443523 on Build GRCh38
rs982446098 Chr.17: 5443524-5443524 on Build GRCh38
rs951076907 Chr.17: 5443529-5443530 on Build GRCh38
rs947582219 Chr.17: 5443407-5443407 on Build GRCh38
rs903272524 Chr.17: 5443414-5443414 on Build GRCh38
rs796203643 Chr.17: 5443541-5443541 on Build GRCh38
rs928305928 Chr.17: 5443535-5443535 on Build GRCh38
rs998825768 Chr.17: 5443421-5443421 on Build GRCh38
rs1052521090 Chr.17: 5443639-5443639 on Build GRCh38
rs764074359 Chr.17: 5443561-5443561 on Build GRCh38
rs1004359293 Chr.17: 5443794-5443794 on Build GRCh38
rs1008135150 Chr.17: 5443458-5443458 on Build GRCh38
rs1043121817 Chr.17: 5443408-5443408 on Build GRCh38
rs901628302 Chr.17: 5443570-5443570 on Build GRCh38
rs753717925 Chr.17: 5443562-5443562 on Build GRCh38
rs994112875 Chr.17: 5443778-5443778 on Build GRCh38
rs542581720 Chr.17: 5443646-5443646 on Build GRCh38
rs1026702125 Chr.17: 5443509-5443509 on Build GRCh38
rs117100367 Chr.17: 5443593-5443593 on Build GRCh38
rs147977554 Chr.17: 5443756-5443756 on Build GRCh38
rs947522597 Chr.17: 5443584-5443584 on Build GRCh38
rs189380471 Chr.17: 5443484-5443484 on Build GRCh38
rs756640406 Chr.17: 5443783-5443783 on Build GRCh38
rs751720196 Chr.17: 5443616-5443616 on Build GRCh38
rs753255141 Chr.17: 5443765-5443765 on Build GRCh38
rs983776147 Chr.17: 5443536-5443536 on Build GRCh38
rs992599314 Chr.17: 5443486-5443486 on Build GRCh38
rs1051884425 Chr.17: 5443660-5443660 on Build GRCh38
rs778182453 Chr.17: 5443798-5443798 on Build GRCh38

View Full Product Details