Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6652417-6652922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf100
Entrez Gene ID
388327
Gene Name
chromosome 17 open reading frame 100
Gene Aliases
-
UniGene
Hs.262480
Gene Chromosome Location
Chr.17: 6651739-6653298 on Build GRCh38
Transcripts
NM_001105520.1

SNP Detail

SNP ID Chromosome Location
rs976632882 Chr.17: 6652444-6652444 on Build GRCh38
rs1018015498 Chr.17: 6652762-6652762 on Build GRCh38
rs144808411 Chr.17: 6652875-6652876 on Build GRCh38
rs893998315 Chr.17: 6652486-6652486 on Build GRCh38
rs954097815 Chr.17: 6652665-6652665 on Build GRCh38
rs914563204 Chr.17: 6652846-6652846 on Build GRCh38
rs768398868 Chr.17: 6652519-6652521 on Build GRCh38
rs998104772 Chr.17: 6652593-6652593 on Build GRCh38
rs370883861 Chr.17: 6652577-6652577 on Build GRCh38
rs989946946 Chr.17: 6652795-6652795 on Build GRCh38
rs7226265 Chr.17: 6652703-6652703 on Build GRCh38
rs59663818 Chr.17: 6652879-6652880 on Build GRCh38
rs775566097 Chr.17: 6652613-6652613 on Build GRCh38
rs1029624716 Chr.17: 6652595-6652595 on Build GRCh38
rs79612349 Chr.17: 6652763-6652763 on Build GRCh38
rs1017158677 Chr.17: 6652679-6652679 on Build GRCh38
rs537762111 Chr.17: 6652599-6652599 on Build GRCh38
rs150414123 Chr.17: 6652876-6652877 on Build GRCh38
rs947408990 Chr.17: 6652881-6652881 on Build GRCh38
rs539248536 Chr.17: 6652868-6652868 on Build GRCh38
rs61741085 Chr.17: 6652677-6652677 on Build GRCh38
rs568982588 Chr.17: 6652500-6652500 on Build GRCh38
rs531196897 Chr.17: 6652543-6652543 on Build GRCh38
rs567908690 Chr.17: 6652683-6652683 on Build GRCh38
rs956622796 Chr.17: 6652798-6652798 on Build GRCh38
rs926964603 Chr.17: 6652752-6652752 on Build GRCh38
rs897367120 Chr.17: 6652698-6652698 on Build GRCh38
rs368229510 Chr.17: 6652573-6652573 on Build GRCh38
rs988806940 Chr.17: 6652597-6652597 on Build GRCh38
rs989833694 Chr.17: 6652838-6652838 on Build GRCh38
rs937088419 Chr.17: 6652461-6652461 on Build GRCh38
rs570210182 Chr.17: 6652505-6652505 on Build GRCh38
rs915389879 Chr.17: 6652852-6652852 on Build GRCh38
rs796987373 Chr.17: 6652628-6652629 on Build GRCh38
rs1011564453 Chr.17: 6652488-6652488 on Build GRCh38
rs1055678021 Chr.17: 6652463-6652463 on Build GRCh38
rs902510641 Chr.17: 6652514-6652514 on Build GRCh38
rs973052901 Chr.17: 6652693-6652693 on Build GRCh38
rs935251692 Chr.17: 6652544-6652544 on Build GRCh38
rs1031253368 Chr.17: 6652789-6652789 on Build GRCh38
rs776252634 Chr.17: 6652737-6652737 on Build GRCh38
rs946946934 Chr.17: 6652887-6652887 on Build GRCh38
rs971312779 Chr.17: 6652730-6652730 on Build GRCh38
rs888139279 Chr.17: 6652734-6652734 on Build GRCh38
rs548215514 Chr.17: 6652632-6652632 on Build GRCh38
rs61741086 Chr.17: 6652688-6652688 on Build GRCh38
rs976519670 Chr.17: 6652773-6652773 on Build GRCh38
rs1026620507 Chr.17: 6652716-6652716 on Build GRCh38

View Full Product Details