Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7022965-7023457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL6B
Entrez Gene ID
255877
Gene Name
B-cell CLL/lymphoma 6B
Gene Aliases
BAZF, ZBTB28, ZNF62
UniGene
Hs.22575
Gene Chromosome Location
Chr.17: 7023050-7029642 on Build GRCh38
Transcripts
NM_181844.3

SNP Detail

SNP ID Chromosome Location
rs958185589 Chr.17: 7023374-7023375 on Build GRCh38
rs537283305 Chr.17: 7023032-7023032 on Build GRCh38
rs909908939 Chr.17: 7023132-7023132 on Build GRCh38
rs905735606 Chr.17: 7023428-7023428 on Build GRCh38
rs950027170 Chr.17: 7023414-7023414 on Build GRCh38
rs967002896 Chr.17: 7023280-7023280 on Build GRCh38
rs565831590 Chr.17: 7023384-7023384 on Build GRCh38
rs35902568 Chr.17: 7023154-7023155 on Build GRCh38
rs1045582427 Chr.17: 7023185-7023185 on Build GRCh38
rs564830614 Chr.17: 7023213-7023213 on Build GRCh38
rs1045955892 Chr.17: 7023420-7023420 on Build GRCh38
rs947430791 Chr.17: 7023377-7023377 on Build GRCh38
rs556843460 Chr.17: 7023052-7023052 on Build GRCh38
rs886582616 Chr.17: 7023252-7023252 on Build GRCh38
rs967247560 Chr.17: 7023061-7023061 on Build GRCh38
rs898927735 Chr.17: 7023290-7023290 on Build GRCh38
rs534615194 Chr.17: 7022986-7022986 on Build GRCh38
rs1009916275 Chr.17: 7023260-7023260 on Build GRCh38
rs868822103 Chr.17: 7023041-7023041 on Build GRCh38
rs1032707805 Chr.17: 7023015-7023015 on Build GRCh38
rs142025015 Chr.17: 7023006-7023006 on Build GRCh38
rs73979725 Chr.17: 7023317-7023317 on Build GRCh38
rs779154778 Chr.17: 7023314-7023314 on Build GRCh38
rs544676370 Chr.17: 7023210-7023210 on Build GRCh38
rs976948714 Chr.17: 7023080-7023080 on Build GRCh38
rs9889241 Chr.17: 7023396-7023396 on Build GRCh38
rs978597958 Chr.17: 7023338-7023338 on Build GRCh38
rs146311225 Chr.17: 7023429-7023429 on Build GRCh38
rs909877798 Chr.17: 7023345-7023345 on Build GRCh38
rs949688057 Chr.17: 7023218-7023218 on Build GRCh38
rs945875761 Chr.17: 7023108-7023108 on Build GRCh38
rs979736929 Chr.17: 7023008-7023008 on Build GRCh38
rs1037054732 Chr.17: 7023389-7023389 on Build GRCh38
rs754127097 Chr.17: 7023017-7023017 on Build GRCh38
rs1005027873 Chr.17: 7023263-7023263 on Build GRCh38
rs553011984 Chr.17: 7023128-7023128 on Build GRCh38
rs941330636 Chr.17: 7023370-7023370 on Build GRCh38
rs991212254 Chr.17: 7023376-7023376 on Build GRCh38
rs925348965 Chr.17: 7023340-7023340 on Build GRCh38
rs1013996402 Chr.17: 7022995-7022995 on Build GRCh38
rs529351617 Chr.17: 7023352-7023352 on Build GRCh38
rs996237324 Chr.17: 7023304-7023304 on Build GRCh38
rs187528429 Chr.17: 7023169-7023169 on Build GRCh38
rs879819206 Chr.17: 7023387-7023387 on Build GRCh38
rs1037389526 Chr.17: 7023171-7023171 on Build GRCh38
rs746405444 Chr.17: 7023058-7023058 on Build GRCh38
rs1014480965 Chr.17: 7023182-7023182 on Build GRCh38
rs150931981 Chr.17: 7023354-7023355 on Build GRCh38
rs954110638 Chr.17: 7023327-7023327 on Build GRCh38
rs558077873 Chr.17: 7022989-7022989 on Build GRCh38
rs1028681977 Chr.17: 7023308-7023308 on Build GRCh38
rs897073755 Chr.17: 7023177-7023177 on Build GRCh38
rs1020229731 Chr.17: 7023325-7023325 on Build GRCh38
rs985548546 Chr.17: 7023337-7023337 on Build GRCh38
rs957062082 Chr.17: 7023250-7023250 on Build GRCh38
rs949429339 Chr.17: 7023412-7023412 on Build GRCh38
rs932750138 Chr.17: 7023112-7023112 on Build GRCh38
rs533442653 Chr.17: 7023249-7023249 on Build GRCh38
rs573802230 Chr.17: 7023063-7023063 on Build GRCh38
rs958260197 Chr.17: 7023016-7023016 on Build GRCh38
rs542696420 Chr.17: 7023081-7023081 on Build GRCh38
rs544013526 Chr.17: 7023271-7023271 on Build GRCh38
rs936631854 Chr.17: 7023147-7023147 on Build GRCh38
rs549027864 Chr.17: 7023365-7023365 on Build GRCh38
rs1001311702 Chr.17: 7023242-7023242 on Build GRCh38
rs777124969 Chr.17: 7022997-7022997 on Build GRCh38
rs370839658 Chr.17: 7023012-7023012 on Build GRCh38

View Full Product Details