Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7026859-7027347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL6B
Entrez Gene ID
255877
Gene Name
B-cell CLL/lymphoma 6B
Gene Aliases
BAZF, ZBTB28, ZNF62
UniGene
Hs.22575
Gene Chromosome Location
Chr.17: 7023050-7029642 on Build GRCh38
Transcripts
NM_181844.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5420447 c.1198C>T p.R400W Chr.17: 7026962-7026962 on Build GRCh38
983720 c.1290C>T p.H430H Chr.17: 7027054-7027054 on Build GRCh38
6847646 c.1277C>T p.T426I Chr.17: 7027041-7027041 on Build GRCh38
6390053 c.1323C>A p.H441Q Chr.17: 7027087-7027087 on Build GRCh38
3691759 c.1218C>T p.H406H Chr.17: 7026982-7026982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776129332 Chr.17: 7027071-7027071 on Build GRCh38
rs750470292 Chr.17: 7027022-7027022 on Build GRCh38
rs748869804 Chr.17: 7027054-7027054 on Build GRCh38
rs766305029 Chr.17: 7027313-7027313 on Build GRCh38
rs553827023 Chr.17: 7027170-7027170 on Build GRCh38
rs541080613 Chr.17: 7027205-7027205 on Build GRCh38
rs1027716100 Chr.17: 7027259-7027259 on Build GRCh38
rs752543663 Chr.17: 7027032-7027032 on Build GRCh38
rs991976413 Chr.17: 7027066-7027066 on Build GRCh38
rs1043783343 Chr.17: 7026881-7026881 on Build GRCh38
rs778315859 Chr.17: 7027063-7027063 on Build GRCh38
rs772665232 Chr.17: 7026984-7026984 on Build GRCh38
rs761008657 Chr.17: 7026880-7026880 on Build GRCh38
rs200881376 Chr.17: 7026936-7026936 on Build GRCh38
rs1031880959 Chr.17: 7026938-7026938 on Build GRCh38
rs770313082 Chr.17: 7026970-7026970 on Build GRCh38
rs202012338 Chr.17: 7026988-7026988 on Build GRCh38
rs763286941 Chr.17: 7027139-7027139 on Build GRCh38
rs748891591 Chr.17: 7026967-7026967 on Build GRCh38
rs925024437 Chr.17: 7027106-7027106 on Build GRCh38
rs747901995 Chr.17: 7026950-7026950 on Build GRCh38
rs754856274 Chr.17: 7026939-7026939 on Build GRCh38
rs764346335 Chr.17: 7026919-7026919 on Build GRCh38
rs999603731 Chr.17: 7026933-7026933 on Build GRCh38
rs975118247 Chr.17: 7027309-7027309 on Build GRCh38
rs376936803 Chr.17: 7026946-7026946 on Build GRCh38
rs556723329 Chr.17: 7027042-7027042 on Build GRCh38
rs763004269 Chr.17: 7027132-7027132 on Build GRCh38
rs910012037 Chr.17: 7027297-7027297 on Build GRCh38
rs769111877 Chr.17: 7027087-7027087 on Build GRCh38
rs1040516885 Chr.17: 7027204-7027204 on Build GRCh38
rs35057803 Chr.17: 7027177-7027178 on Build GRCh38
rs548786963 Chr.17: 7026922-7026922 on Build GRCh38
rs769447393 Chr.17: 7026960-7026960 on Build GRCh38
rs892033523 Chr.17: 7027191-7027191 on Build GRCh38
rs753783380 Chr.17: 7026892-7026892 on Build GRCh38
rs761994879 Chr.17: 7027014-7027014 on Build GRCh38
rs951740360 Chr.17: 7027255-7027255 on Build GRCh38
rs761967216 Chr.17: 7027103-7027103 on Build GRCh38
rs571811101 Chr.17: 7027172-7027172 on Build GRCh38
rs752832864 Chr.17: 7027115-7027115 on Build GRCh38
rs371647213 Chr.17: 7027059-7027059 on Build GRCh38
rs186677055 Chr.17: 7027045-7027045 on Build GRCh38
rs1009127213 Chr.17: 7027198-7027198 on Build GRCh38
rs754002967 Chr.17: 7027011-7027011 on Build GRCh38
rs1043446742 Chr.17: 7027088-7027088 on Build GRCh38
rs200507334 Chr.17: 7026971-7026971 on Build GRCh38
rs952040528 Chr.17: 7027035-7027035 on Build GRCh38
rs903554678 Chr.17: 7026902-7026902 on Build GRCh38
rs373527238 Chr.17: 7027094-7027094 on Build GRCh38
rs765213795 Chr.17: 7027016-7027016 on Build GRCh38
rs117980493 Chr.17: 7027078-7027078 on Build GRCh38
rs764470868 Chr.17: 7026997-7026997 on Build GRCh38
rs754941923 Chr.17: 7027023-7027023 on Build GRCh38
rs1039485013 Chr.17: 7026943-7026943 on Build GRCh38
rs773173142 Chr.17: 7027125-7027125 on Build GRCh38
rs781047103 Chr.17: 7027024-7027024 on Build GRCh38
rs189158063 Chr.17: 7027163-7027163 on Build GRCh38
rs541639358 Chr.17: 7027162-7027162 on Build GRCh38
rs964746709 Chr.17: 7027015-7027015 on Build GRCh38
rs572482756 Chr.17: 7027284-7027284 on Build GRCh38
rs539445999 Chr.17: 7027069-7027069 on Build GRCh38
rs939280665 Chr.17: 7027061-7027061 on Build GRCh38
rs776000574 Chr.17: 7026985-7026985 on Build GRCh38
rs904595988 Chr.17: 7027137-7027137 on Build GRCh38
rs1004874011 Chr.17: 7027299-7027299 on Build GRCh38
rs367582163 Chr.17: 7027305-7027305 on Build GRCh38
rs761064240 Chr.17: 7026986-7026986 on Build GRCh38
rs996752473 Chr.17: 7027239-7027239 on Build GRCh38
rs777388875 Chr.17: 7026963-7026963 on Build GRCh38
rs201281948 Chr.17: 7027108-7027108 on Build GRCh38
rs752348909 Chr.17: 7027225-7027225 on Build GRCh38
rs568588238 Chr.17: 7026923-7026923 on Build GRCh38
rs900813422 Chr.17: 7026956-7026956 on Build GRCh38

View Full Product Details