Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7178203-7178696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASGR1
Entrez Gene ID
432
Gene Name
asialoglycoprotein receptor 1
Gene Aliases
ASGPR, ASGPR1, CLEC4H1, HL-1
UniGene
Hs.12056
Gene Chromosome Location
Chr.17: 7173431-7179564 on Build GRCh38
Transcripts
NM_001197216.2 NM_001671.4 XM_011523861.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6189885 c.61C>A p.L21I Chr.17: 7178503-7178503 on Build GRCh38
5055731 c.54_56delCCA p.H19delH Chr.17: 7178508-7178510 on Build GRCh38
5055732 c.54_56delCCA p.H19delH Chr.17: 7178508-7178510 on Build GRCh38
4880168 c.184C>A p.L62I Chr.17: 7178380-7178380 on Build GRCh38
3521546 c.42G>A p.E14E Chr.17: 7178522-7178522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745819327 Chr.17: 7178620-7178620 on Build GRCh38
rs749587351 Chr.17: 7178487-7178487 on Build GRCh38
rs138040706 Chr.17: 7178548-7178548 on Build GRCh38
rs759563985 Chr.17: 7178507-7178507 on Build GRCh38
rs779143556 Chr.17: 7178605-7178605 on Build GRCh38
rs770114856 Chr.17: 7178482-7178482 on Build GRCh38
rs561364325 Chr.17: 7178437-7178437 on Build GRCh38
rs748668624 Chr.17: 7178479-7178479 on Build GRCh38
rs749364552 Chr.17: 7178407-7178407 on Build GRCh38
rs781590979 Chr.17: 7178472-7178472 on Build GRCh38
rs749675436 Chr.17: 7178566-7178566 on Build GRCh38
rs779798527 Chr.17: 7178462-7178462 on Build GRCh38
rs112059971 Chr.17: 7178589-7178589 on Build GRCh38
rs925328672 Chr.17: 7178578-7178578 on Build GRCh38
rs77369802 Chr.17: 7178442-7178442 on Build GRCh38
rs979786294 Chr.17: 7178329-7178329 on Build GRCh38
rs201915673 Chr.17: 7178535-7178535 on Build GRCh38
rs969780460 Chr.17: 7178312-7178312 on Build GRCh38
rs1004606390 Chr.17: 7178319-7178319 on Build GRCh38
rs373175252 Chr.17: 7178528-7178528 on Build GRCh38
rs996134498 Chr.17: 7178372-7178372 on Build GRCh38
rs778107086 Chr.17: 7178484-7178484 on Build GRCh38
rs772157233 Chr.17: 7178511-7178511 on Build GRCh38
rs945575966 Chr.17: 7178539-7178539 on Build GRCh38
rs561987493 Chr.17: 7178576-7178576 on Build GRCh38
rs772631211 Chr.17: 7178581-7178581 on Build GRCh38
rs901283372 Chr.17: 7178559-7178559 on Build GRCh38
rs771154885 Chr.17: 7178489-7178489 on Build GRCh38
rs746291841 Chr.17: 7178525-7178527 on Build GRCh38
rs1015484180 Chr.17: 7178235-7178235 on Build GRCh38
rs765098282 Chr.17: 7178451-7178451 on Build GRCh38
rs763099208 Chr.17: 7178393-7178394 on Build GRCh38
rs917018141 Chr.17: 7178265-7178265 on Build GRCh38
rs888462825 Chr.17: 7178640-7178640 on Build GRCh38
rs932721042 Chr.17: 7178568-7178568 on Build GRCh38
rs752497233 Chr.17: 7178275-7178275 on Build GRCh38
rs537927114 Chr.17: 7178293-7178293 on Build GRCh38
rs150368459 Chr.17: 7178537-7178537 on Build GRCh38
rs200273995 Chr.17: 7178456-7178456 on Build GRCh38
rs554673961 Chr.17: 7178364-7178364 on Build GRCh38
rs761550199 Chr.17: 7178450-7178450 on Build GRCh38
rs199577129 Chr.17: 7178245-7178245 on Build GRCh38
rs754497570 Chr.17: 7178546-7178546 on Build GRCh38
rs757413548 Chr.17: 7178464-7178464 on Build GRCh38
rs138199073 Chr.17: 7178519-7178519 on Build GRCh38
rs141866734 Chr.17: 7178527-7178527 on Build GRCh38
rs377207552 Chr.17: 7178580-7178580 on Build GRCh38
rs762792513 Chr.17: 7178530-7178530 on Build GRCh38
rs145652363 Chr.17: 7178494-7178494 on Build GRCh38
rs754474579 Chr.17: 7178468-7178468 on Build GRCh38
rs761792778 Chr.17: 7178518-7178518 on Build GRCh38
rs1016350320 Chr.17: 7178341-7178342 on Build GRCh38
rs925626733 Chr.17: 7178347-7178347 on Build GRCh38
rs778009812 Chr.17: 7178249-7178249 on Build GRCh38
rs756648015 Chr.17: 7178554-7178554 on Build GRCh38
rs935623829 Chr.17: 7178438-7178438 on Build GRCh38
rs757577814 Chr.17: 7178570-7178570 on Build GRCh38
rs776479397 Chr.17: 7178516-7178516 on Build GRCh38
rs568084983 Chr.17: 7178374-7178374 on Build GRCh38
rs868300471 Chr.17: 7178412-7178412 on Build GRCh38
rs778194735 Chr.17: 7178564-7178564 on Build GRCh38
rs967206490 Chr.17: 7178542-7178542 on Build GRCh38
rs753765568 Chr.17: 7178449-7178449 on Build GRCh38
rs962999623 Chr.17: 7178345-7178345 on Build GRCh38
rs758132232 Chr.17: 7178460-7178460 on Build GRCh38

View Full Product Details