Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7179427-7179937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASGR1
Entrez Gene ID
432
Gene Name
asialoglycoprotein receptor 1
Gene Aliases
ASGPR, ASGPR1, CLEC4H1, HL-1
UniGene
Hs.12056
Gene Chromosome Location
Chr.17: 7173431-7179564 on Build GRCh38
Transcripts
NM_001197216.2 NM_001671.4

SNP Detail

SNP ID Chromosome Location
rs927223931 Chr.17: 7179853-7179853 on Build GRCh38
rs764213396 Chr.17: 7179501-7179501 on Build GRCh38
rs950057704 Chr.17: 7179527-7179527 on Build GRCh38
rs1029911513 Chr.17: 7179645-7179645 on Build GRCh38
rs1054223647 Chr.17: 7179608-7179608 on Build GRCh38
rs567180346 Chr.17: 7179712-7179712 on Build GRCh38
rs1050171822 Chr.17: 7179518-7179518 on Build GRCh38
rs997554252 Chr.17: 7179629-7179629 on Build GRCh38
rs953519873 Chr.17: 7179763-7179763 on Build GRCh38
rs1045581033 Chr.17: 7179894-7179894 on Build GRCh38
rs753028246 Chr.17: 7179874-7179874 on Build GRCh38
rs954279008 Chr.17: 7179670-7179670 on Build GRCh38
rs944377068 Chr.17: 7179558-7179558 on Build GRCh38
rs538546213 Chr.17: 7179608-7179608 on Build GRCh38
rs924236202 Chr.17: 7179842-7179842 on Build GRCh38
rs892915085 Chr.17: 7179590-7179590 on Build GRCh38
rs547073913 Chr.17: 7179675-7179675 on Build GRCh38
rs1037498928 Chr.17: 7179502-7179502 on Build GRCh38
rs1001403386 Chr.17: 7179561-7179561 on Build GRCh38
rs962585266 Chr.17: 7179482-7179482 on Build GRCh38
rs143478857 Chr.17: 7179880-7179880 on Build GRCh38
rs894272791 Chr.17: 7179609-7179609 on Build GRCh38
rs965749745 Chr.17: 7179626-7179626 on Build GRCh38
rs192207637 Chr.17: 7179739-7179739 on Build GRCh38
rs985908875 Chr.17: 7179775-7179775 on Build GRCh38
rs918627347 Chr.17: 7179811-7179811 on Build GRCh38
rs559169097 Chr.17: 7179751-7179751 on Build GRCh38
rs911630282 Chr.17: 7179787-7179787 on Build GRCh38
rs575453157 Chr.17: 7179781-7179781 on Build GRCh38
rs115109403 Chr.17: 7179489-7179489 on Build GRCh38
rs751712967 Chr.17: 7179726-7179726 on Build GRCh38
rs920728844 Chr.17: 7179497-7179497 on Build GRCh38
rs905776201 Chr.17: 7179557-7179557 on Build GRCh38
rs892984138 Chr.17: 7179884-7179884 on Build GRCh38
rs985592850 Chr.17: 7179485-7179485 on Build GRCh38
rs920804446 Chr.17: 7179741-7179741 on Build GRCh38
rs894346448 Chr.17: 7179881-7179881 on Build GRCh38
rs554650310 Chr.17: 7179851-7179851 on Build GRCh38
rs937224194 Chr.17: 7179868-7179868 on Build GRCh38
rs941433447 Chr.17: 7179492-7179492 on Build GRCh38
rs903116355 Chr.17: 7179586-7179586 on Build GRCh38
rs972848757 Chr.17: 7179771-7179771 on Build GRCh38
rs568123487 Chr.17: 7179478-7179478 on Build GRCh38
rs768061296 Chr.17: 7179839-7179839 on Build GRCh38
rs766586422 Chr.17: 7179459-7179459 on Build GRCh38
rs1012324583 Chr.17: 7179622-7179622 on Build GRCh38
rs950117864 Chr.17: 7179831-7179831 on Build GRCh38
rs138060620 Chr.17: 7179522-7179522 on Build GRCh38
rs867423296 Chr.17: 7179569-7179569 on Build GRCh38
rs1015311068 Chr.17: 7179729-7179729 on Build GRCh38
rs1020426418 Chr.17: 7179615-7179615 on Build GRCh38
rs963173343 Chr.17: 7179760-7179760 on Build GRCh38
rs182244108 Chr.17: 7179850-7179850 on Build GRCh38
rs973614146 Chr.17: 7179740-7179740 on Build GRCh38
rs550810733 Chr.17: 7179797-7179797 on Build GRCh38
rs1045680850 Chr.17: 7179539-7179539 on Build GRCh38

View Full Product Details