Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7190198-7190699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG4
Entrez Gene ID
1742
Gene Name
discs large MAGUK scaffold protein 4
Gene Aliases
PSD95, SAP-90, SAP90
UniGene
Hs.463928
Gene Chromosome Location
Chr.17: 7189890-7220050 on Build GRCh38
Transcripts
NM_001128827.2 NM_001321074.1 NM_001321075.1 NM_001321076.1 NM_001321077.1 NM_001365.4 NR_135527.1 XM_005256491.1 XM_011523698.1 XM_011523699.2 XM_011523702.1 XM_017024288.1 XM_017024289.1 XM_017024290.1

SNP Detail

SNP ID Chromosome Location
rs1020298629 Chr.17: 7190367-7190368 on Build GRCh38
rs1037508336 Chr.17: 7190258-7190258 on Build GRCh38
rs916273608 Chr.17: 7190440-7190440 on Build GRCh38
rs966952349 Chr.17: 7190388-7190388 on Build GRCh38
rs560944237 Chr.17: 7190619-7190619 on Build GRCh38
rs1049600151 Chr.17: 7190498-7190498 on Build GRCh38
rs376222775 Chr.17: 7190624-7190625 on Build GRCh38
rs999774359 Chr.17: 7190643-7190643 on Build GRCh38
rs370698277 Chr.17: 7190618-7190618 on Build GRCh38
rs746076481 Chr.17: 7190665-7190665 on Build GRCh38
rs896778470 Chr.17: 7190328-7190328 on Build GRCh38
rs564867795 Chr.17: 7190464-7190464 on Build GRCh38
rs896847846 Chr.17: 7190572-7190572 on Build GRCh38
rs906808498 Chr.17: 7190637-7190637 on Build GRCh38
rs374801889 Chr.17: 7190525-7190525 on Build GRCh38
rs1032509737 Chr.17: 7190405-7190405 on Build GRCh38
rs966730501 Chr.17: 7190484-7190484 on Build GRCh38
rs1002315264 Chr.17: 7190390-7190390 on Build GRCh38
rs181496750 Chr.17: 7190305-7190305 on Build GRCh38
rs958175209 Chr.17: 7190647-7190647 on Build GRCh38
rs766118390 Chr.17: 7190532-7190532 on Build GRCh38
rs1036748840 Chr.17: 7190556-7190556 on Build GRCh38
rs35747999 Chr.17: 7190246-7190247 on Build GRCh38
rs1005275825 Chr.17: 7190308-7190308 on Build GRCh38
rs760313375 Chr.17: 7190507-7190507 on Build GRCh38
rs772099517 Chr.17: 7190670-7190670 on Build GRCh38
rs185380227 Chr.17: 7190487-7190487 on Build GRCh38
rs572944836 Chr.17: 7190262-7190262 on Build GRCh38
rs957807280 Chr.17: 7190418-7190418 on Build GRCh38
rs932499704 Chr.17: 7190489-7190489 on Build GRCh38
rs1008651657 Chr.17: 7190601-7190601 on Build GRCh38
rs922460091 Chr.17: 7190224-7190224 on Build GRCh38
rs531186127 Chr.17: 7190618-7190619 on Build GRCh38
rs765823538 Chr.17: 7190488-7190488 on Build GRCh38
rs932468541 Chr.17: 7190233-7190233 on Build GRCh38
rs958135320 Chr.17: 7190452-7190452 on Build GRCh38
rs1025223110 Chr.17: 7190339-7190339 on Build GRCh38
rs868086620 Chr.17: 7190465-7190465 on Build GRCh38
rs550814978 Chr.17: 7190533-7190533 on Build GRCh38
rs931419284 Chr.17: 7190547-7190547 on Build GRCh38
rs1047116339 Chr.17: 7190624-7190624 on Build GRCh38
rs386795008 Chr.17: 7190530-7190532 on Build GRCh38
rs536003363 Chr.17: 7190628-7190628 on Build GRCh38
rs990555970 Chr.17: 7190423-7190423 on Build GRCh38
rs1008202774 Chr.17: 7190337-7190337 on Build GRCh38
rs775631780 Chr.17: 7190674-7190674 on Build GRCh38
rs920423455 Chr.17: 7190526-7190526 on Build GRCh38
rs970894099 Chr.17: 7190344-7190344 on Build GRCh38
rs1032540870 Chr.17: 7190645-7190645 on Build GRCh38
rs898589489 Chr.17: 7190272-7190272 on Build GRCh38
rs1033781206 Chr.17: 7190443-7190443 on Build GRCh38
rs1049817395 Chr.17: 7190554-7190554 on Build GRCh38
rs970819219 Chr.17: 7190482-7190483 on Build GRCh38
rs568548018 Chr.17: 7190431-7190431 on Build GRCh38
rs200598498 Chr.17: 7190656-7190656 on Build GRCh38
rs1049753304 Chr.17: 7190333-7190333 on Build GRCh38
rs994163633 Chr.17: 7190334-7190334 on Build GRCh38
rs1037122275 Chr.17: 7190320-7190320 on Build GRCh38

View Full Product Details