Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7227806-7228207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL2
Entrez Gene ID
1856
Gene Name
dishevelled segment polarity protein 2
Gene Aliases
-
UniGene
Hs.118640
Gene Chromosome Location
Chr.17: 7225342-7234548 on Build GRCh38
Transcripts
NM_004422.2 XM_005256502.1 XM_017024298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4580052 c.1092T>A p.T364T Chr.17: 7227987-7227987 on Build GRCh38
4909640 c.1100G>T p.R367L Chr.17: 7227979-7227979 on Build GRCh38
2976661 c.1075C>A p.P359T Chr.17: 7228004-7228004 on Build GRCh38
4069210 c.1076C>A p.P359H Chr.17: 7228003-7228003 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs938821105 Chr.17: 7227947-7227947 on Build GRCh38
rs756414359 Chr.17: 7228084-7228084 on Build GRCh38
rs763785958 Chr.17: 7228050-7228050 on Build GRCh38
rs985871518 Chr.17: 7227838-7227838 on Build GRCh38
rs145282702 Chr.17: 7227980-7227980 on Build GRCh38
rs764026787 Chr.17: 7227956-7227956 on Build GRCh38
rs963116020 Chr.17: 7227844-7227844 on Build GRCh38
rs761710493 Chr.17: 7228053-7228053 on Build GRCh38
rs1026789448 Chr.17: 7228109-7228109 on Build GRCh38
rs77412278 Chr.17: 7227874-7227874 on Build GRCh38
rs763476780 Chr.17: 7227941-7227942 on Build GRCh38
rs747135469 Chr.17: 7227966-7227966 on Build GRCh38
rs535994494 Chr.17: 7228064-7228064 on Build GRCh38
rs143084690 Chr.17: 7228004-7228004 on Build GRCh38
rs1053193592 Chr.17: 7228023-7228023 on Build GRCh38
rs1030137895 Chr.17: 7227834-7227834 on Build GRCh38
rs1017290510 Chr.17: 7227841-7227841 on Build GRCh38
rs533533057 Chr.17: 7228073-7228073 on Build GRCh38
rs374113002 Chr.17: 7227935-7227935 on Build GRCh38
rs897029333 Chr.17: 7228099-7228099 on Build GRCh38
rs1044093221 Chr.17: 7228060-7228060 on Build GRCh38
rs137865252 Chr.17: 7227981-7227981 on Build GRCh38
rs781405704 Chr.17: 7228074-7228074 on Build GRCh38
rs538930879 Chr.17: 7228172-7228172 on Build GRCh38
rs749743656 Chr.17: 7227997-7227997 on Build GRCh38
rs1017342579 Chr.17: 7228141-7228141 on Build GRCh38
rs759343637 Chr.17: 7228180-7228180 on Build GRCh38
rs369472570 Chr.17: 7227932-7227932 on Build GRCh38
rs928803265 Chr.17: 7227911-7227911 on Build GRCh38
rs930224652 Chr.17: 7227895-7227895 on Build GRCh38
rs568437108 Chr.17: 7227915-7227915 on Build GRCh38
rs753715250 Chr.17: 7227959-7227959 on Build GRCh38
rs781418768 Chr.17: 7227979-7227979 on Build GRCh38
rs952085183 Chr.17: 7227852-7227852 on Build GRCh38
rs866811166 Chr.17: 7227872-7227872 on Build GRCh38
rs774059800 Chr.17: 7227835-7227835 on Build GRCh38
rs546945495 Chr.17: 7228135-7228135 on Build GRCh38
rs879485788 Chr.17: 7227904-7227904 on Build GRCh38
rs765980463 Chr.17: 7227830-7227830 on Build GRCh38
rs1026419544 Chr.17: 7228168-7228168 on Build GRCh38
rs371100454 Chr.17: 7228101-7228101 on Build GRCh38
rs965064301 Chr.17: 7228162-7228162 on Build GRCh38
rs890310506 Chr.17: 7228069-7228069 on Build GRCh38
rs188458420 Chr.17: 7228157-7228157 on Build GRCh38
rs775018358 Chr.17: 7228026-7228026 on Build GRCh38
rs994558085 Chr.17: 7228161-7228161 on Build GRCh38
rs760443700 Chr.17: 7228040-7228040 on Build GRCh38
rs780087339 Chr.17: 7227965-7227965 on Build GRCh38
rs773689180 Chr.17: 7227994-7227994 on Build GRCh38
rs375577751 Chr.17: 7227954-7227954 on Build GRCh38
rs752853482 Chr.17: 7228077-7228077 on Build GRCh38
rs370287833 Chr.17: 7227987-7227987 on Build GRCh38
rs866399492 Chr.17: 7228042-7228042 on Build GRCh38
rs752496332 Chr.17: 7227937-7227937 on Build GRCh38
rs751594569 Chr.17: 7228071-7228071 on Build GRCh38
rs1006291277 Chr.17: 7228144-7228144 on Build GRCh38
rs767229371 Chr.17: 7228018-7228018 on Build GRCh38
rs62059164 Chr.17: 7228052-7228052 on Build GRCh38
rs757244896 Chr.17: 7227960-7227960 on Build GRCh38
rs777974146 Chr.17: 7228088-7228088 on Build GRCh38
rs905525697 Chr.17: 7228065-7228065 on Build GRCh38
rs983009869 Chr.17: 7227902-7227902 on Build GRCh38
rs766408450 Chr.17: 7228068-7228069 on Build GRCh38
rs547857082 Chr.17: 7228014-7228014 on Build GRCh38
rs755154551 Chr.17: 7227978-7227978 on Build GRCh38
rs145138687 Chr.17: 7228013-7228013 on Build GRCh38
rs771612069 Chr.17: 7228001-7228001 on Build GRCh38
rs918896609 Chr.17: 7227883-7227883 on Build GRCh38
rs376020506 Chr.17: 7227982-7227982 on Build GRCh38
rs952135925 Chr.17: 7228128-7228128 on Build GRCh38

View Full Product Details