Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7259516-7260012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN7
Entrez Gene ID
1366
Gene Name
claudin 7
Gene Aliases
CEPTRL2, CLDN-7, CPETRL2, Hs.84359, claudin-1
UniGene
Hs.513915
Gene Chromosome Location
Chr.17: 7259903-7263193 on Build GRCh38
Transcripts
NM_001185022.1 NM_001185023.1 NM_001307.5 NM_015362.4 NM_203413.2 NM_203414.2 NM_203415.1 XM_011523779.2

Gene Symbol
ELP5
Entrez Gene ID
23587
Gene Name
elongator acetyltransferase complex subunit 5
Gene Aliases
C17orf81, DERP6, HSPC002, MST071, MSTP071
UniGene
Hs.417029
Gene Chromosome Location
Chr.17: 7252053-7259940 on Build GRCh38
Transcripts
NM_001185022.1 NM_001185023.1 NM_001307.5 NM_015362.4 NM_203413.2 NM_203414.2 NM_203415.1 XM_011523779.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1246584 c.884T>G p.I295S Chr.17: 7259618-7259618 on Build GRCh38
1246585 c.928C>A p.P310T Chr.17: 7259662-7259662 on Build GRCh38
6998740 c.910G>A p.D304N Chr.17: 7259644-7259644 on Build GRCh38
1610710 c.925G>T p.D309Y Chr.17: 7259659-7259659 on Build GRCh38
4437018 c.931G>C p.D311H Chr.17: 7259665-7259665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751763722 Chr.17: 7259671-7259671 on Build GRCh38
rs763628506 Chr.17: 7259568-7259571 on Build GRCh38
rs975948432 Chr.17: 7259604-7259604 on Build GRCh38
rs750522156 Chr.17: 7259588-7259588 on Build GRCh38
rs958914936 Chr.17: 7259971-7259971 on Build GRCh38
rs937613304 Chr.17: 7259772-7259772 on Build GRCh38
rs79033455 Chr.17: 7259881-7259881 on Build GRCh38
rs1026141799 Chr.17: 7259912-7259912 on Build GRCh38
rs928820753 Chr.17: 7259950-7259950 on Build GRCh38
rs755169129 Chr.17: 7259685-7259685 on Build GRCh38
rs981552311 Chr.17: 7259918-7259918 on Build GRCh38
rs375900480 Chr.17: 7259720-7259720 on Build GRCh38
rs201957922 Chr.17: 7259587-7259587 on Build GRCh38
rs538863089 Chr.17: 7259623-7259623 on Build GRCh38
rs143238064 Chr.17: 7259895-7259895 on Build GRCh38
rs753873628 Chr.17: 7259666-7259666 on Build GRCh38
rs759194711 Chr.17: 7259628-7259628 on Build GRCh38
rs543910780 Chr.17: 7259805-7259805 on Build GRCh38
rs766596642 Chr.17: 7259670-7259670 on Build GRCh38
rs574395150 Chr.17: 7259851-7259851 on Build GRCh38
rs201910708 Chr.17: 7259724-7259724 on Build GRCh38
rs754745755 Chr.17: 7259811-7259811 on Build GRCh38
rs763074889 Chr.17: 7259668-7259668 on Build GRCh38
rs1005796959 Chr.17: 7259857-7259857 on Build GRCh38
rs1054689358 Chr.17: 7259778-7259778 on Build GRCh38
rs772079374 Chr.17: 7259766-7259766 on Build GRCh38
rs981609553 Chr.17: 7259743-7259743 on Build GRCh38
rs1043865413 Chr.17: 7259817-7259817 on Build GRCh38
rs1014272634 Chr.17: 7259836-7259836 on Build GRCh38
rs17849664 Chr.17: 7259641-7259641 on Build GRCh38
rs758587929 Chr.17: 7259589-7259589 on Build GRCh38
rs755327625 Chr.17: 7259598-7259598 on Build GRCh38
rs141204395 Chr.17: 7259612-7259612 on Build GRCh38
rs748775737 Chr.17: 7259622-7259622 on Build GRCh38
rs987824996 Chr.17: 7259635-7259635 on Build GRCh38
rs751227713 Chr.17: 7259784-7259784 on Build GRCh38
rs536208932 Chr.17: 7259545-7259545 on Build GRCh38
rs753485217 Chr.17: 7259571-7259572 on Build GRCh38
rs148656210 Chr.17: 7259590-7259590 on Build GRCh38
rs1022095404 Chr.17: 7259558-7259558 on Build GRCh38
rs756553203 Chr.17: 7259701-7259701 on Build GRCh38
rs995559536 Chr.17: 7259902-7259902 on Build GRCh38
rs773955632 Chr.17: 7259624-7259624 on Build GRCh38
rs891022495 Chr.17: 7259878-7259878 on Build GRCh38
rs771846696 Chr.17: 7259630-7259630 on Build GRCh38
rs749829467 Chr.17: 7259711-7259711 on Build GRCh38
rs778411564 Chr.17: 7259722-7259722 on Build GRCh38
rs913606465 Chr.17: 7259916-7259916 on Build GRCh38
rs377718990 Chr.17: 7259932-7259934 on Build GRCh38
rs781414906 Chr.17: 7259601-7259601 on Build GRCh38
rs1017188443 Chr.17: 7259859-7259859 on Build GRCh38
rs558309811 Chr.17: 7259707-7259707 on Build GRCh38
rs894812447 Chr.17: 7259801-7259801 on Build GRCh38
rs752618499 Chr.17: 7259562-7259562 on Build GRCh38
rs937900774 Chr.17: 7259967-7259967 on Build GRCh38
rs779497605 Chr.17: 7259986-7259986 on Build GRCh38
rs930131576 Chr.17: 7259726-7259726 on Build GRCh38
rs745445881 Chr.17: 7259723-7259723 on Build GRCh38
rs955304804 Chr.17: 7259591-7259591 on Build GRCh38
rs557697234 Chr.17: 7259978-7259978 on Build GRCh38
rs528757529 Chr.17: 7259883-7259883 on Build GRCh38
rs561746637 Chr.17: 7259561-7259561 on Build GRCh38
rs748629916 Chr.17: 7259602-7259602 on Build GRCh38
rs565471914 Chr.17: 7259921-7259921 on Build GRCh38
rs548498221 Chr.17: 7259818-7259818 on Build GRCh38
rs539811443 Chr.17: 7259871-7259871 on Build GRCh38
rs756894928 Chr.17: 7259683-7259686 on Build GRCh38
rs199964921 Chr.17: 7259593-7259593 on Build GRCh38
rs763903125 Chr.17: 7259661-7259661 on Build GRCh38
rs770198667 Chr.17: 7259608-7259608 on Build GRCh38
rs371307004 Chr.17: 7259692-7259692 on Build GRCh38
rs1047079880 Chr.17: 7259838-7259838 on Build GRCh38
rs775138826 Chr.17: 7259636-7259636 on Build GRCh38
rs1050097 Chr.17: 7259753-7259753 on Build GRCh38
rs752973958 Chr.17: 7259697-7259697 on Build GRCh38
rs951524203 Chr.17: 7259915-7259915 on Build GRCh38
rs926572189 Chr.17: 7259964-7259965 on Build GRCh38
rs760582477 Chr.17: 7259658-7259658 on Build GRCh38
rs999407865 Chr.17: 7259570-7259570 on Build GRCh38
rs375018055 Chr.17: 7259548-7259548 on Build GRCh38
rs999349296 Chr.17: 7259839-7259839 on Build GRCh38

View Full Product Details