Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7260743-7261243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN7
Entrez Gene ID
1366
Gene Name
claudin 7
Gene Aliases
CEPTRL2, CLDN-7, CPETRL2, Hs.84359, claudin-1
UniGene
Hs.513915
Gene Chromosome Location
Chr.17: 7259903-7263193 on Build GRCh38
Transcripts
NM_001185022.1 NM_001185023.1 NM_001307.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6440114 c.270_271GG>AA p.G91S Chr.17: 7260938-7260939 on Build GRCh38
4965086 c.224-8G>C p.? Chr.17: 7260993-7260993 on Build GRCh38
3521665 c.271G>A p.G91S Chr.17: 7260938-7260938 on Build GRCh38
6348463 c.245C>A p.A82D Chr.17: 7260964-7260964 on Build GRCh38
983903 c.315G>A p.T105T Chr.17: 7260894-7260894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373478236 Chr.17: 7260910-7260910 on Build GRCh38
rs112566076 Chr.17: 7261180-7261180 on Build GRCh38
rs142682069 Chr.17: 7260887-7260887 on Build GRCh38
rs959916609 Chr.17: 7261176-7261176 on Build GRCh38
rs774011626 Chr.17: 7260994-7260994 on Build GRCh38
rs768087014 Chr.17: 7260796-7260796 on Build GRCh38
rs754006543 Chr.17: 7260857-7260857 on Build GRCh38
rs755628370 Chr.17: 7260818-7260818 on Build GRCh38
rs758959441 Chr.17: 7260914-7260914 on Build GRCh38
rs1008692911 Chr.17: 7261202-7261202 on Build GRCh38
rs746209260 Chr.17: 7260873-7260873 on Build GRCh38
rs867519822 Chr.17: 7261091-7261091 on Build GRCh38
rs547957587 Chr.17: 7260929-7260929 on Build GRCh38
rs369146758 Chr.17: 7260883-7260883 on Build GRCh38
rs750701862 Chr.17: 7260941-7260941 on Build GRCh38
rs867054456 Chr.17: 7260886-7260886 on Build GRCh38
rs369509007 Chr.17: 7260791-7260791 on Build GRCh38
rs145889982 Chr.17: 7260801-7260801 on Build GRCh38
rs776184919 Chr.17: 7260781-7260781 on Build GRCh38
rs747479920 Chr.17: 7260882-7260882 on Build GRCh38
rs774841188 Chr.17: 7260773-7260773 on Build GRCh38
rs544848539 Chr.17: 7261150-7261150 on Build GRCh38
rs963744067 Chr.17: 7261059-7261059 on Build GRCh38
rs568192125 Chr.17: 7260945-7260945 on Build GRCh38
rs954296922 Chr.17: 7260953-7260953 on Build GRCh38
rs113692621 Chr.17: 7261051-7261051 on Build GRCh38
rs750216913 Chr.17: 7260805-7260805 on Build GRCh38
rs986873517 Chr.17: 7260958-7260958 on Build GRCh38
rs781744000 Chr.17: 7260968-7260968 on Build GRCh38
rs575993475 Chr.17: 7261086-7261086 on Build GRCh38
rs1019252206 Chr.17: 7261053-7261053 on Build GRCh38
rs765416503 Chr.17: 7261027-7261027 on Build GRCh38
rs776934659 Chr.17: 7261105-7261105 on Build GRCh38
rs769041772 Chr.17: 7260967-7260967 on Build GRCh38
rs878992849 Chr.17: 7260987-7260987 on Build GRCh38
rs756152576 Chr.17: 7260846-7260846 on Build GRCh38
rs111255205 Chr.17: 7260993-7260993 on Build GRCh38
rs1019137268 Chr.17: 7261151-7261151 on Build GRCh38
rs751474604 Chr.17: 7260819-7260819 on Build GRCh38
rs952595706 Chr.17: 7261120-7261120 on Build GRCh38
rs772811592 Chr.17: 7260992-7260992 on Build GRCh38
rs533639505 Chr.17: 7260990-7260990 on Build GRCh38
rs769316794 Chr.17: 7260991-7260991 on Build GRCh38
rs779314588 Chr.17: 7260862-7260862 on Build GRCh38
rs144832882 Chr.17: 7260912-7260912 on Build GRCh38
rs576767384 Chr.17: 7260995-7260995 on Build GRCh38
rs762687113 Chr.17: 7260909-7260909 on Build GRCh38
rs996951812 Chr.17: 7261078-7261078 on Build GRCh38
rs908450344 Chr.17: 7260925-7260925 on Build GRCh38
rs999467896 Chr.17: 7260772-7260772 on Build GRCh38
rs750619958 Chr.17: 7261029-7261029 on Build GRCh38
rs929702652 Chr.17: 7260929-7260943 on Build GRCh38
rs963756863 Chr.17: 7260824-7260824 on Build GRCh38
rs761455420 Chr.17: 7260892-7260892 on Build GRCh38
rs568777982 Chr.17: 7260878-7260878 on Build GRCh38
rs940602923 Chr.17: 7261090-7261090 on Build GRCh38
rs915866614 Chr.17: 7261204-7261204 on Build GRCh38
rs756267361 Chr.17: 7260771-7260772 on Build GRCh38
rs762778751 Chr.17: 7260808-7260808 on Build GRCh38
rs757414858 Chr.17: 7260859-7260859 on Build GRCh38
rs758773722 Chr.17: 7260944-7260944 on Build GRCh38
rs769406740 Chr.17: 7260894-7260894 on Build GRCh38
rs746299150 Chr.17: 7260780-7260780 on Build GRCh38
rs890686847 Chr.17: 7261047-7261047 on Build GRCh38
rs149308129 Chr.17: 7260947-7260947 on Build GRCh38
rs761388518 Chr.17: 7260800-7260800 on Build GRCh38
rs779224586 Chr.17: 7260933-7260933 on Build GRCh38
rs757459277 Chr.17: 7260926-7260926 on Build GRCh38
rs999018882 Chr.17: 7261031-7261031 on Build GRCh38
rs948772806 Chr.17: 7261003-7261003 on Build GRCh38
rs777744847 Chr.17: 7260990-7260995 on Build GRCh38
rs772901497 Chr.17: 7260904-7260904 on Build GRCh38
rs539414705 Chr.17: 7261044-7261044 on Build GRCh38
rs749461011 Chr.17: 7261007-7261008 on Build GRCh38
rs775516909 Chr.17: 7261015-7261015 on Build GRCh38
rs760178187 Chr.17: 7260792-7260792 on Build GRCh38
rs758683602 Chr.17: 7261030-7261030 on Build GRCh38
rs767332527 Chr.17: 7260998-7260998 on Build GRCh38
rs1019664423 Chr.17: 7260981-7260981 on Build GRCh38
rs201481204 Chr.17: 7260798-7260798 on Build GRCh38
rs560743926 Chr.17: 7260948-7260948 on Build GRCh38
rs982557887 Chr.17: 7261133-7261133 on Build GRCh38
rs995871763 Chr.17: 7261124-7261124 on Build GRCh38
rs777728095 Chr.17: 7260854-7260854 on Build GRCh38

View Full Product Details