Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 10711087-10711636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRM
Entrez Gene ID
56985
Gene Name
ADP-ribose/CDP-alcohol diphosphatase, manganese dependent
Gene Aliases
C17orf48, MDS006, NBLA03831
UniGene
Hs.47668
Gene Chromosome Location
Chr.17: 10697610-10711558 on Build GRCh38
Transcripts
NM_020233.4 XM_005256738.2 XM_017024616.1 XM_017024617.1

Gene Symbol
TMEM220
Entrez Gene ID
388335
Gene Name
transmembrane protein 220
Gene Aliases
-
UniGene
Hs.462230
Gene Chromosome Location
Chr.17: 10711084-10730329 on Build GRCh38
Transcripts
NM_020233.4 XM_005256738.2 XM_017024616.1 XM_017024617.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3765978 c.1010A>G p.E337G Chr.17: 10711125-10711125 on Build GRCh38
6595362 c.1005G>T p.K335N Chr.17: 10711120-10711120 on Build GRCh38
6595363 c.1028A>C p.*343S Chr.17: 10711143-10711143 on Build GRCh38
6220821 c.982delC p.P328fs*5 Chr.17: 10711097-10711097 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs970014796 Chr.17: 10711423-10711424 on Build GRCh38
rs931879014 Chr.17: 10711554-10711554 on Build GRCh38
rs900412212 Chr.17: 10711351-10711351 on Build GRCh38
rs386795326 Chr.17: 10711550-10711552 on Build GRCh38
rs756075874 Chr.17: 10711564-10711564 on Build GRCh38
rs1042769567 Chr.17: 10711333-10711333 on Build GRCh38
rs185933947 Chr.17: 10711353-10711353 on Build GRCh38
rs1014326249 Chr.17: 10711399-10711399 on Build GRCh38
rs73974820 Chr.17: 10711470-10711470 on Build GRCh38
rs1051646964 Chr.17: 10711572-10711572 on Build GRCh38
rs118080120 Chr.17: 10711320-10711320 on Build GRCh38
rs578207345 Chr.17: 10711409-10711409 on Build GRCh38
rs140005480 Chr.17: 10711141-10711141 on Build GRCh38
rs556610845 Chr.17: 10711393-10711393 on Build GRCh38
rs982362535 Chr.17: 10711442-10711442 on Build GRCh38
rs947024437 Chr.17: 10711324-10711324 on Build GRCh38
rs190733292 Chr.17: 10711457-10711457 on Build GRCh38
rs445928 Chr.17: 10711356-10711356 on Build GRCh38
rs379403 Chr.17: 10711550-10711550 on Build GRCh38
rs921688034 Chr.17: 10711542-10711542 on Build GRCh38
rs573939100 Chr.17: 10711365-10711365 on Build GRCh38
rs912974168 Chr.17: 10711274-10711274 on Build GRCh38
rs868190726 Chr.17: 10711589-10711589 on Build GRCh38
rs995241103 Chr.17: 10711403-10711403 on Build GRCh38
rs947191003 Chr.17: 10711512-10711512 on Build GRCh38
rs890399274 Chr.17: 10711579-10711579 on Build GRCh38
rs944490453 Chr.17: 10711275-10711275 on Build GRCh38
rs531433630 Chr.17: 10711599-10711599 on Build GRCh38
rs912970939 Chr.17: 10711500-10711500 on Build GRCh38
rs957551937 Chr.17: 10711496-10711496 on Build GRCh38
rs1005709308 Chr.17: 10711354-10711354 on Build GRCh38
rs1053402094 Chr.17: 10711214-10711214 on Build GRCh38
rs538529693 Chr.17: 10711385-10711385 on Build GRCh38
rs1015407769 Chr.17: 10711384-10711384 on Build GRCh38
rs749023265 Chr.17: 10711189-10711189 on Build GRCh38
rs551768488 Chr.17: 10711336-10711336 on Build GRCh38
rs777849706 Chr.17: 10711267-10711267 on Build GRCh38
rs567932336 Chr.17: 10711236-10711236 on Build GRCh38
rs387022 Chr.17: 10711552-10711552 on Build GRCh38
rs970010423 Chr.17: 10711415-10711415 on Build GRCh38
rs146377569 Chr.17: 10711352-10711352 on Build GRCh38
rs1023670851 Chr.17: 10711410-10711410 on Build GRCh38
rs759991903 Chr.17: 10711276-10711276 on Build GRCh38
rs928092349 Chr.17: 10711455-10711455 on Build GRCh38
rs1024410637 Chr.17: 10711415-10711418 on Build GRCh38
rs1004893329 Chr.17: 10711364-10711364 on Build GRCh38
rs572130702 Chr.17: 10711534-10711534 on Build GRCh38
rs770073148 Chr.17: 10711137-10711137 on Build GRCh38
rs406446 Chr.17: 10711125-10711125 on Build GRCh38
rs1014440810 Chr.17: 10711600-10711600 on Build GRCh38

View Full Product Details