Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 12992693-12993190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAC2
Entrez Gene ID
60528
Gene Name
elaC ribonuclease Z 2
Gene Aliases
COXPD17, ELC2, HPC2
UniGene
Hs.434232
Gene Chromosome Location
Chr.17: 12991612-13018064 on Build GRCh38
Transcripts
NM_001165962.1 NM_018127.6 NM_173717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5921155 c.2440C>T p.H814Y Chr.17: 12992859-12992859 on Build GRCh38
4064030 c.2404G>A p.G802S Chr.17: 12992895-12992895 on Build GRCh38
4680695 c.2337G>A p.E779E Chr.17: 12992962-12992962 on Build GRCh38
302123 c.2403C>T p.G801G Chr.17: 12992896-12992896 on Build GRCh38
3514399 c.2353C>T p.R785W Chr.17: 12992946-12992946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755369174 Chr.17: 12992938-12992938 on Build GRCh38
rs770865114 Chr.17: 12992958-12992958 on Build GRCh38
rs755662266 Chr.17: 12992769-12992769 on Build GRCh38
rs142986208 Chr.17: 12993139-12993139 on Build GRCh38
rs370729050 Chr.17: 12993066-12993066 on Build GRCh38
rs761628384 Chr.17: 12992854-12992855 on Build GRCh38
rs958202463 Chr.17: 12992990-12992990 on Build GRCh38
rs200771997 Chr.17: 12992785-12992785 on Build GRCh38
rs878944913 Chr.17: 12993047-12993058 on Build GRCh38
rs747444748 Chr.17: 12992892-12992892 on Build GRCh38
rs374981739 Chr.17: 12992743-12992743 on Build GRCh38
rs762446590 Chr.17: 12993091-12993091 on Build GRCh38
rs892274034 Chr.17: 12992786-12992786 on Build GRCh38
rs11545304 Chr.17: 12992731-12992731 on Build GRCh38
rs550032568 Chr.17: 12992980-12992980 on Build GRCh38
rs777147755 Chr.17: 12992896-12992896 on Build GRCh38
rs1014032579 Chr.17: 12993137-12993137 on Build GRCh38
rs894210423 Chr.17: 12993136-12993136 on Build GRCh38
rs770669443 Chr.17: 12992883-12992883 on Build GRCh38
rs989543533 Chr.17: 12992802-12992802 on Build GRCh38
rs745926343 Chr.17: 12993058-12993058 on Build GRCh38
rs971374535 Chr.17: 12992976-12992976 on Build GRCh38
rs775820542 Chr.17: 12993062-12993062 on Build GRCh38
rs371543722 Chr.17: 12993004-12993004 on Build GRCh38
rs200249647 Chr.17: 12992783-12992783 on Build GRCh38
rs991035115 Chr.17: 12992759-12992759 on Build GRCh38
rs749996506 Chr.17: 12992937-12992937 on Build GRCh38
rs781469896 Chr.17: 12992939-12992939 on Build GRCh38
rs119484086 Chr.17: 12992957-12992957 on Build GRCh38
rs774414219 Chr.17: 12992921-12992921 on Build GRCh38
rs759606875 Chr.17: 12992973-12992973 on Build GRCh38
rs748669284 Chr.17: 12992947-12992947 on Build GRCh38
rs775479429 Chr.17: 12992974-12992974 on Build GRCh38
rs762659201 Chr.17: 12992954-12992954 on Build GRCh38
rs769868918 Chr.17: 12992902-12992902 on Build GRCh38
rs767100578 Chr.17: 12993147-12993147 on Build GRCh38
rs760538357 Chr.17: 12992979-12992979 on Build GRCh38
rs768527037 Chr.17: 12993069-12993069 on Build GRCh38
rs1045782172 Chr.17: 12993141-12993141 on Build GRCh38
rs761410635 Chr.17: 12992992-12992992 on Build GRCh38
rs143840985 Chr.17: 12992929-12992929 on Build GRCh38
rs201700166 Chr.17: 12992890-12992890 on Build GRCh38
rs778221408 Chr.17: 12992949-12992949 on Build GRCh38
rs750097249 Chr.17: 12993003-12993003 on Build GRCh38
rs779668862 Chr.17: 12993008-12993008 on Build GRCh38
rs763169771 Chr.17: 12992846-12992846 on Build GRCh38
rs776663945 Chr.17: 12993074-12993074 on Build GRCh38
rs199998904 Chr.17: 12993024-12993024 on Build GRCh38
rs748616092 Chr.17: 12992899-12992899 on Build GRCh38
rs753701032 Chr.17: 12992983-12992983 on Build GRCh38
rs180717697 Chr.17: 12992923-12992923 on Build GRCh38
rs1005336453 Chr.17: 12992719-12992719 on Build GRCh38
rs772658366 Chr.17: 12993089-12993089 on Build GRCh38
rs756198729 Chr.17: 12993066-12993067 on Build GRCh38
rs753128082 Chr.17: 12993019-12993019 on Build GRCh38
rs777588472 Chr.17: 12992772-12992772 on Build GRCh38
rs773456696 Chr.17: 12992842-12992842 on Build GRCh38
rs199747602 Chr.17: 12992801-12992801 on Build GRCh38
rs773219330 Chr.17: 12992906-12992906 on Build GRCh38
rs888296975 Chr.17: 12992909-12992909 on Build GRCh38
rs767378629 Chr.17: 12992924-12992924 on Build GRCh38
rs147779718 Chr.17: 12992895-12992895 on Build GRCh38
rs1055673968 Chr.17: 12993124-12993124 on Build GRCh38
rs553919919 Chr.17: 12993056-12993056 on Build GRCh38
rs749717060 Chr.17: 12993044-12993044 on Build GRCh38
rs958086224 Chr.17: 12992799-12992799 on Build GRCh38
rs922510869 Chr.17: 12992873-12992873 on Build GRCh38
rs866746571 Chr.17: 12992817-12992817 on Build GRCh38
rs764787999 Chr.17: 12992946-12992947 on Build GRCh38
rs774664159 Chr.17: 12992966-12992966 on Build GRCh38
rs386795520 Chr.17: 12992875-12992877 on Build GRCh38
rs751437403 Chr.17: 12992862-12992862 on Build GRCh38
rs991895115 Chr.17: 12992723-12992723 on Build GRCh38
rs767611235 Chr.17: 12992865-12992865 on Build GRCh38
rs13641 Chr.17: 12992757-12992757 on Build GRCh38
rs372201309 Chr.17: 12992787-12992787 on Build GRCh38
rs540886198 Chr.17: 12992766-12992766 on Build GRCh38
rs761933183 Chr.17: 12992809-12992809 on Build GRCh38
rs941164325 Chr.17: 12993111-12993111 on Build GRCh38
rs567494102 Chr.17: 12992936-12992936 on Build GRCh38
rs926500016 Chr.17: 12992925-12992926 on Build GRCh38
rs777762288 Chr.17: 12993080-12993083 on Build GRCh38
rs574860371 Chr.17: 12992818-12992818 on Build GRCh38
rs746853159 Chr.17: 12993064-12993064 on Build GRCh38
rs200827637 Chr.17: 12992948-12992948 on Build GRCh38
rs148819805 Chr.17: 12992927-12992927 on Build GRCh38
rs756694442 Chr.17: 12992930-12992930 on Build GRCh38
rs778365597 Chr.17: 12993027-12993027 on Build GRCh38
rs902866028 Chr.17: 12993148-12993148 on Build GRCh38
rs376222314 Chr.17: 12993059-12993059 on Build GRCh38
rs186366777 Chr.17: 12993053-12993053 on Build GRCh38
rs553830170 Chr.17: 12992725-12992725 on Build GRCh38
rs560083267 Chr.17: 12992864-12992864 on Build GRCh38
rs766100666 Chr.17: 12992850-12992850 on Build GRCh38
rs749364898 Chr.17: 12993080-12993085 on Build GRCh38
rs989490256 Chr.17: 12993021-12993021 on Build GRCh38
rs771045547 Chr.17: 12993051-12993051 on Build GRCh38
rs917643446 Chr.17: 12992915-12992915 on Build GRCh38
rs144228112 Chr.17: 12992881-12992881 on Build GRCh38
rs764615178 Chr.17: 12993001-12993001 on Build GRCh38
rs779706680 Chr.17: 12992833-12992833 on Build GRCh38
rs922658791 Chr.17: 12993050-12993050 on Build GRCh38
rs913810768 Chr.17: 12993158-12993158 on Build GRCh38
rs752005883 Chr.17: 12992948-12992949 on Build GRCh38
rs562627181 Chr.17: 12992816-12992816 on Build GRCh38
rs756987850 Chr.17: 12992885-12992885 on Build GRCh38
rs1025606264 Chr.17: 12993083-12993083 on Build GRCh38
rs752630201 Chr.17: 12992868-12992868 on Build GRCh38
rs987811809 Chr.17: 12993140-12993140 on Build GRCh38
rs372457406 Chr.17: 12993094-12993094 on Build GRCh38
rs748902752 Chr.17: 12992882-12992882 on Build GRCh38
rs764105172 Chr.17: 12992928-12992928 on Build GRCh38
rs755713504 Chr.17: 12992872-12992872 on Build GRCh38
rs141048290 Chr.17: 12993010-12993010 on Build GRCh38
rs771225119 Chr.17: 12992907-12992907 on Build GRCh38
rs73978302 Chr.17: 12993081-12993081 on Build GRCh38
rs533278704 Chr.17: 12992819-12992819 on Build GRCh38
rs777094768 Chr.17: 12992805-12992805 on Build GRCh38
rs776429479 Chr.17: 12992833-12992835 on Build GRCh38
rs969148799 Chr.17: 12993086-12993086 on Build GRCh38
rs751155734 Chr.17: 12993018-12993018 on Build GRCh38
rs764819139 Chr.17: 12993085-12993085 on Build GRCh38
rs878855051 Chr.17: 12992883-12992884 on Build GRCh38
rs977140471 Chr.17: 12993036-12993036 on Build GRCh38
rs755267324 Chr.17: 12992949-12992950 on Build GRCh38
rs376515152 Chr.17: 12992832-12992832 on Build GRCh38
rs765254360 Chr.17: 12992814-12992814 on Build GRCh38
rs1045139477 Chr.17: 12992762-12992765 on Build GRCh38
rs768102931 Chr.17: 12992952-12992953 on Build GRCh38
rs1025638975 Chr.17: 12992764-12992764 on Build GRCh38
rs996184495 Chr.17: 12992855-12992855 on Build GRCh38
rs1041310323 Chr.17: 12992967-12992967 on Build GRCh38
rs753154933 Chr.17: 12992990-12992991 on Build GRCh38

View Full Product Details