Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 12994286-12994826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAC2
Entrez Gene ID
60528
Gene Name
elaC ribonuclease Z 2
Gene Aliases
COXPD17, ELC2, HPC2
UniGene
Hs.434232
Gene Chromosome Location
Chr.17: 12991612-13018064 on Build GRCh38
Transcripts
NM_001165962.1 NM_018127.6 NM_173717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
472317 c.2036A>G p.D679G Chr.17: 12994497-12994497 on Build GRCh38
6259728 c.2102C>A p.T701K Chr.17: 12994431-12994431 on Build GRCh38
4895232 c.1991C>T p.S664F Chr.17: 12994802-12994802 on Build GRCh38
3514400 c.2023C>T p.R675W Chr.17: 12994770-12994770 on Build GRCh38
4064032 c.2099A>C p.K700T Chr.17: 12994434-12994434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750302074 Chr.17: 12994773-12994773 on Build GRCh38
rs757021921 Chr.17: 12994408-12994408 on Build GRCh38
rs374690415 Chr.17: 12994542-12994542 on Build GRCh38
rs548603402 Chr.17: 12994509-12994509 on Build GRCh38
rs116634952 Chr.17: 12994738-12994738 on Build GRCh38
rs757071308 Chr.17: 12994543-12994543 on Build GRCh38
rs760764029 Chr.17: 12994743-12994743 on Build GRCh38
rs760710912 Chr.17: 12994525-12994525 on Build GRCh38
rs752085443 Chr.17: 12994375-12994375 on Build GRCh38
rs774724028 Chr.17: 12994507-12994507 on Build GRCh38
rs775705116 Chr.17: 12994518-12994518 on Build GRCh38
rs959478306 Chr.17: 12994529-12994529 on Build GRCh38
rs746212999 Chr.17: 12994736-12994736 on Build GRCh38
rs753137788 Chr.17: 12994568-12994568 on Build GRCh38
rs751495465 Chr.17: 12994781-12994781 on Build GRCh38
rs1005165508 Chr.17: 12994367-12994367 on Build GRCh38
rs907170903 Chr.17: 12994601-12994601 on Build GRCh38
rs776342488 Chr.17: 12994543-12994544 on Build GRCh38
rs775834425 Chr.17: 12994740-12994740 on Build GRCh38
rs753669740 Chr.17: 12994399-12994399 on Build GRCh38
rs767897056 Chr.17: 12994515-12994515 on Build GRCh38
rs764640672 Chr.17: 12994732-12994732 on Build GRCh38
rs1033730754 Chr.17: 12994505-12994505 on Build GRCh38
rs780790017 Chr.17: 12994786-12994786 on Build GRCh38
rs964399559 Chr.17: 12994530-12994530 on Build GRCh38
rs764948567 Chr.17: 12994547-12994547 on Build GRCh38
rs747764248 Chr.17: 12994725-12994725 on Build GRCh38
rs746879525 Chr.17: 12994466-12994467 on Build GRCh38
rs541359069 Chr.17: 12994473-12994473 on Build GRCh38
rs766389446 Chr.17: 12994777-12994777 on Build GRCh38
rs377587005 Chr.17: 12994407-12994407 on Build GRCh38
rs530418086 Chr.17: 12994591-12994591 on Build GRCh38
rs369900061 Chr.17: 12994735-12994735 on Build GRCh38
rs761967944 Chr.17: 12994763-12994763 on Build GRCh38
rs761385155 Chr.17: 12994784-12994784 on Build GRCh38
rs779854666 Chr.17: 12994719-12994719 on Build GRCh38
rs771434310 Chr.17: 12994733-12994733 on Build GRCh38
rs199942266 Chr.17: 12994411-12994411 on Build GRCh38
rs746289536 Chr.17: 12994801-12994801 on Build GRCh38
rs771375007 Chr.17: 12994500-12994500 on Build GRCh38
rs138254982 Chr.17: 12994469-12994469 on Build GRCh38
rs1011025531 Chr.17: 12994729-12994729 on Build GRCh38
rs954534026 Chr.17: 12994556-12994556 on Build GRCh38
rs747296926 Chr.17: 12994677-12994677 on Build GRCh38
rs765368974 Chr.17: 12994769-12994769 on Build GRCh38
rs568498595 Chr.17: 12994557-12994557 on Build GRCh38
rs186237722 Chr.17: 12994320-12994320 on Build GRCh38
rs201882092 Chr.17: 12994382-12994382 on Build GRCh38
rs577239094 Chr.17: 12994457-12994457 on Build GRCh38
rs764116549 Chr.17: 12994745-12994745 on Build GRCh38
rs749721094 Chr.17: 12994497-12994497 on Build GRCh38
rs959320530 Chr.17: 12994310-12994310 on Build GRCh38
rs989490333 Chr.17: 12994312-12994312 on Build GRCh38
rs200759780 Chr.17: 12994776-12994776 on Build GRCh38
rs138847889 Chr.17: 12994549-12994549 on Build GRCh38
rs776392466 Chr.17: 12994493-12994493 on Build GRCh38
rs1021441356 Chr.17: 12994461-12994461 on Build GRCh38
rs1045612223 Chr.17: 12994605-12994605 on Build GRCh38
rs371343827 Chr.17: 12994538-12994538 on Build GRCh38
rs755897599 Chr.17: 12994797-12994797 on Build GRCh38
rs757313871 Chr.17: 12994791-12994791 on Build GRCh38
rs145824949 Chr.17: 12994371-12994371 on Build GRCh38
rs948810469 Chr.17: 12994589-12994589 on Build GRCh38
rs755559592 Chr.17: 12994372-12994372 on Build GRCh38
rs78988571 Chr.17: 12994479-12994479 on Build GRCh38
rs1002756423 Chr.17: 12994393-12994393 on Build GRCh38
rs190562664 Chr.17: 12994412-12994412 on Build GRCh38
rs895214427 Chr.17: 12994700-12994700 on Build GRCh38
rs1011457495 Chr.17: 12994645-12994645 on Build GRCh38
rs754567726 Chr.17: 12994782-12994782 on Build GRCh38
rs563445630 Chr.17: 12994582-12994582 on Build GRCh38
rs974420238 Chr.17: 12994331-12994331 on Build GRCh38
rs889538036 Chr.17: 12994365-12994365 on Build GRCh38
rs547932956 Chr.17: 12994581-12994581 on Build GRCh38
rs200293959 Chr.17: 12994398-12994398 on Build GRCh38
rs767033102 Chr.17: 12994374-12994374 on Build GRCh38
rs779563218 Chr.17: 12994447-12994447 on Build GRCh38
rs757853122 Chr.17: 12994413-12994413 on Build GRCh38
rs56329828 Chr.17: 12994352-12994352 on Build GRCh38
rs934138477 Chr.17: 12994569-12994569 on Build GRCh38
rs551786671 Chr.17: 12994612-12994612 on Build GRCh38
rs548083572 Chr.17: 12994723-12994723 on Build GRCh38
rs964452238 Chr.17: 12994316-12994316 on Build GRCh38
rs756416403 Chr.17: 12994387-12994387 on Build GRCh38
rs920105697 Chr.17: 12994584-12994584 on Build GRCh38
rs777258865 Chr.17: 12994800-12994800 on Build GRCh38
rs750343273 Chr.17: 12994548-12994548 on Build GRCh38
rs376231184 Chr.17: 12994790-12994790 on Build GRCh38
rs372106847 Chr.17: 12994385-12994385 on Build GRCh38
rs891708688 Chr.17: 12994632-12994632 on Build GRCh38
rs961586097 Chr.17: 12994793-12994793 on Build GRCh38
rs908675358 Chr.17: 12994353-12994353 on Build GRCh38
rs907121704 Chr.17: 12994609-12994609 on Build GRCh38
rs768715038 Chr.17: 12994506-12994506 on Build GRCh38
rs1053167590 Chr.17: 12994617-12994617 on Build GRCh38
rs781379178 Chr.17: 12994805-12994805 on Build GRCh38
rs766056867 Chr.17: 12994345-12994345 on Build GRCh38
rs776845488 Chr.17: 12994759-12994759 on Build GRCh38
rs764345429 Chr.17: 12994537-12994537 on Build GRCh38
rs537368002 Chr.17: 12994337-12994337 on Build GRCh38
rs865979216 Chr.17: 12994659-12994659 on Build GRCh38
rs879251188 Chr.17: 12994593-12994593 on Build GRCh38
rs538916762 Chr.17: 12994721-12994721 on Build GRCh38
rs759557739 Chr.17: 12994377-12994377 on Build GRCh38
rs940163499 Chr.17: 12994355-12994355 on Build GRCh38
rs989480488 Chr.17: 12994534-12994534 on Build GRCh38
rs752239590 Chr.17: 12994787-12994787 on Build GRCh38
rs148869775 Chr.17: 12994698-12994698 on Build GRCh38

View Full Product Details