Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 13000071-13000470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAC2
Entrez Gene ID
60528
Gene Name
elaC ribonuclease Z 2
Gene Aliases
COXPD17, ELC2, HPC2
UniGene
Hs.434232
Gene Chromosome Location
Chr.17: 12991612-13018064 on Build GRCh38
Transcripts
NM_001165962.1 NM_018127.6 NM_173717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
310851 c.1349C>T p.A450V Chr.17: 13000230-13000230 on Build GRCh38
4464297 c.1327C>T p.P443S Chr.17: 13000252-13000252 on Build GRCh38
3514409 c.1369C>T p.Q457* Chr.17: 13000210-13000210 on Build GRCh38
436051 c.1400C>T p.A467V Chr.17: 13000179-13000179 on Build GRCh38
6429815 c.1347G>T p.E449D Chr.17: 13000232-13000232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528816334 Chr.17: 13000403-13000403 on Build GRCh38
rs768766446 Chr.17: 13000282-13000285 on Build GRCh38
rs928412781 Chr.17: 13000450-13000450 on Build GRCh38
rs372663216 Chr.17: 13000174-13000174 on Build GRCh38
rs779254943 Chr.17: 13000276-13000276 on Build GRCh38
rs910841714 Chr.17: 13000109-13000109 on Build GRCh38
rs202196332 Chr.17: 13000280-13000281 on Build GRCh38
rs192492803 Chr.17: 13000303-13000303 on Build GRCh38
rs373251586 Chr.17: 13000145-13000145 on Build GRCh38
rs776280164 Chr.17: 13000226-13000226 on Build GRCh38
rs1004578729 Chr.17: 13000133-13000133 on Build GRCh38
rs768570432 Chr.17: 13000293-13000293 on Build GRCh38
rs757842867 Chr.17: 13000439-13000439 on Build GRCh38
rs996163635 Chr.17: 13000442-13000442 on Build GRCh38
rs758768902 Chr.17: 13000157-13000157 on Build GRCh38
rs200149324 Chr.17: 13000189-13000189 on Build GRCh38
rs753958264 Chr.17: 13000267-13000267 on Build GRCh38
rs138269137 Chr.17: 13000237-13000237 on Build GRCh38
rs373784693 Chr.17: 13000317-13000317 on Build GRCh38
rs375565327 Chr.17: 13000229-13000229 on Build GRCh38
rs752185416 Chr.17: 13000384-13000384 on Build GRCh38
rs575025761 Chr.17: 13000349-13000349 on Build GRCh38
rs373125478 Chr.17: 13000260-13000260 on Build GRCh38
rs768144008 Chr.17: 13000215-13000215 on Build GRCh38
rs140939761 Chr.17: 13000264-13000264 on Build GRCh38
rs771858858 Chr.17: 13000284-13000284 on Build GRCh38
rs200848500 Chr.17: 13000112-13000112 on Build GRCh38
rs763673924 Chr.17: 13000148-13000148 on Build GRCh38
rs370459292 Chr.17: 13000391-13000391 on Build GRCh38
rs759143366 Chr.17: 13000181-13000181 on Build GRCh38
rs774440711 Chr.17: 13000115-13000115 on Build GRCh38
rs769139794 Chr.17: 13000291-13000292 on Build GRCh38
rs200410915 Chr.17: 13000279-13000280 on Build GRCh38
rs146858023 Chr.17: 13000178-13000178 on Build GRCh38
rs564077271 Chr.17: 13000435-13000435 on Build GRCh38
rs371133505 Chr.17: 13000306-13000306 on Build GRCh38
rs769488454 Chr.17: 13000301-13000301 on Build GRCh38
rs931689766 Chr.17: 13000371-13000371 on Build GRCh38
rs746908751 Chr.17: 13000288-13000288 on Build GRCh38
rs931724133 Chr.17: 13000104-13000104 on Build GRCh38
rs370493014 Chr.17: 13000282-13000283 on Build GRCh38
rs754016032 Chr.17: 13000326-13000326 on Build GRCh38
rs776190388 Chr.17: 13000294-13000294 on Build GRCh38
rs749460308 Chr.17: 13000179-13000179 on Build GRCh38
rs753667852 Chr.17: 13000130-13000130 on Build GRCh38
rs772807173 Chr.17: 13000302-13000302 on Build GRCh38
rs552514338 Chr.17: 13000143-13000143 on Build GRCh38
rs3760316 Chr.17: 13000282-13000282 on Build GRCh38
rs528583908 Chr.17: 13000124-13000124 on Build GRCh38
rs758221167 Chr.17: 13000182-13000184 on Build GRCh38
rs779858810 Chr.17: 13000286-13000286 on Build GRCh38
rs761308485 Chr.17: 13000316-13000316 on Build GRCh38
rs753211422 Chr.17: 13000248-13000248 on Build GRCh38
rs985592295 Chr.17: 13000362-13000362 on Build GRCh38
rs369821953 Chr.17: 13000107-13000107 on Build GRCh38
rs143878748 Chr.17: 13000172-13000172 on Build GRCh38
rs775449483 Chr.17: 13000201-13000201 on Build GRCh38
rs3760317 Chr.17: 13000273-13000273 on Build GRCh38
rs770780181 Chr.17: 13000114-13000114 on Build GRCh38
rs751907634 Chr.17: 13000171-13000171 on Build GRCh38
rs183823186 Chr.17: 13000446-13000446 on Build GRCh38
rs76249486 Chr.17: 13000440-13000440 on Build GRCh38
rs1015826110 Chr.17: 13000135-13000135 on Build GRCh38
rs779782886 Chr.17: 13000210-13000210 on Build GRCh38
rs201830045 Chr.17: 13000281-13000282 on Build GRCh38
rs375798344 Chr.17: 13000337-13000337 on Build GRCh38
rs940364487 Chr.17: 13000412-13000412 on Build GRCh38
rs766674723 Chr.17: 13000163-13000163 on Build GRCh38
rs767481385 Chr.17: 13000118-13000118 on Build GRCh38
rs753447554 Chr.17: 13000152-13000152 on Build GRCh38
rs376568538 Chr.17: 13000447-13000447 on Build GRCh38
rs759926685 Chr.17: 13000230-13000230 on Build GRCh38
rs77972324 Chr.17: 13000190-13000190 on Build GRCh38
rs761281443 Chr.17: 13000295-13000295 on Build GRCh38
rs766134149 Chr.17: 13000123-13000123 on Build GRCh38
rs1029817912 Chr.17: 13000335-13000335 on Build GRCh38
rs759601377 Chr.17: 13000116-13000116 on Build GRCh38
rs911665526 Chr.17: 13000399-13000399 on Build GRCh38
rs760132087 Chr.17: 13000204-13000204 on Build GRCh38
rs761514025 Chr.17: 13000228-13000228 on Build GRCh38

View Full Product Details