Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 14074231-14074738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX10
Entrez Gene ID
1352
Gene Name
COX10, heme A:farnesyltransferase cytochrome c oxidase assembly factor
Gene Aliases
-
UniGene
Hs.462278
Gene Chromosome Location
Chr.17: 14069402-14208679 on Build GRCh38
Transcripts
NM_001303.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3771269 c.100C>T p.H34Y Chr.17: 14074379-14074379 on Build GRCh38
4064057 c.96C>A p.S32S Chr.17: 14074375-14074375 on Build GRCh38
460416 c.120C>G p.L40L Chr.17: 14074399-14074399 on Build GRCh38
2738155 c.78A>T p.R26S Chr.17: 14074357-14074357 on Build GRCh38
2738158 c.127G>A p.V43I Chr.17: 14074406-14074406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976068459 Chr.17: 14074658-14074658 on Build GRCh38
rs769008440 Chr.17: 14074668-14074668 on Build GRCh38
rs149438557 Chr.17: 14074707-14074707 on Build GRCh38
rs139374994 Chr.17: 14074678-14074678 on Build GRCh38
rs1047847173 Chr.17: 14074710-14074710 on Build GRCh38
rs1012589953 Chr.17: 14074404-14074404 on Build GRCh38
rs1052598527 Chr.17: 14074257-14074257 on Build GRCh38
rs990458514 Chr.17: 14074255-14074255 on Build GRCh38
rs753369804 Chr.17: 14074359-14074359 on Build GRCh38
rs143814384 Chr.17: 14074496-14074497 on Build GRCh38
rs561075405 Chr.17: 14074310-14074310 on Build GRCh38
rs1023600507 Chr.17: 14074365-14074365 on Build GRCh38
rs372619044 Chr.17: 14074282-14074282 on Build GRCh38
rs899009773 Chr.17: 14074666-14074666 on Build GRCh38
rs78734063 Chr.17: 14074261-14074261 on Build GRCh38
rs552157137 Chr.17: 14074600-14074600 on Build GRCh38
rs771562895 Chr.17: 14074509-14074512 on Build GRCh38
rs759643676 Chr.17: 14074320-14074320 on Build GRCh38
rs771080222 Chr.17: 14074442-14074442 on Build GRCh38
rs543596288 Chr.17: 14074473-14074473 on Build GRCh38
rs748497811 Chr.17: 14074354-14074356 on Build GRCh38
rs750158907 Chr.17: 14074367-14074367 on Build GRCh38
rs943383703 Chr.17: 14074278-14074278 on Build GRCh38
rs977225626 Chr.17: 14074530-14074530 on Build GRCh38
rs990775725 Chr.17: 14074583-14074583 on Build GRCh38
rs540737897 Chr.17: 14074343-14074343 on Build GRCh38
rs375695653 Chr.17: 14074408-14074408 on Build GRCh38
rs532416333 Chr.17: 14074597-14074597 on Build GRCh38
rs746158624 Chr.17: 14074451-14074451 on Build GRCh38
rs772223730 Chr.17: 14074452-14074452 on Build GRCh38
rs772542140 Chr.17: 14074500-14074500 on Build GRCh38
rs899124768 Chr.17: 14074308-14074308 on Build GRCh38
rs749733605 Chr.17: 14074434-14074434 on Build GRCh38
rs59113375 Chr.17: 14074499-14074500 on Build GRCh38
rs921577205 Chr.17: 14074536-14074536 on Build GRCh38
rs151129445 Chr.17: 14074497-14074498 on Build GRCh38
rs146895519 Chr.17: 14074624-14074624 on Build GRCh38
rs548701564 Chr.17: 14074704-14074704 on Build GRCh38
rs778156711 Chr.17: 14074494-14074495 on Build GRCh38
rs987211130 Chr.17: 14074556-14074556 on Build GRCh38
rs1047324564 Chr.17: 14074341-14074341 on Build GRCh38
rs886052599 Chr.17: 14074402-14074402 on Build GRCh38
rs763311988 Chr.17: 14074578-14074578 on Build GRCh38
rs1032236193 Chr.17: 14074537-14074537 on Build GRCh38
rs757977403 Chr.17: 14074371-14074371 on Build GRCh38
rs764862405 Chr.17: 14074496-14074496 on Build GRCh38
rs756716339 Chr.17: 14074374-14074374 on Build GRCh38
rs112124267 Chr.17: 14074260-14074260 on Build GRCh38
rs778372171 Chr.17: 14074410-14074410 on Build GRCh38
rs746426795 Chr.17: 14074328-14074328 on Build GRCh38
rs748889500 Chr.17: 14074327-14074327 on Build GRCh38
rs778853299 Chr.17: 14074450-14074450 on Build GRCh38
rs16948978 Chr.17: 14074362-14074362 on Build GRCh38
rs894112128 Chr.17: 14074400-14074400 on Build GRCh38
rs761654126 Chr.17: 14074492-14074492 on Build GRCh38
rs763979314 Chr.17: 14074353-14074353 on Build GRCh38
rs369261621 Chr.17: 14074485-14074485 on Build GRCh38
rs141481210 Chr.17: 14074372-14074372 on Build GRCh38
rs957596856 Chr.17: 14074561-14074561 on Build GRCh38
rs775269071 Chr.17: 14074456-14074456 on Build GRCh38
rs781688641 Chr.17: 14074283-14074283 on Build GRCh38
rs200849035 Chr.17: 14074479-14074479 on Build GRCh38
rs868800789 Chr.17: 14074690-14074690 on Build GRCh38
rs770371159 Chr.17: 14074303-14074303 on Build GRCh38
rs923304678 Chr.17: 14074669-14074669 on Build GRCh38
rs113449951 Chr.17: 14074502-14074502 on Build GRCh38
rs770147932 Chr.17: 14074480-14074481 on Build GRCh38
rs761436926 Chr.17: 14074361-14074361 on Build GRCh38
rs372205534 Chr.17: 14074431-14074431 on Build GRCh38
rs748725533 Chr.17: 14074291-14074291 on Build GRCh38
rs778001030 Chr.17: 14074307-14074307 on Build GRCh38
rs775937770 Chr.17: 14074446-14074446 on Build GRCh38
rs114096923 Chr.17: 14074623-14074623 on Build GRCh38
rs375225357 Chr.17: 14074480-14074480 on Build GRCh38
rs201075211 Chr.17: 14074316-14074316 on Build GRCh38

View Full Product Details