Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 14159799-14160303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX10
Entrez Gene ID
1352
Gene Name
COX10, heme A:farnesyltransferase cytochrome c oxidase assembly factor
Gene Aliases
-
UniGene
Hs.462278
Gene Chromosome Location
Chr.17: 14069402-14208679 on Build GRCh38
Transcripts
NM_001303.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4830179 c.647C>G p.S216* Chr.17: 14159899-14159899 on Build GRCh38
5479264 c.626T>G p.F209C Chr.17: 14159878-14159878 on Build GRCh38
5724265 c.682C>T p.R228C Chr.17: 14159934-14159934 on Build GRCh38
3514450 c.629T>C p.F210S Chr.17: 14159881-14159881 on Build GRCh38
6359524 c.625-10C>A p.? Chr.17: 14159867-14159867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044830432 Chr.17: 14159867-14159867 on Build GRCh38
rs771710928 Chr.17: 14160263-14160263 on Build GRCh38
rs947781568 Chr.17: 14160090-14160090 on Build GRCh38
rs987334064 Chr.17: 14160053-14160053 on Build GRCh38
rs1035030899 Chr.17: 14160151-14160152 on Build GRCh38
rs931632877 Chr.17: 14160210-14160210 on Build GRCh38
rs767464568 Chr.17: 14160277-14160277 on Build GRCh38
rs1014866611 Chr.17: 14160064-14160064 on Build GRCh38
rs199609301 Chr.17: 14159927-14159927 on Build GRCh38
rs770099528 Chr.17: 14159833-14159833 on Build GRCh38
rs374962346 Chr.17: 14159941-14159941 on Build GRCh38
rs975868252 Chr.17: 14159998-14159998 on Build GRCh38
rs532822584 Chr.17: 14160261-14160261 on Build GRCh38
rs1003704965 Chr.17: 14159896-14159896 on Build GRCh38
rs778376135 Chr.17: 14159965-14159976 on Build GRCh38
rs759459962 Chr.17: 14159868-14159868 on Build GRCh38
rs7212977 Chr.17: 14160072-14160072 on Build GRCh38
rs752138772 Chr.17: 14159891-14159891 on Build GRCh38
rs907724562 Chr.17: 14160054-14160054 on Build GRCh38
rs151110639 Chr.17: 14160005-14160005 on Build GRCh38
rs767032314 Chr.17: 14159877-14159877 on Build GRCh38
rs892651237 Chr.17: 14160163-14160163 on Build GRCh38
rs1036221187 Chr.17: 14160276-14160276 on Build GRCh38
rs104894556 Chr.17: 14159926-14159926 on Build GRCh38
rs1022692169 Chr.17: 14159964-14159964 on Build GRCh38
rs947066094 Chr.17: 14159939-14159939 on Build GRCh38
rs575464707 Chr.17: 14160227-14160227 on Build GRCh38
rs1022023277 Chr.17: 14160152-14160152 on Build GRCh38
rs193249989 Chr.17: 14160270-14160270 on Build GRCh38
rs111972427 Chr.17: 14160057-14160057 on Build GRCh38
rs1029741338 Chr.17: 14160255-14160255 on Build GRCh38
rs769969857 Chr.17: 14159974-14159974 on Build GRCh38
rs1004678613 Chr.17: 14160044-14160044 on Build GRCh38
rs16949103 Chr.17: 14159975-14159975 on Build GRCh38
rs369503902 Chr.17: 14159995-14159995 on Build GRCh38
rs564891503 Chr.17: 14160279-14160279 on Build GRCh38
rs747531770 Chr.17: 14159945-14159945 on Build GRCh38
rs150243023 Chr.17: 14160088-14160088 on Build GRCh38
rs975686085 Chr.17: 14160183-14160183 on Build GRCh38
rs141116519 Chr.17: 14160017-14160017 on Build GRCh38
rs563873207 Chr.17: 14160260-14160260 on Build GRCh38
rs552965087 Chr.17: 14160091-14160091 on Build GRCh38
rs569926153 Chr.17: 14160039-14160039 on Build GRCh38
rs968292952 Chr.17: 14160178-14160178 on Build GRCh38
rs187898435 Chr.17: 14160191-14160191 on Build GRCh38
rs956889118 Chr.17: 14160152-14160152 on Build GRCh38
rs201826759 Chr.17: 14159958-14159958 on Build GRCh38
rs988356756 Chr.17: 14159931-14159931 on Build GRCh38
rs372357167 Chr.17: 14159957-14159957 on Build GRCh38
rs751576984 Chr.17: 14159860-14159860 on Build GRCh38
rs573109130 Chr.17: 14160034-14160034 on Build GRCh38
rs766053272 Chr.17: 14159849-14159849 on Build GRCh38
rs939508898 Chr.17: 14160144-14160144 on Build GRCh38
rs762128926 Chr.17: 14159828-14159828 on Build GRCh38
rs769217886 Chr.17: 14159946-14159946 on Build GRCh38
rs903626023 Chr.17: 14160161-14160161 on Build GRCh38
rs114521946 Chr.17: 14159934-14159934 on Build GRCh38
rs772901907 Chr.17: 14159841-14159841 on Build GRCh38
rs184369205 Chr.17: 14160139-14160139 on Build GRCh38
rs915395698 Chr.17: 14160178-14160179 on Build GRCh38
rs78809149 Chr.17: 14160077-14160077 on Build GRCh38
rs949692588 Chr.17: 14160126-14160138 on Build GRCh38
rs770820715 Chr.17: 14159999-14159999 on Build GRCh38
rs554598847 Chr.17: 14160179-14160179 on Build GRCh38
rs368124325 Chr.17: 14159846-14159846 on Build GRCh38
rs1011534039 Chr.17: 14160171-14160171 on Build GRCh38

View Full Product Details