Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 14207488-14207992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX10
Entrez Gene ID
1352
Gene Name
COX10, heme A:farnesyltransferase cytochrome c oxidase assembly factor
Gene Aliases
-
UniGene
Hs.462278
Gene Chromosome Location
Chr.17: 14069402-14208679 on Build GRCh38
Transcripts
NM_001303.3

SNP Detail

SNP ID Chromosome Location
rs886052606 Chr.17: 14207804-14207805 on Build GRCh38
rs1013789256 Chr.17: 14207577-14207577 on Build GRCh38
rs1010760230 Chr.17: 14207636-14207636 on Build GRCh38
rs563239177 Chr.17: 14207739-14207739 on Build GRCh38
rs557278632 Chr.17: 14207913-14207913 on Build GRCh38
rs143758001 Chr.17: 14207518-14207518 on Build GRCh38
rs1000537587 Chr.17: 14207916-14207916 on Build GRCh38
rs949020072 Chr.17: 14207836-14207836 on Build GRCh38
rs1022607159 Chr.17: 14207640-14207640 on Build GRCh38
rs907245531 Chr.17: 14207845-14207845 on Build GRCh38
rs1011146047 Chr.17: 14207899-14207899 on Build GRCh38
rs757031938 Chr.17: 14207671-14207671 on Build GRCh38
rs899745912 Chr.17: 14207968-14207968 on Build GRCh38
rs537449689 Chr.17: 14207866-14207866 on Build GRCh38
rs1045703166 Chr.17: 14207841-14207841 on Build GRCh38
rs552101577 Chr.17: 14207856-14207856 on Build GRCh38
rs189358985 Chr.17: 14207858-14207858 on Build GRCh38
rs8076247 Chr.17: 14207510-14207510 on Build GRCh38
rs370121669 Chr.17: 14207888-14207888 on Build GRCh38
rs941546985 Chr.17: 14207512-14207512 on Build GRCh38
rs1030782217 Chr.17: 14207597-14207597 on Build GRCh38
rs573026653 Chr.17: 14207530-14207530 on Build GRCh38
rs965399188 Chr.17: 14207676-14207676 on Build GRCh38
rs893235842 Chr.17: 14207880-14207880 on Build GRCh38
rs907276647 Chr.17: 14207569-14207569 on Build GRCh38
rs1045759022 Chr.17: 14207568-14207568 on Build GRCh38
rs529638594 Chr.17: 14207697-14207697 on Build GRCh38
rs368791319 Chr.17: 14207532-14207532 on Build GRCh38
rs908293617 Chr.17: 14207718-14207718 on Build GRCh38
rs916135773 Chr.17: 14207772-14207772 on Build GRCh38
rs1802618 Chr.17: 14207970-14207970 on Build GRCh38
rs955102373 Chr.17: 14207673-14207673 on Build GRCh38
rs886052605 Chr.17: 14207777-14207778 on Build GRCh38
rs1032138928 Chr.17: 14207956-14207956 on Build GRCh38
rs79156487 Chr.17: 14207793-14207793 on Build GRCh38
rs1000674901 Chr.17: 14207594-14207594 on Build GRCh38
rs966542131 Chr.17: 14207593-14207593 on Build GRCh38
rs974280791 Chr.17: 14207748-14207748 on Build GRCh38
rs934668785 Chr.17: 14207847-14207847 on Build GRCh38
rs543545046 Chr.17: 14207725-14207725 on Build GRCh38
rs896170780 Chr.17: 14207838-14207838 on Build GRCh38
rs1013303600 Chr.17: 14207564-14207564 on Build GRCh38
rs11078234 Chr.17: 14207584-14207584 on Build GRCh38
rs953971887 Chr.17: 14207639-14207639 on Build GRCh38
rs886052603 Chr.17: 14207621-14207621 on Build GRCh38
rs886052608 Chr.17: 14207952-14207952 on Build GRCh38
rs557163590 Chr.17: 14207514-14207514 on Build GRCh38
rs1022154516 Chr.17: 14207915-14207915 on Build GRCh38
rs1057374685 Chr.17: 14207773-14207773 on Build GRCh38
rs982494983 Chr.17: 14207696-14207696 on Build GRCh38
rs886052604 Chr.17: 14207748-14207748 on Build GRCh38
rs1048606415 Chr.17: 14207712-14207712 on Build GRCh38
rs7214082 Chr.17: 14207859-14207859 on Build GRCh38
rs953879063 Chr.17: 14207967-14207967 on Build GRCh38
rs973257196 Chr.17: 14207680-14207680 on Build GRCh38
rs751457905 Chr.17: 14207667-14207667 on Build GRCh38
rs560578836 Chr.17: 14207666-14207666 on Build GRCh38
rs140898783 Chr.17: 14207865-14207865 on Build GRCh38
rs886052607 Chr.17: 14207841-14207841 on Build GRCh38
rs146848128 Chr.17: 14207650-14207650 on Build GRCh38
rs11078233 Chr.17: 14207535-14207535 on Build GRCh38
rs918660319 Chr.17: 14207681-14207681 on Build GRCh38
rs75823746 Chr.17: 14207651-14207651 on Build GRCh38
rs532204640 Chr.17: 14207780-14207780 on Build GRCh38
rs183903455 Chr.17: 14207511-14207511 on Build GRCh38
rs956502633 Chr.17: 14207612-14207612 on Build GRCh38
rs931361027 Chr.17: 14207698-14207698 on Build GRCh38

View Full Product Details