Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15619094-15619605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDRT1
Entrez Gene ID
374286
Gene Name
CMT1A duplicated region transcript 1
Gene Aliases
C170RF1, C17ORF1, C17ORF1A, FBXW10B, FBXW10P1, HREP, SM25H2
UniGene
Hs.548021
Gene Chromosome Location
Chr.17: 15565482-15619704 on Build GRCh38
Transcripts
NM_001282540.1 NM_006382.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7234988 c.27G>A p.K9K Chr.17: 15619486-15619486 on Build GRCh38
5851871 c.70C>T p.P24S Chr.17: 15619443-15619443 on Build GRCh38
5882813 c.279C>A p.D93E Chr.17: 15619234-15619234 on Build GRCh38
3771272 c.44G>A p.R15H Chr.17: 15619469-15619469 on Build GRCh38
7119063 c.311C>T p.S104F Chr.17: 15619202-15619202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775972046 Chr.17: 15619274-15619274 on Build GRCh38
rs368697298 Chr.17: 15619325-15619325 on Build GRCh38
rs564630646 Chr.17: 15619549-15619549 on Build GRCh38
rs1014002360 Chr.17: 15619284-15619284 on Build GRCh38
rs185295167 Chr.17: 15619453-15619453 on Build GRCh38
rs759668077 Chr.17: 15619546-15619546 on Build GRCh38
rs764315653 Chr.17: 15619183-15619183 on Build GRCh38
rs754248281 Chr.17: 15619302-15619302 on Build GRCh38
rs764146099 Chr.17: 15619553-15619553 on Build GRCh38
rs369747385 Chr.17: 15619545-15619545 on Build GRCh38
rs200834889 Chr.17: 15619193-15619193 on Build GRCh38
rs758193164 Chr.17: 15619197-15619197 on Build GRCh38
rs372982872 Chr.17: 15619203-15619203 on Build GRCh38
rs181555831 Chr.17: 15619247-15619247 on Build GRCh38
rs188826833 Chr.17: 15619141-15619141 on Build GRCh38
rs1055572790 Chr.17: 15619277-15619277 on Build GRCh38
rs376457568 Chr.17: 15619514-15619514 on Build GRCh38
rs752735042 Chr.17: 15619497-15619497 on Build GRCh38
rs779867096 Chr.17: 15619350-15619350 on Build GRCh38
rs781206380 Chr.17: 15619392-15619392 on Build GRCh38
rs757983860 Chr.17: 15619341-15619342 on Build GRCh38
rs373629485 Chr.17: 15619280-15619280 on Build GRCh38
rs762981756 Chr.17: 15619467-15619467 on Build GRCh38
rs751725401 Chr.17: 15619478-15619478 on Build GRCh38
rs755895222 Chr.17: 15619552-15619552 on Build GRCh38
rs764691195 Chr.17: 15619298-15619298 on Build GRCh38
rs76061382 Chr.17: 15619137-15619137 on Build GRCh38
rs967202688 Chr.17: 15619331-15619331 on Build GRCh38
rs748440590 Chr.17: 15619395-15619397 on Build GRCh38
rs747717927 Chr.17: 15619242-15619242 on Build GRCh38
rs370245211 Chr.17: 15619241-15619241 on Build GRCh38
rs770467424 Chr.17: 15619476-15619476 on Build GRCh38
rs768212627 Chr.17: 15619119-15619119 on Build GRCh38
rs373696841 Chr.17: 15619531-15619531 on Build GRCh38
rs751389528 Chr.17: 15619351-15619351 on Build GRCh38
rs772693457 Chr.17: 15619139-15619139 on Build GRCh38
rs777379873 Chr.17: 15619507-15619507 on Build GRCh38
rs778388519 Chr.17: 15619514-15619514 on Build GRCh38
rs202021798 Chr.17: 15619470-15619470 on Build GRCh38
rs747972847 Chr.17: 15619400-15619400 on Build GRCh38
rs551945332 Chr.17: 15619562-15619562 on Build GRCh38
rs778560362 Chr.17: 15619557-15619557 on Build GRCh38
rs754722107 Chr.17: 15619381-15619381 on Build GRCh38
rs377174530 Chr.17: 15619207-15619207 on Build GRCh38
rs377296198 Chr.17: 15619169-15619169 on Build GRCh38
rs772926254 Chr.17: 15619249-15619249 on Build GRCh38
rs541937180 Chr.17: 15619523-15619523 on Build GRCh38
rs747000595 Chr.17: 15619219-15619219 on Build GRCh38
rs769131182 Chr.17: 15619520-15619520 on Build GRCh38
rs762686486 Chr.17: 15619264-15619264 on Build GRCh38
rs535994502 Chr.17: 15619237-15619237 on Build GRCh38
rs754811270 Chr.17: 15619481-15619481 on Build GRCh38
rs761413180 Chr.17: 15619136-15619136 on Build GRCh38
rs748835732 Chr.17: 15619508-15619508 on Build GRCh38
rs375199803 Chr.17: 15619433-15619433 on Build GRCh38
rs777130119 Chr.17: 15619419-15619419 on Build GRCh38
rs369929383 Chr.17: 15619519-15619519 on Build GRCh38
rs753768870 Chr.17: 15619555-15619555 on Build GRCh38
rs150696081 Chr.17: 15619541-15619541 on Build GRCh38
rs372428411 Chr.17: 15619449-15619449 on Build GRCh38
rs753320119 Chr.17: 15619155-15619155 on Build GRCh38
rs756830802 Chr.17: 15619556-15619556 on Build GRCh38
rs911276568 Chr.17: 15619128-15619134 on Build GRCh38
rs765162712 Chr.17: 15619318-15619318 on Build GRCh38
rs767215913 Chr.17: 15619495-15619495 on Build GRCh38
rs772618342 Chr.17: 15619265-15619265 on Build GRCh38
rs201659993 Chr.17: 15619434-15619434 on Build GRCh38
rs778719597 Chr.17: 15619512-15619512 on Build GRCh38
rs552589315 Chr.17: 15619440-15619440 on Build GRCh38
rs922811114 Chr.17: 15619492-15619492 on Build GRCh38
rs761966306 Chr.17: 15619305-15619305 on Build GRCh38
rs559969871 Chr.17: 15619236-15619236 on Build GRCh38
rs200161785 Chr.17: 15619446-15619446 on Build GRCh38
rs776837705 Chr.17: 15619522-15619522 on Build GRCh38
rs998786666 Chr.17: 15619399-15619399 on Build GRCh38
rs750553231 Chr.17: 15619194-15619194 on Build GRCh38
rs781663759 Chr.17: 15619355-15619361 on Build GRCh38
rs375341389 Chr.17: 15619452-15619452 on Build GRCh38
rs755979035 Chr.17: 15619502-15619502 on Build GRCh38
rs201445711 Chr.17: 15619445-15619445 on Build GRCh38
rs750026145 Chr.17: 15619197-15619199 on Build GRCh38
rs747398970 Chr.17: 15619438-15619438 on Build GRCh38
rs773663032 Chr.17: 15619537-15619537 on Build GRCh38
rs369804816 Chr.17: 15619187-15619187 on Build GRCh38
rs776185302 Chr.17: 15619135-15619135 on Build GRCh38
rs767930659 Chr.17: 15619147-15619147 on Build GRCh38
rs767391044 Chr.17: 15619547-15619547 on Build GRCh38
rs370886190 Chr.17: 15619415-15619415 on Build GRCh38
rs756777625 Chr.17: 15619511-15619511 on Build GRCh38
rs750702564 Chr.17: 15619321-15619321 on Build GRCh38
rs763036918 Chr.17: 15619539-15619539 on Build GRCh38
rs374209015 Chr.17: 15619192-15619192 on Build GRCh38
rs748993346 Chr.17: 15619421-15619421 on Build GRCh38
rs1005603398 Chr.17: 15619268-15619268 on Build GRCh38

View Full Product Details