Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15945467-15945969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA2B
Entrez Gene ID
136
Gene Name
adenosine A2b receptor
Gene Aliases
ADORA2
UniGene
Hs.167046
Gene Chromosome Location
Chr.17: 15927781-15975896 on Build GRCh38
Transcripts
NM_000676.2 XM_011523659.2 XM_011523661.2 XM_017024197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1749877 c.291G>T p.L97L Chr.17: 15945539-15945539 on Build GRCh38
6334941 c.255G>T p.V85V Chr.17: 15945503-15945503 on Build GRCh38
5956180 c.277A>G p.I93V Chr.17: 15945525-15945525 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749186713 Chr.17: 15945523-15945523 on Build GRCh38
rs977154208 Chr.17: 15945724-15945724 on Build GRCh38
rs983142385 Chr.17: 15945711-15945711 on Build GRCh38
rs768179791 Chr.17: 15945937-15945937 on Build GRCh38
rs867677913 Chr.17: 15945727-15945727 on Build GRCh38
rs1050751022 Chr.17: 15945573-15945573 on Build GRCh38
rs948955695 Chr.17: 15945561-15945561 on Build GRCh38
rs950352905 Chr.17: 15945706-15945706 on Build GRCh38
rs964585556 Chr.17: 15945687-15945687 on Build GRCh38
rs1011681846 Chr.17: 15945648-15945648 on Build GRCh38
rs2041456 Chr.17: 15945743-15945743 on Build GRCh38
rs769414671 Chr.17: 15945873-15945873 on Build GRCh38
rs550487634 Chr.17: 15945657-15945657 on Build GRCh38
rs775307627 Chr.17: 15945544-15945544 on Build GRCh38
rs947422924 Chr.17: 15945813-15945813 on Build GRCh38
rs34680670 Chr.17: 15945832-15945832 on Build GRCh38
rs900328804 Chr.17: 15945827-15945827 on Build GRCh38
rs925661188 Chr.17: 15945916-15945916 on Build GRCh38
rs113146623 Chr.17: 15945913-15945913 on Build GRCh38
rs934642855 Chr.17: 15945786-15945786 on Build GRCh38
rs910216308 Chr.17: 15945941-15945941 on Build GRCh38
rs2041457 Chr.17: 15945892-15945892 on Build GRCh38
rs201752442 Chr.17: 15945595-15945595 on Build GRCh38
rs897650949 Chr.17: 15945796-15945798 on Build GRCh38
rs998846773 Chr.17: 15945625-15945625 on Build GRCh38
rs530853855 Chr.17: 15945630-15945630 on Build GRCh38
rs748757348 Chr.17: 15945918-15945918 on Build GRCh38
rs927510577 Chr.17: 15945686-15945687 on Build GRCh38
rs746301942 Chr.17: 15945838-15945838 on Build GRCh38
rs772704634 Chr.17: 15945782-15945787 on Build GRCh38
rs369345398 Chr.17: 15945530-15945530 on Build GRCh38
rs1034612039 Chr.17: 15945855-15945855 on Build GRCh38
rs560614929 Chr.17: 15945488-15945488 on Build GRCh38
rs759087654 Chr.17: 15945541-15945541 on Build GRCh38
rs1004822821 Chr.17: 15945907-15945907 on Build GRCh38
rs963435975 Chr.17: 15945736-15945736 on Build GRCh38
rs1039118457 Chr.17: 15945823-15945823 on Build GRCh38
rs986160763 Chr.17: 15945928-15945928 on Build GRCh38
rs767329319 Chr.17: 15945543-15945543 on Build GRCh38
rs994207986 Chr.17: 15945639-15945639 on Build GRCh38
rs570294747 Chr.17: 15945704-15945704 on Build GRCh38
rs781359164 Chr.17: 15945501-15945501 on Build GRCh38
rs997387725 Chr.17: 15945846-15945846 on Build GRCh38
rs529225004 Chr.17: 15945492-15945492 on Build GRCh38
rs553305531 Chr.17: 15945551-15945551 on Build GRCh38
rs73274394 Chr.17: 15945597-15945597 on Build GRCh38
rs34502015 Chr.17: 15945807-15945807 on Build GRCh38
rs1017829077 Chr.17: 15945672-15945672 on Build GRCh38
rs140304774 Chr.17: 15945534-15945534 on Build GRCh38
rs756262039 Chr.17: 15945619-15945619 on Build GRCh38
rs546288326 Chr.17: 15945757-15945757 on Build GRCh38
rs763425220 Chr.17: 15945577-15945577 on Build GRCh38
rs760366487 Chr.17: 15945554-15945554 on Build GRCh38
rs766900823 Chr.17: 15945591-15945591 on Build GRCh38
rs997397593 Chr.17: 15945691-15945691 on Build GRCh38
rs948892982 Chr.17: 15945760-15945760 on Build GRCh38
rs993368239 Chr.17: 15945700-15945700 on Build GRCh38
rs780014133 Chr.17: 15945489-15945489 on Build GRCh38
rs893147755 Chr.17: 15945812-15945812 on Build GRCh38
rs777442017 Chr.17: 15945626-15945626 on Build GRCh38
rs959255469 Chr.17: 15945879-15945879 on Build GRCh38
rs150319645 Chr.17: 15945560-15945560 on Build GRCh38
rs965393659 Chr.17: 15945637-15945639 on Build GRCh38
rs958912158 Chr.17: 15945699-15945699 on Build GRCh38
rs763861196 Chr.17: 15945667-15945667 on Build GRCh38
rs146701592 Chr.17: 15945518-15945518 on Build GRCh38
rs1024505491 Chr.17: 15945893-15945893 on Build GRCh38
rs750745874 Chr.17: 15945568-15945568 on Build GRCh38
rs368963660 Chr.17: 15945777-15945777 on Build GRCh38
rs935639259 Chr.17: 15945734-15945734 on Build GRCh38
rs752946237 Chr.17: 15945618-15945618 on Build GRCh38
rs893235106 Chr.17: 15945647-15945647 on Build GRCh38
rs747095169 Chr.17: 15945491-15945491 on Build GRCh38
rs566568199 Chr.17: 15945769-15945769 on Build GRCh38
rs769597741 Chr.17: 15945512-15945512 on Build GRCh38

View Full Product Details